Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45920856-45921349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT7B
Entrez Gene ID
7477
Gene Name
Wnt family member 7B
Gene Aliases
-
UniGene
Hs.512714
Gene Chromosome Location
Chr.22: 45920362-45977128 on Build GRCh38
Transcripts
NM_058238.2 XM_011530366.1

SNP Detail

SNP ID Chromosome Location
rs576656136 Chr.22: 45920953-45920953 on Build GRCh38
rs556124340 Chr.22: 45921321-45921321 on Build GRCh38
rs11544453 Chr.22: 45920983-45920983 on Build GRCh38
rs112416708 Chr.22: 45920999-45920999 on Build GRCh38
rs531622291 Chr.22: 45921142-45921142 on Build GRCh38
rs548310195 Chr.22: 45921214-45921214 on Build GRCh38
rs917283307 Chr.22: 45921262-45921262 on Build GRCh38
rs979124022 Chr.22: 45920985-45920985 on Build GRCh38
rs898805975 Chr.22: 45921092-45921092 on Build GRCh38
rs755910829 Chr.22: 45921154-45921154 on Build GRCh38
rs1047211540 Chr.22: 45921065-45921065 on Build GRCh38
rs950230618 Chr.22: 45920885-45920885 on Build GRCh38
rs1047278286 Chr.22: 45920902-45920902 on Build GRCh38
rs1028709175 Chr.22: 45921134-45921134 on Build GRCh38
rs1019178542 Chr.22: 45921194-45921194 on Build GRCh38
rs1054803676 Chr.22: 45921107-45921107 on Build GRCh38
rs555532900 Chr.22: 45920972-45920972 on Build GRCh38
rs186742442 Chr.22: 45921103-45921103 on Build GRCh38
rs758099832 Chr.22: 45920888-45920888 on Build GRCh38
rs952217792 Chr.22: 45921223-45921223 on Build GRCh38
rs368125342 Chr.22: 45921225-45921225 on Build GRCh38
rs1006252955 Chr.22: 45920917-45920917 on Build GRCh38
rs887314114 Chr.22: 45921068-45921068 on Build GRCh38
rs188844737 Chr.22: 45921220-45921220 on Build GRCh38
rs999714174 Chr.22: 45921193-45921193 on Build GRCh38
rs961170799 Chr.22: 45921245-45921245 on Build GRCh38
rs895973102 Chr.22: 45920884-45920884 on Build GRCh38
rs570483209 Chr.22: 45920911-45920911 on Build GRCh38
rs894887606 Chr.22: 45921120-45921120 on Build GRCh38
rs1008684640 Chr.22: 45921163-45921163 on Build GRCh38
rs75848132 Chr.22: 45921192-45921192 on Build GRCh38
rs1020177808 Chr.22: 45920973-45920973 on Build GRCh38
rs984919474 Chr.22: 45921235-45921235 on Build GRCh38
rs996541443 Chr.22: 45921216-45921216 on Build GRCh38
rs1022502698 Chr.22: 45920969-45920969 on Build GRCh38
rs958709185 Chr.22: 45921226-45921226 on Build GRCh38
rs570294261 Chr.22: 45921326-45921326 on Build GRCh38
rs570548774 Chr.22: 45921261-45921261 on Build GRCh38
rs995929056 Chr.22: 45920931-45920931 on Build GRCh38
rs966290688 Chr.22: 45921195-45921195 on Build GRCh38
rs967357051 Chr.22: 45920970-45920970 on Build GRCh38
rs79828342 Chr.22: 45921310-45921310 on Build GRCh38
rs536156921 Chr.22: 45921329-45921329 on Build GRCh38
rs939977729 Chr.22: 45920921-45920921 on Build GRCh38
rs372229104 Chr.22: 45920934-45920934 on Build GRCh38
rs113866700 Chr.22: 45921052-45921052 on Build GRCh38
rs191549873 Chr.22: 45921141-45921141 on Build GRCh38
rs925955937 Chr.22: 45920996-45920996 on Build GRCh38
rs1039054638 Chr.22: 45921079-45921079 on Build GRCh38
rs978369014 Chr.22: 45920981-45920981 on Build GRCh38
rs984358966 Chr.22: 45921059-45921059 on Build GRCh38
rs556620582 Chr.22: 45920930-45920930 on Build GRCh38
rs184162816 Chr.22: 45921209-45921209 on Build GRCh38
rs917348767 Chr.22: 45921047-45921047 on Build GRCh38
rs75343830 Chr.22: 45920925-45920925 on Build GRCh38
rs765867790 Chr.22: 45921032-45921032 on Build GRCh38
rs542105873 Chr.22: 45920961-45920961 on Build GRCh38
rs987306508 Chr.22: 45920962-45920962 on Build GRCh38
rs1028761201 Chr.22: 45920940-45920940 on Build GRCh38
rs941604197 Chr.22: 45921077-45921077 on Build GRCh38
rs761847840 Chr.22: 45920920-45920920 on Build GRCh38
rs909687732 Chr.22: 45921236-45921236 on Build GRCh38
rs1044272202 Chr.22: 45921164-45921164 on Build GRCh38
rs547532882 Chr.22: 45921250-45921250 on Build GRCh38
rs564206157 Chr.22: 45921026-45921026 on Build GRCh38
rs991504013 Chr.22: 45921041-45921041 on Build GRCh38

View Full Product Details