Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10146542-10147011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VHL
Entrez Gene ID
7428
Gene Name
von Hippel-Lindau tumor suppressor
Gene Aliases
HRCA1, RCA1, VHL1, pVHL
UniGene
Hs.517792
Gene Chromosome Location
Chr.3: 10141635-10153670 on Build GRCh38
Transcripts
NM_000551.3 NM_198156.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7253800 c.403_404insATTT p.L135fs*10 Chr.3: 10146576-10146577 on Build GRCh38
308465 c.401_404delAATT p.E134fs*24 Chr.3: 10146574-10146577 on Build GRCh38
17896 c.378_379insT p.G127fs*5 Chr.3: 10146551-10146552 on Build GRCh38
33993 c.404T>A p.L135* Chr.3: 10146577-10146577 on Build GRCh38
5075579 c.438_439insT p.I147fs*27 Chr.3: 10146611-10146612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869025654 Chr.3: 10146617-10146617 on Build GRCh38
rs5030814 Chr.3: 10146638-10146638 on Build GRCh38
rs587780732 Chr.3: 10146589-10146589 on Build GRCh38
rs750211277 Chr.3: 10146670-10146670 on Build GRCh38
rs869025651 Chr.3: 10146604-10146604 on Build GRCh38
rs869025657 Chr.3: 10146637-10146637 on Build GRCh38
rs369465430 Chr.3: 10146593-10146593 on Build GRCh38
rs879253989 Chr.3: 10146640-10146640 on Build GRCh38
rs119103278 Chr.3: 10146578-10146578 on Build GRCh38
rs943062970 Chr.3: 10146895-10146895 on Build GRCh38
rs869025652 Chr.3: 10146608-10146609 on Build GRCh38
rs112997247 Chr.3: 10146848-10146848 on Build GRCh38
rs554826167 Chr.3: 10146826-10146826 on Build GRCh38
rs1034783527 Chr.3: 10146981-10146981 on Build GRCh38
rs193922611 Chr.3: 10146631-10146631 on Build GRCh38
rs749704215 Chr.3: 10146606-10146606 on Build GRCh38
rs879131097 Chr.3: 10146810-10146810 on Build GRCh38
rs876659313 Chr.3: 10146624-10146624 on Build GRCh38
rs758157229 Chr.3: 10146676-10146676 on Build GRCh38
rs575360985 Chr.3: 10146798-10146798 on Build GRCh38
rs760184234 Chr.3: 10146622-10146622 on Build GRCh38
rs869025646 Chr.3: 10146575-10146575 on Build GRCh38
rs869025650 Chr.3: 10146603-10146603 on Build GRCh38
rs546327119 Chr.3: 10146731-10146731 on Build GRCh38
rs892067264 Chr.3: 10146850-10146850 on Build GRCh38
rs587780077 Chr.3: 10146618-10146618 on Build GRCh38
rs763525739 Chr.3: 10146623-10146623 on Build GRCh38
rs878937319 Chr.3: 10146609-10146609 on Build GRCh38
rs869025648 Chr.3: 10146587-10146587 on Build GRCh38
rs1678607 Chr.3: 10146744-10146744 on Build GRCh38
rs1033284192 Chr.3: 10146977-10146977 on Build GRCh38
rs864622313 Chr.3: 10146607-10146607 on Build GRCh38
rs1042724883 Chr.3: 10146964-10146964 on Build GRCh38
rs929479307 Chr.3: 10146845-10146845 on Build GRCh38
rs775038762 Chr.3: 10146617-10146617 on Build GRCh38
rs545480782 Chr.3: 10146921-10146921 on Build GRCh38
rs34661876 Chr.3: 10146679-10146679 on Build GRCh38
rs5030833 Chr.3: 10146580-10146580 on Build GRCh38
rs878854125 Chr.3: 10146581-10146581 on Build GRCh38
rs773556807 Chr.3: 10146602-10146602 on Build GRCh38
rs946385444 Chr.3: 10146960-10146960 on Build GRCh38
rs959045706 Chr.3: 10146982-10146982 on Build GRCh38
rs869025653 Chr.3: 10146617-10146618 on Build GRCh38
rs991457989 Chr.3: 10146716-10146716 on Build GRCh38
rs201482330 Chr.3: 10146575-10146575 on Build GRCh38
rs909295775 Chr.3: 10146760-10146760 on Build GRCh38
rs143594610 Chr.3: 10146599-10146599 on Build GRCh38
rs869025658 Chr.3: 10146673-10146675 on Build GRCh38
rs869025656 Chr.3: 10146627-10146653 on Build GRCh38
rs1003364563 Chr.3: 10146641-10146641 on Build GRCh38
rs536176038 Chr.3: 10146806-10146806 on Build GRCh38
rs766571541 Chr.3: 10146677-10146677 on Build GRCh38
rs143844761 Chr.3: 10146652-10146652 on Build GRCh38
rs200928782 Chr.3: 10146674-10146674 on Build GRCh38
rs893514720 Chr.3: 10146930-10146930 on Build GRCh38
rs1052028970 Chr.3: 10146829-10146829 on Build GRCh38
rs869025649 Chr.3: 10146592-10146593 on Build GRCh38
rs780178275 Chr.3: 10146586-10146586 on Build GRCh38
rs1057517560 Chr.3: 10146612-10146612 on Build GRCh38
rs534301494 Chr.3: 10146721-10146721 on Build GRCh38
rs755014728 Chr.3: 10146687-10146687 on Build GRCh38
rs794727253 Chr.3: 10146622-10146622 on Build GRCh38
rs781248221 Chr.3: 10146688-10146688 on Build GRCh38
rs1043653911 Chr.3: 10146889-10146889 on Build GRCh38
rs869025655 Chr.3: 10146625-10146625 on Build GRCh38
rs915946923 Chr.3: 10146743-10146743 on Build GRCh38
rs768637170 Chr.3: 10146591-10146591 on Build GRCh38
rs5030813 Chr.3: 10146567-10146567 on Build GRCh38
rs397516442 Chr.3: 10146581-10146581 on Build GRCh38
rs781618785 Chr.3: 10146596-10146596 on Build GRCh38
rs761828544 Chr.3: 10146651-10146651 on Build GRCh38
rs1003790426 Chr.3: 10146978-10146978 on Build GRCh38
rs869025659 Chr.3: 10146636-10146636 on Build GRCh38
rs35334687 Chr.3: 10146853-10146853 on Build GRCh38
rs555191817 Chr.3: 10146805-10146805 on Build GRCh38
rs397516443 Chr.3: 10146638-10146638 on Build GRCh38
rs578147586 Chr.3: 10146915-10146915 on Build GRCh38
rs1010403360 Chr.3: 10146878-10146878 on Build GRCh38
rs556747524 Chr.3: 10146775-10146775 on Build GRCh38
rs372757722 Chr.3: 10146600-10146600 on Build GRCh38
rs750301189 Chr.3: 10146572-10146572 on Build GRCh38
rs765313182 Chr.3: 10146667-10146667 on Build GRCh38
rs894974805 Chr.3: 10146985-10146985 on Build GRCh38
rs754416002 Chr.3: 10146813-10146813 on Build GRCh38
rs763511251 Chr.3: 10146967-10146967 on Build GRCh38
rs541428506 Chr.3: 10146856-10146856 on Build GRCh38
rs5030834 Chr.3: 10146644-10146644 on Build GRCh38
rs140172229 Chr.3: 10146810-10146810 on Build GRCh38
rs147014327 Chr.3: 10146766-10146766 on Build GRCh38
rs143659897 Chr.3: 10146892-10146892 on Build GRCh38
rs771727849 Chr.3: 10146608-10146608 on Build GRCh38

View Full Product Details