Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12152976-12153482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYN2
Entrez Gene ID
6854
Gene Name
synapsin II
Gene Aliases
SYNII
UniGene
Hs.445503
Gene Chromosome Location
Chr.3: 12004360-12192032 on Build GRCh38
Transcripts
NM_003178.5 NM_133625.4 XM_006713311.3 XM_006713312.3 XM_006713313.2 NM_003256.3

Gene Symbol
TIMP4
Entrez Gene ID
7079
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
-
UniGene
Hs.591665
Gene Chromosome Location
Chr.3: 12153068-12159351 on Build GRCh38
Transcripts
NM_003178.5 NM_133625.4 XM_006713311.3 XM_006713312.3 XM_006713313.2 NM_003256.3

SNP Detail

SNP ID Chromosome Location
rs1014787876 Chr.3: 12153406-12153421 on Build GRCh38
rs1026838803 Chr.3: 12153036-12153036 on Build GRCh38
rs56180253 Chr.3: 12153000-12153000 on Build GRCh38
rs549809998 Chr.3: 12153231-12153231 on Build GRCh38
rs182058887 Chr.3: 12153318-12153318 on Build GRCh38
rs886178050 Chr.3: 12153276-12153276 on Build GRCh38
rs948880133 Chr.3: 12153155-12153155 on Build GRCh38
rs962937858 Chr.3: 12153074-12153074 on Build GRCh38
rs996666219 Chr.3: 12153346-12153346 on Build GRCh38
rs890057543 Chr.3: 12153416-12153416 on Build GRCh38
rs570283785 Chr.3: 12153154-12153154 on Build GRCh38
rs993499141 Chr.3: 12153017-12153017 on Build GRCh38
rs17035943 Chr.3: 12153084-12153084 on Build GRCh38
rs560426325 Chr.3: 12153123-12153123 on Build GRCh38
rs959970070 Chr.3: 12153424-12153424 on Build GRCh38
rs1019461752 Chr.3: 12153055-12153055 on Build GRCh38
rs562742388 Chr.3: 12153060-12153060 on Build GRCh38
rs977711159 Chr.3: 12153408-12153408 on Build GRCh38
rs949050753 Chr.3: 12153208-12153208 on Build GRCh38
rs1009023748 Chr.3: 12153222-12153222 on Build GRCh38
rs1009363307 Chr.3: 12153049-12153049 on Build GRCh38
rs995698207 Chr.3: 12153402-12153402 on Build GRCh38
rs966760874 Chr.3: 12153452-12153452 on Build GRCh38
rs977135623 Chr.3: 12153108-12153108 on Build GRCh38
rs890413137 Chr.3: 12153214-12153214 on Build GRCh38
rs17035945 Chr.3: 12153128-12153128 on Build GRCh38
rs871222 Chr.3: 12153164-12153164 on Build GRCh38
rs940446737 Chr.3: 12153328-12153328 on Build GRCh38
rs1028317203 Chr.3: 12153089-12153089 on Build GRCh38
rs548280901 Chr.3: 12153100-12153100 on Build GRCh38
rs963904106 Chr.3: 12153344-12153344 on Build GRCh38
rs1017694767 Chr.3: 12153248-12153248 on Build GRCh38
rs924245891 Chr.3: 12153127-12153127 on Build GRCh38
rs1037945198 Chr.3: 12153156-12153156 on Build GRCh38
rs551969712 Chr.3: 12153137-12153137 on Build GRCh38
rs968679742 Chr.3: 12153359-12153359 on Build GRCh38
rs1060661 Chr.3: 12153347-12153347 on Build GRCh38
rs915647455 Chr.3: 12153426-12153426 on Build GRCh38
rs992568801 Chr.3: 12153425-12153425 on Build GRCh38
rs533502122 Chr.3: 12153366-12153366 on Build GRCh38
rs1021447133 Chr.3: 12153357-12153357 on Build GRCh38
rs753534619 Chr.3: 12153391-12153391 on Build GRCh38
rs994132448 Chr.3: 12153180-12153180 on Build GRCh38
rs111600647 Chr.3: 12153316-12153316 on Build GRCh38
rs899205024 Chr.3: 12153002-12153002 on Build GRCh38
rs924301258 Chr.3: 12153420-12153420 on Build GRCh38
rs1056766225 Chr.3: 12153136-12153136 on Build GRCh38
rs531312010 Chr.3: 12153169-12153169 on Build GRCh38
rs916501680 Chr.3: 12153192-12153192 on Build GRCh38
rs954836175 Chr.3: 12153041-12153041 on Build GRCh38
rs571957033 Chr.3: 12153184-12153184 on Build GRCh38
rs535482539 Chr.3: 12153429-12153429 on Build GRCh38
rs975362773 Chr.3: 12153067-12153067 on Build GRCh38

View Full Product Details