Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 13500634-13501135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC11
Entrez Gene ID
79885
Gene Name
histone deacetylase 11
Gene Aliases
HD11
UniGene
Hs.744132
Gene Chromosome Location
Chr.3: 13480171-13506424 on Build GRCh38
Transcripts
NM_001136041.2 NM_024827.3 XM_005265491.1 XM_005265492.1 XM_006713338.1 XM_011534131.1 XM_011534132.2 XM_011534135.1 XM_017007234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1566255 c.468G>A p.A156A Chr.3: 13500768-13500768 on Build GRCh38
119031 c.489+1G>C p.? Chr.3: 13500790-13500790 on Build GRCh38
5708483 c.436A>G p.S146G Chr.3: 13500736-13500736 on Build GRCh38
240082 c.448G>A p.G150R Chr.3: 13500748-13500748 on Build GRCh38
1130335 c.447C>T p.G149G Chr.3: 13500747-13500747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991116434 Chr.3: 13500934-13500934 on Build GRCh38
rs928323144 Chr.3: 13500684-13500684 on Build GRCh38
rs374085819 Chr.3: 13500790-13500790 on Build GRCh38
rs963667767 Chr.3: 13500868-13500868 on Build GRCh38
rs775554632 Chr.3: 13500743-13500743 on Build GRCh38
rs998965154 Chr.3: 13500810-13500810 on Build GRCh38
rs553483978 Chr.3: 13501059-13501059 on Build GRCh38
rs760095628 Chr.3: 13501082-13501082 on Build GRCh38
rs140627816 Chr.3: 13500768-13500768 on Build GRCh38
rs770859645 Chr.3: 13500735-13500735 on Build GRCh38
rs752248385 Chr.3: 13500786-13500786 on Build GRCh38
rs774342289 Chr.3: 13500738-13500738 on Build GRCh38
rs115992302 Chr.3: 13500826-13500826 on Build GRCh38
rs758668290 Chr.3: 13500782-13500782 on Build GRCh38
rs555565718 Chr.3: 13501026-13501026 on Build GRCh38
rs766875680 Chr.3: 13500747-13500747 on Build GRCh38
rs374445837 Chr.3: 13500835-13500835 on Build GRCh38
rs576811489 Chr.3: 13501096-13501096 on Build GRCh38
rs995466655 Chr.3: 13500882-13500882 on Build GRCh38
rs929752030 Chr.3: 13500657-13500657 on Build GRCh38
rs982467901 Chr.3: 13500676-13500676 on Build GRCh38
rs757651652 Chr.3: 13500673-13500673 on Build GRCh38
rs535683038 Chr.3: 13501002-13501002 on Build GRCh38
rs765562088 Chr.3: 13501004-13501004 on Build GRCh38
rs1026496091 Chr.3: 13500884-13500884 on Build GRCh38
rs748790709 Chr.3: 13500823-13500823 on Build GRCh38
rs908828410 Chr.3: 13500937-13500937 on Build GRCh38
rs377504911 Chr.3: 13500693-13500693 on Build GRCh38
rs767824703 Chr.3: 13500742-13500742 on Build GRCh38
rs754725710 Chr.3: 13500694-13500694 on Build GRCh38
rs142239259 Chr.3: 13500902-13500902 on Build GRCh38
rs754307930 Chr.3: 13500758-13500758 on Build GRCh38
rs982520239 Chr.3: 13500888-13500888 on Build GRCh38
rs113257876 Chr.3: 13501056-13501056 on Build GRCh38
rs778688152 Chr.3: 13500832-13500832 on Build GRCh38
rs773310211 Chr.3: 13500713-13500713 on Build GRCh38
rs765720762 Chr.3: 13500777-13500777 on Build GRCh38
rs567772750 Chr.3: 13501037-13501037 on Build GRCh38
rs147179913 Chr.3: 13500680-13500680 on Build GRCh38
rs959737166 Chr.3: 13500919-13500919 on Build GRCh38
rs749063001 Chr.3: 13500662-13500663 on Build GRCh38
rs1051881801 Chr.3: 13501070-13501070 on Build GRCh38
rs764120706 Chr.3: 13500767-13500767 on Build GRCh38
rs199861928 Chr.3: 13500784-13500784 on Build GRCh38
rs979258305 Chr.3: 13501071-13501071 on Build GRCh38
rs950924724 Chr.3: 13500885-13500885 on Build GRCh38
rs754296174 Chr.3: 13500667-13500667 on Build GRCh38
rs762250500 Chr.3: 13500775-13500775 on Build GRCh38
rs751029334 Chr.3: 13500895-13500895 on Build GRCh38
rs779182556 Chr.3: 13500691-13500691 on Build GRCh38
rs1017910812 Chr.3: 13500844-13500844 on Build GRCh38
rs760834867 Chr.3: 13500752-13500752 on Build GRCh38
rs754349464 Chr.3: 13500774-13500774 on Build GRCh38
rs558128004 Chr.3: 13501015-13501015 on Build GRCh38
rs756118717 Chr.3: 13500664-13500664 on Build GRCh38
rs750839749 Chr.3: 13500780-13500780 on Build GRCh38
rs562135997 Chr.3: 13500662-13500662 on Build GRCh38
rs377073273 Chr.3: 13500815-13500815 on Build GRCh38
rs569636082 Chr.3: 13500708-13500708 on Build GRCh38
rs754322872 Chr.3: 13501072-13501072 on Build GRCh38
rs550493153 Chr.3: 13500872-13500872 on Build GRCh38
rs759249442 Chr.3: 13500739-13500739 on Build GRCh38
rs915527290 Chr.3: 13500951-13500951 on Build GRCh38
rs969859467 Chr.3: 13500770-13500770 on Build GRCh38
rs764637837 Chr.3: 13500666-13500666 on Build GRCh38
rs749167601 Chr.3: 13500716-13500716 on Build GRCh38
rs371080961 Chr.3: 13500709-13500709 on Build GRCh38

View Full Product Details