Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14472119-14472613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A6
Entrez Gene ID
6533
Gene Name
solute carrier family 6 member 6
Gene Aliases
TAUT
UniGene
Hs.529488
Gene Chromosome Location
Chr.3: 14402576-14489349 on Build GRCh38
Transcripts
NM_001134367.3 NM_003043.5 NR_103507.2 XM_006713307.2 XM_011534030.2 XM_011534031.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3408310 c.1154C>G p.T385R Chr.3: 14472262-14472262 on Build GRCh38
4136971 c.1209+5G>C p.? Chr.3: 14472322-14472322 on Build GRCh38
3588377 c.1105C>T p.L369L Chr.3: 14472213-14472213 on Build GRCh38
3588378 c.1135A>T p.T379S Chr.3: 14472243-14472243 on Build GRCh38
1419708 c.1169delT p.F392fs*31 Chr.3: 14472277-14472277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2289128 Chr.3: 14472187-14472187 on Build GRCh38
rs141030841 Chr.3: 14472258-14472258 on Build GRCh38
rs986001037 Chr.3: 14472552-14472552 on Build GRCh38
rs985732391 Chr.3: 14472404-14472404 on Build GRCh38
rs2289127 Chr.3: 14472417-14472417 on Build GRCh38
rs1029711757 Chr.3: 14472542-14472542 on Build GRCh38
rs924523171 Chr.3: 14472419-14472419 on Build GRCh38
rs373663427 Chr.3: 14472344-14472344 on Build GRCh38
rs910099204 Chr.3: 14472414-14472414 on Build GRCh38
rs115042746 Chr.3: 14472174-14472174 on Build GRCh38
rs767284946 Chr.3: 14472546-14472546 on Build GRCh38
rs771428962 Chr.3: 14472314-14472314 on Build GRCh38
rs754183618 Chr.3: 14472183-14472183 on Build GRCh38
rs1033304540 Chr.3: 14472246-14472246 on Build GRCh38
rs1054466063 Chr.3: 14472469-14472469 on Build GRCh38
rs1045448423 Chr.3: 14472447-14472447 on Build GRCh38
rs762167686 Chr.3: 14472354-14472354 on Build GRCh38
rs535211121 Chr.3: 14472453-14472453 on Build GRCh38
rs893112722 Chr.3: 14472478-14472478 on Build GRCh38
rs760689457 Chr.3: 14472343-14472343 on Build GRCh38
rs781533049 Chr.3: 14472243-14472243 on Build GRCh38
rs1026969541 Chr.3: 14472233-14472233 on Build GRCh38
rs937153471 Chr.3: 14472477-14472477 on Build GRCh38
rs759115097 Chr.3: 14472296-14472296 on Build GRCh38
rs905708880 Chr.3: 14472466-14472466 on Build GRCh38
rs962449751 Chr.3: 14472377-14472377 on Build GRCh38
rs780631640 Chr.3: 14472195-14472195 on Build GRCh38
rs1029872226 Chr.3: 14472373-14472373 on Build GRCh38
rs750171386 Chr.3: 14472586-14472586 on Build GRCh38
rs760562863 Chr.3: 14472171-14472171 on Build GRCh38
rs762706237 Chr.3: 14472258-14472259 on Build GRCh38
rs765529121 Chr.3: 14472355-14472355 on Build GRCh38
rs758371962 Chr.3: 14472192-14472192 on Build GRCh38
rs761703735 Chr.3: 14472398-14472398 on Build GRCh38
rs200619667 Chr.3: 14472254-14472254 on Build GRCh38
rs921121335 Chr.3: 14472410-14472410 on Build GRCh38
rs907397365 Chr.3: 14472548-14472548 on Build GRCh38
rs780890296 Chr.3: 14472376-14472376 on Build GRCh38
rs1054587556 Chr.3: 14472168-14472168 on Build GRCh38
rs1017086612 Chr.3: 14472558-14472558 on Build GRCh38
rs377659535 Chr.3: 14472155-14472155 on Build GRCh38
rs986070818 Chr.3: 14472153-14472153 on Build GRCh38
rs769012111 Chr.3: 14472223-14472223 on Build GRCh38
rs1044480419 Chr.3: 14472493-14472493 on Build GRCh38
rs903214303 Chr.3: 14472148-14472148 on Build GRCh38
rs1026605748 Chr.3: 14472564-14472564 on Build GRCh38
rs545666681 Chr.3: 14472346-14472346 on Build GRCh38
rs1010138715 Chr.3: 14472489-14472489 on Build GRCh38
rs536018464 Chr.3: 14472363-14472363 on Build GRCh38
rs770545612 Chr.3: 14472255-14472255 on Build GRCh38
rs535310567 Chr.3: 14472418-14472418 on Build GRCh38
rs371071996 Chr.3: 14472381-14472381 on Build GRCh38
rs202084938 Chr.3: 14472169-14472169 on Build GRCh38
rs1054391950 Chr.3: 14472482-14472482 on Build GRCh38
rs1003445108 Chr.3: 14472549-14472549 on Build GRCh38
rs998279938 Chr.3: 14472511-14472511 on Build GRCh38
rs1011296537 Chr.3: 14472259-14472259 on Build GRCh38
rs147546223 Chr.3: 14472488-14472488 on Build GRCh38
rs757539811 Chr.3: 14472185-14472185 on Build GRCh38
rs902510014 Chr.3: 14472494-14472494 on Build GRCh38
rs371141646 Chr.3: 14472186-14472186 on Build GRCh38
rs886698678 Chr.3: 14472570-14472570 on Build GRCh38
rs934652344 Chr.3: 14472160-14472160 on Build GRCh38
rs199794619 Chr.3: 14472212-14472212 on Build GRCh38
rs370209276 Chr.3: 14472321-14472321 on Build GRCh38

View Full Product Details