Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15024025-15024531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2C2
Entrez Gene ID
7182
Gene Name
nuclear receptor subfamily 2 group C member 2
Gene Aliases
TAK1, TR4
UniGene
Hs.555973
Gene Chromosome Location
Chr.3: 14947584-15049279 on Build GRCh38
Transcripts
NM_001291694.1 NM_003298.4 XM_011534058.2 XM_011534059.2 XM_011534061.2 XM_011534063.2 XM_011534064.2 XM_011534065.2 XM_011534066.2 XM_017007118.1 XM_017007119.1 XM_017007120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1039902 c.847G>A p.D283N Chr.3: 15024200-15024200 on Build GRCh38
3003211 c.819T>C p.R273R Chr.3: 15024172-15024172 on Build GRCh38
6729148 c.817C>T p.R273C Chr.3: 15024170-15024170 on Build GRCh38
3974219 c.777T>A p.L259L Chr.3: 15024130-15024130 on Build GRCh38
6272102 c.793G>T p.V265L Chr.3: 15024146-15024146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969945140 Chr.3: 15024504-15024504 on Build GRCh38
rs754363959 Chr.3: 15024099-15024099 on Build GRCh38
rs1039810060 Chr.3: 15024378-15024378 on Build GRCh38
rs1037971697 Chr.3: 15024054-15024054 on Build GRCh38
rs990708723 Chr.3: 15024408-15024409 on Build GRCh38
rs78460072 Chr.3: 15024493-15024493 on Build GRCh38
rs746370304 Chr.3: 15024124-15024126 on Build GRCh38
rs1026405430 Chr.3: 15024344-15024344 on Build GRCh38
rs1039322220 Chr.3: 15024483-15024483 on Build GRCh38
rs551591752 Chr.3: 15024332-15024332 on Build GRCh38
rs769455192 Chr.3: 15024157-15024157 on Build GRCh38
rs928266398 Chr.3: 15024290-15024290 on Build GRCh38
rs941410949 Chr.3: 15024374-15024374 on Build GRCh38
rs994900195 Chr.3: 15024288-15024288 on Build GRCh38
rs950896976 Chr.3: 15024351-15024351 on Build GRCh38
rs890419241 Chr.3: 15024420-15024420 on Build GRCh38
rs770008045 Chr.3: 15024059-15024059 on Build GRCh38
rs1024182360 Chr.3: 15024270-15024270 on Build GRCh38
rs767622562 Chr.3: 15024070-15024070 on Build GRCh38
rs529430729 Chr.3: 15024199-15024199 on Build GRCh38
rs967688347 Chr.3: 15024426-15024426 on Build GRCh38
rs537094994 Chr.3: 15024366-15024366 on Build GRCh38
rs752662965 Chr.3: 15024437-15024437 on Build GRCh38
rs1008251993 Chr.3: 15024268-15024271 on Build GRCh38
rs778822349 Chr.3: 15024399-15024399 on Build GRCh38
rs145356913 Chr.3: 15024198-15024198 on Build GRCh38
rs374396474 Chr.3: 15024075-15024075 on Build GRCh38
rs753882482 Chr.3: 15024253-15024253 on Build GRCh38
rs528516137 Chr.3: 15024284-15024284 on Build GRCh38
rs886506387 Chr.3: 15024502-15024502 on Build GRCh38
rs372044110 Chr.3: 15024227-15024227 on Build GRCh38
rs890712515 Chr.3: 15024205-15024205 on Build GRCh38
rs982740994 Chr.3: 15024283-15024283 on Build GRCh38
rs772579729 Chr.3: 15024222-15024233 on Build GRCh38
rs34660472 Chr.3: 15024174-15024175 on Build GRCh38
rs768409674 Chr.3: 15024146-15024146 on Build GRCh38
rs368645270 Chr.3: 15024461-15024461 on Build GRCh38
rs1003374387 Chr.3: 15024357-15024357 on Build GRCh38
rs752671969 Chr.3: 15024440-15024440 on Build GRCh38
rs931863524 Chr.3: 15024393-15024393 on Build GRCh38
rs374825136 Chr.3: 15024479-15024479 on Build GRCh38
rs959305369 Chr.3: 15024396-15024396 on Build GRCh38
rs982784878 Chr.3: 15024365-15024365 on Build GRCh38
rs753653299 Chr.3: 15024229-15024229 on Build GRCh38
rs923707988 Chr.3: 15024443-15024443 on Build GRCh38
rs757720867 Chr.3: 15024102-15024102 on Build GRCh38
rs946437682 Chr.3: 15024125-15024125 on Build GRCh38
rs779288620 Chr.3: 15024111-15024111 on Build GRCh38
rs759247878 Chr.3: 15024182-15024182 on Build GRCh38
rs149255046 Chr.3: 15024160-15024160 on Build GRCh38
rs374459337 Chr.3: 15024438-15024438 on Build GRCh38
rs914883620 Chr.3: 15024413-15024413 on Build GRCh38
rs200291251 Chr.3: 15024115-15024115 on Build GRCh38
rs189921815 Chr.3: 15024463-15024463 on Build GRCh38
rs571603515 Chr.3: 15024345-15024345 on Build GRCh38
rs76791637 Chr.3: 15024485-15024485 on Build GRCh38
rs1052100581 Chr.3: 15024403-15024403 on Build GRCh38
rs367955638 Chr.3: 15024094-15024094 on Build GRCh38
rs957809071 Chr.3: 15024252-15024252 on Build GRCh38
rs757807162 Chr.3: 15024259-15024259 on Build GRCh38
rs777772620 Chr.3: 15024086-15024086 on Build GRCh38
rs780700639 Chr.3: 15024148-15024148 on Build GRCh38
rs900305813 Chr.3: 15024386-15024386 on Build GRCh38
rs201956313 Chr.3: 15024213-15024213 on Build GRCh38
rs1047798284 Chr.3: 15024499-15024499 on Build GRCh38
rs756153277 Chr.3: 15024077-15024077 on Build GRCh38
rs763027702 Chr.3: 15024170-15024170 on Build GRCh38
rs943573183 Chr.3: 15024480-15024480 on Build GRCh38
rs760341630 Chr.3: 15024161-15024161 on Build GRCh38
rs770972367 Chr.3: 15024171-15024171 on Build GRCh38
rs747764904 Chr.3: 15024149-15024149 on Build GRCh38
rs143442810 Chr.3: 15024450-15024450 on Build GRCh38
rs1030263426 Chr.3: 15024189-15024189 on Build GRCh38
rs145414944 Chr.3: 15024133-15024133 on Build GRCh38
rs41284019 Chr.3: 15024278-15024278 on Build GRCh38
rs140658786 Chr.3: 15024191-15024191 on Build GRCh38

View Full Product Details