Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 96769538-96770049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTBP2
Entrez Gene ID
58155
Gene Name
polypyrimidine tract binding protein 2
Gene Aliases
PTBLP, brPTB, nPTB
UniGene
Hs.596061
Gene Chromosome Location
Chr.1: 96721605-96815049 on Build GRCh38
Transcripts
NM_001300985.1 NM_001300986.1 NM_001300987.1 NM_001300988.1 NM_001300989.1 NM_001300990.1 NM_021190.3 NR_125356.1 NR_125357.1 XM_011541874.2 XM_011541875.2 XM_011541876.2 XM_017001951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806186 c.191G>A p.R64Q Chr.1: 96769778-96769778 on Build GRCh38
7299782 c.170C>A p.P57H Chr.1: 96769757-96769757 on Build GRCh38
4718559 c.248G>T p.G83V Chr.1: 96769835-96769835 on Build GRCh38
284335 c.123T>C p.G41G Chr.1: 96769710-96769710 on Build GRCh38
4718558 c.144_146delAGG p.G49delG Chr.1: 96769731-96769733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764131452 Chr.1: 96769660-96769660 on Build GRCh38
rs145363773 Chr.1: 96769767-96769767 on Build GRCh38
rs546889559 Chr.1: 96769793-96769793 on Build GRCh38
rs762761950 Chr.1: 96769914-96769914 on Build GRCh38
rs893370005 Chr.1: 96769590-96769590 on Build GRCh38
rs976513180 Chr.1: 96770005-96770005 on Build GRCh38
rs201169288 Chr.1: 96769669-96769669 on Build GRCh38
rs771488962 Chr.1: 96769894-96769894 on Build GRCh38
rs969524403 Chr.1: 96769572-96769572 on Build GRCh38
rs1056389185 Chr.1: 96770003-96770003 on Build GRCh38
rs908306359 Chr.1: 96769629-96769629 on Build GRCh38
rs147916074 Chr.1: 96769794-96769794 on Build GRCh38
rs771165543 Chr.1: 96769732-96769732 on Build GRCh38
rs1041968003 Chr.1: 96769929-96769929 on Build GRCh38
rs375282955 Chr.1: 96769871-96769871 on Build GRCh38
rs751668384 Chr.1: 96769901-96769901 on Build GRCh38
rs531865117 Chr.1: 96769621-96769621 on Build GRCh38
rs773962988 Chr.1: 96769927-96769927 on Build GRCh38
rs768251633 Chr.1: 96769915-96769915 on Build GRCh38
rs568969186 Chr.1: 96770013-96770013 on Build GRCh38
rs368013391 Chr.1: 96769880-96769880 on Build GRCh38
rs1016689252 Chr.1: 96770025-96770025 on Build GRCh38
rs765144770 Chr.1: 96769715-96769715 on Build GRCh38
rs972588373 Chr.1: 96769751-96769751 on Build GRCh38
rs907346593 Chr.1: 96769641-96769641 on Build GRCh38
rs372119324 Chr.1: 96769655-96769655 on Build GRCh38
rs370745002 Chr.1: 96769675-96769675 on Build GRCh38
rs778918337 Chr.1: 96769708-96769708 on Build GRCh38
rs1054454323 Chr.1: 96769885-96769885 on Build GRCh38
rs41300329 Chr.1: 96769883-96769883 on Build GRCh38
rs199797146 Chr.1: 96769909-96769909 on Build GRCh38
rs191765179 Chr.1: 96769656-96769656 on Build GRCh38
rs748262408 Chr.1: 96769713-96769713 on Build GRCh38
rs899702361 Chr.1: 96769644-96769644 on Build GRCh38
rs1003027984 Chr.1: 96770002-96770002 on Build GRCh38
rs918523724 Chr.1: 96769756-96769756 on Build GRCh38
rs931001658 Chr.1: 96769618-96769618 on Build GRCh38
rs139414147 Chr.1: 96769734-96769734 on Build GRCh38
rs867295961 Chr.1: 96769765-96769765 on Build GRCh38
rs189849284 Chr.1: 96769582-96769582 on Build GRCh38
rs1013697976 Chr.1: 96769829-96769829 on Build GRCh38
rs1027241947 Chr.1: 96769925-96769925 on Build GRCh38
rs750256953 Chr.1: 96769849-96769849 on Build GRCh38
rs370081109 Chr.1: 96769696-96769696 on Build GRCh38
rs747614212 Chr.1: 96769716-96769716 on Build GRCh38
rs779116460 Chr.1: 96769865-96769865 on Build GRCh38
rs982482392 Chr.1: 96769949-96769949 on Build GRCh38
rs908234284 Chr.1: 96769987-96769989 on Build GRCh38
rs986626333 Chr.1: 96769805-96769805 on Build GRCh38
rs932495967 Chr.1: 96770021-96770021 on Build GRCh38
rs764251551 Chr.1: 96769900-96769900 on Build GRCh38
rs761851890 Chr.1: 96769795-96769795 on Build GRCh38
rs1012441741 Chr.1: 96770017-96770017 on Build GRCh38
rs758465346 Chr.1: 96769879-96769879 on Build GRCh38
rs911292281 Chr.1: 96769848-96769848 on Build GRCh38
rs1010521595 Chr.1: 96769606-96769606 on Build GRCh38
rs751310546 Chr.1: 96769661-96769661 on Build GRCh38
rs769600627 Chr.1: 96769755-96769755 on Build GRCh38
rs374967810 Chr.1: 96769902-96769902 on Build GRCh38
rs549238211 Chr.1: 96770008-96770008 on Build GRCh38
rs762810463 Chr.1: 96769750-96769750 on Build GRCh38
rs762566196 Chr.1: 96769783-96769783 on Build GRCh38
rs755156298 Chr.1: 96769856-96769856 on Build GRCh38
rs1040757768 Chr.1: 96769643-96769643 on Build GRCh38
rs1042989802 Chr.1: 96769664-96769664 on Build GRCh38
rs761098728 Chr.1: 96769893-96769894 on Build GRCh38
rs777866923 Chr.1: 96769881-96769881 on Build GRCh38
rs376100837 Chr.1: 96769681-96769681 on Build GRCh38
rs907465914 Chr.1: 96769948-96769948 on Build GRCh38
rs767195687 Chr.1: 96769810-96769810 on Build GRCh38
rs953053645 Chr.1: 96769926-96769926 on Build GRCh38
rs920166344 Chr.1: 96769597-96769597 on Build GRCh38

View Full Product Details