Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100006934-100007470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35A3
Entrez Gene ID
23443
Gene Name
solute carrier family 35 member A3
Gene Aliases
AMRS
UniGene
Hs.448979
Gene Chromosome Location
Chr.1: 99969367-100026979 on Build GRCh38
Transcripts
NM_001271684.1 NM_001271685.1 NM_012243.2 XM_005270691.4 XM_011541135.2 XM_011541136.2 XM_011541137.2 XM_011541138.2 XM_017000869.1 XM_017000870.1 XM_017000871.1 XM_017000872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205032 c.357A>G p.L119L Chr.1: 100007048-100007048 on Build GRCh38
4127542 c.446C>T p.T149I Chr.1: 100007137-100007137 on Build GRCh38
4127543 c.572C>T p.T191I Chr.1: 100007137-100007137 on Build GRCh38
4135544 c.347C>T p.T116M Chr.1: 100007038-100007038 on Build GRCh38
4135545 c.473C>T p.T158M Chr.1: 100007038-100007038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374331207 Chr.1: 100007000-100007000 on Build GRCh38
rs377296535 Chr.1: 100007295-100007295 on Build GRCh38
rs565262231 Chr.1: 100007443-100007443 on Build GRCh38
rs886712711 Chr.1: 100007335-100007335 on Build GRCh38
rs988629039 Chr.1: 100007105-100007105 on Build GRCh38
rs746889061 Chr.1: 100007167-100007167 on Build GRCh38
rs895733014 Chr.1: 100006997-100006997 on Build GRCh38
rs940571339 Chr.1: 100007386-100007386 on Build GRCh38
rs916948707 Chr.1: 100007362-100007362 on Build GRCh38
rs1057033790 Chr.1: 100006982-100006985 on Build GRCh38
rs745715064 Chr.1: 100007182-100007182 on Build GRCh38
rs968926309 Chr.1: 100007245-100007245 on Build GRCh38
rs927066196 Chr.1: 100007282-100007282 on Build GRCh38
rs1020846826 Chr.1: 100007242-100007242 on Build GRCh38
rs936998412 Chr.1: 100006981-100006982 on Build GRCh38
rs558927804 Chr.1: 100006984-100006984 on Build GRCh38
rs754300711 Chr.1: 100007085-100007085 on Build GRCh38
rs774263178 Chr.1: 100007038-100007038 on Build GRCh38
rs752315530 Chr.1: 100007104-100007104 on Build GRCh38
rs574333514 Chr.1: 100007039-100007039 on Build GRCh38
rs531826604 Chr.1: 100007311-100007311 on Build GRCh38
rs768989935 Chr.1: 100007025-100007025 on Build GRCh38
rs776180155 Chr.1: 100007161-100007161 on Build GRCh38
rs992487177 Chr.1: 100007358-100007358 on Build GRCh38
rs761280692 Chr.1: 100007069-100007069 on Build GRCh38
rs1055726 Chr.1: 100007130-100007130 on Build GRCh38
rs371189911 Chr.1: 100007076-100007076 on Build GRCh38
rs200749316 Chr.1: 100007061-100007061 on Build GRCh38
rs930693158 Chr.1: 100007318-100007318 on Build GRCh38
rs768735026 Chr.1: 100007183-100007183 on Build GRCh38
rs950152933 Chr.1: 100007222-100007222 on Build GRCh38
rs183838843 Chr.1: 100007001-100007001 on Build GRCh38
rs370413976 Chr.1: 100007019-100007019 on Build GRCh38
rs917058722 Chr.1: 100007106-100007106 on Build GRCh38
rs373264999 Chr.1: 100007175-100007175 on Build GRCh38
rs757866129 Chr.1: 100007124-100007124 on Build GRCh38
rs879102341 Chr.1: 100007401-100007401 on Build GRCh38
rs766567958 Chr.1: 100006990-100006990 on Build GRCh38
rs754559092 Chr.1: 100007096-100007096 on Build GRCh38
rs997906078 Chr.1: 100006996-100006996 on Build GRCh38
rs377027140 Chr.1: 100007020-100007020 on Build GRCh38
rs751482113 Chr.1: 100007319-100007319 on Build GRCh38
rs978883484 Chr.1: 100007282-100007283 on Build GRCh38
rs545179252 Chr.1: 100007399-100007399 on Build GRCh38
rs764989689 Chr.1: 100007100-100007100 on Build GRCh38
rs377356975 Chr.1: 100007186-100007186 on Build GRCh38
rs563367604 Chr.1: 100007304-100007304 on Build GRCh38
rs989098683 Chr.1: 100007220-100007220 on Build GRCh38
rs538804779 Chr.1: 100006973-100006973 on Build GRCh38
rs761498254 Chr.1: 100007209-100007212 on Build GRCh38
rs773812783 Chr.1: 100007066-100007066 on Build GRCh38
rs534252 Chr.1: 100007048-100007048 on Build GRCh38
rs927290246 Chr.1: 100007276-100007276 on Build GRCh38
rs780843963 Chr.1: 100007176-100007176 on Build GRCh38
rs772208958 Chr.1: 100007188-100007188 on Build GRCh38
rs1044453889 Chr.1: 100007230-100007230 on Build GRCh38

View Full Product Details