Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100961730-100962145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A7
Entrez Gene ID
148867
Gene Name
solute carrier family 30 member 7
Gene Aliases
ZNT7, ZnT-7, ZnTL2
UniGene
Hs.533903
Gene Chromosome Location
Chr.1: 100896076-100996913 on Build GRCh38
Transcripts
NM_001144884.1 NM_133496.4 XM_011540779.2 XM_017000400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
893017 c.887G>T p.R296I Chr.1: 100961872-100961872 on Build GRCh38
893016 c.878T>C p.L293S Chr.1: 100961863-100961863 on Build GRCh38
277322 c.846T>G p.V282V Chr.1: 100961831-100961831 on Build GRCh38
5917411 c.850C>T p.P284S Chr.1: 100961835-100961835 on Build GRCh38
6781477 c.91+2T>C p.? Chr.1: 100961920-100961920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925944713 Chr.1: 100962075-100962075 on Build GRCh38
rs140829028 Chr.1: 100961871-100961871 on Build GRCh38
rs568508014 Chr.1: 100962003-100962003 on Build GRCh38
rs935614758 Chr.1: 100962049-100962049 on Build GRCh38
rs749932198 Chr.1: 100961997-100961997 on Build GRCh38
rs1004022942 Chr.1: 100961978-100961978 on Build GRCh38
rs374978623 Chr.1: 100961933-100961933 on Build GRCh38
rs768326939 Chr.1: 100962116-100962116 on Build GRCh38
rs1013343439 Chr.1: 100961851-100961851 on Build GRCh38
rs758416982 Chr.1: 100961898-100961899 on Build GRCh38
rs539119885 Chr.1: 100961757-100961757 on Build GRCh38
rs368329855 Chr.1: 100961790-100961790 on Build GRCh38
rs200456629 Chr.1: 100961812-100961812 on Build GRCh38
rs78664684 Chr.1: 100962004-100962004 on Build GRCh38
rs965486037 Chr.1: 100961994-100961994 on Build GRCh38
rs777133764 Chr.1: 100961872-100961872 on Build GRCh38
rs750999083 Chr.1: 100961808-100961808 on Build GRCh38
rs372317372 Chr.1: 100962082-100962082 on Build GRCh38
rs148169056 Chr.1: 100962018-100962018 on Build GRCh38
rs778651914 Chr.1: 100961846-100961846 on Build GRCh38
rs369961134 Chr.1: 100962008-100962008 on Build GRCh38
rs145862870 Chr.1: 100961988-100961988 on Build GRCh38
rs748977788 Chr.1: 100962081-100962081 on Build GRCh38
rs142163084 Chr.1: 100961908-100961908 on Build GRCh38
rs200148833 Chr.1: 100961895-100961895 on Build GRCh38
rs574984496 Chr.1: 100961935-100961935 on Build GRCh38
rs962438193 Chr.1: 100961991-100961991 on Build GRCh38
rs745868734 Chr.1: 100961804-100961805 on Build GRCh38
rs754216296 Chr.1: 100961832-100961832 on Build GRCh38
rs370595448 Chr.1: 100961833-100961833 on Build GRCh38
rs747666467 Chr.1: 100961862-100961862 on Build GRCh38
rs979155120 Chr.1: 100962068-100962068 on Build GRCh38
rs534279037 Chr.1: 100962080-100962080 on Build GRCh38
rs780608050 Chr.1: 100961827-100961827 on Build GRCh38
rs755680938 Chr.1: 100962017-100962017 on Build GRCh38
rs371661746 Chr.1: 100961897-100961897 on Build GRCh38
rs982972954 Chr.1: 100961989-100961989 on Build GRCh38
rs552577198 Chr.1: 100961765-100961765 on Build GRCh38
rs774626977 Chr.1: 100961942-100961942 on Build GRCh38
rs1027268517 Chr.1: 100961951-100961951 on Build GRCh38
rs1052534072 Chr.1: 100962058-100962058 on Build GRCh38
rs1015719426 Chr.1: 100961992-100961992 on Build GRCh38
rs559817265 Chr.1: 100962044-100962044 on Build GRCh38
rs373639658 Chr.1: 100962000-100962001 on Build GRCh38
rs5776552 Chr.1: 100961827-100961827 on Build GRCh38

View Full Product Details