Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100965752-100966303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A7
Entrez Gene ID
148867
Gene Name
solute carrier family 30 member 7
Gene Aliases
ZNT7, ZnT-7, ZnTL2
UniGene
Hs.533903
Gene Chromosome Location
Chr.1: 100896076-100996913 on Build GRCh38
Transcripts
NM_001144884.1 NM_133496.4 XM_011540779.2 XM_017000400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6343128 c.952G>T p.V318F Chr.1: 100965787-100965787 on Build GRCh38
4019871 c.1075T>A p.F359I Chr.1: 100965910-100965910 on Build GRCh38
5185076 c.1072_1073insG p.I358fs*15 Chr.1: 100965907-100965908 on Build GRCh38
893019 c.1065A>C p.T355T Chr.1: 100965900-100965900 on Build GRCh38
893018 c.1043C>A p.A348D Chr.1: 100965878-100965878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761031883 Chr.1: 100966037-100966037 on Build GRCh38
rs137866293 Chr.1: 100965824-100965824 on Build GRCh38
rs560910165 Chr.1: 100966101-100966101 on Build GRCh38
rs572066666 Chr.1: 100965942-100965942 on Build GRCh38
rs1018132772 Chr.1: 100965836-100965836 on Build GRCh38
rs997123817 Chr.1: 100966006-100966006 on Build GRCh38
rs1011002734 Chr.1: 100966163-100966163 on Build GRCh38
rs765796802 Chr.1: 100965950-100965950 on Build GRCh38
rs190202631 Chr.1: 100965864-100965864 on Build GRCh38
rs773242564 Chr.1: 100965916-100965916 on Build GRCh38
rs562779627 Chr.1: 100966171-100966171 on Build GRCh38
rs944923041 Chr.1: 100966097-100966097 on Build GRCh38
rs760159960 Chr.1: 100965949-100965949 on Build GRCh38
rs191648216 Chr.1: 100966260-100966260 on Build GRCh38
rs928969182 Chr.1: 100966058-100966058 on Build GRCh38
rs915722869 Chr.1: 100965998-100965998 on Build GRCh38
rs748445728 Chr.1: 100965823-100965823 on Build GRCh38
rs981425276 Chr.1: 100966120-100966120 on Build GRCh38
rs1049416765 Chr.1: 100966177-100966177 on Build GRCh38
rs745859109 Chr.1: 100965892-100965892 on Build GRCh38
rs999520692 Chr.1: 100966192-100966192 on Build GRCh38
rs943173860 Chr.1: 100966235-100966235 on Build GRCh38
rs773506102 Chr.1: 100966036-100966036 on Build GRCh38
rs148152153 Chr.1: 100965908-100965908 on Build GRCh38
rs1031570000 Chr.1: 100965925-100965925 on Build GRCh38
rs929200945 Chr.1: 100966173-100966173 on Build GRCh38
rs72987213 Chr.1: 100966038-100966038 on Build GRCh38
rs897734771 Chr.1: 100965855-100965856 on Build GRCh38
rs779931775 Chr.1: 100965902-100965902 on Build GRCh38
rs763803107 Chr.1: 100965956-100965956 on Build GRCh38
rs556434388 Chr.1: 100965832-100965832 on Build GRCh38
rs375117246 Chr.1: 100965869-100965869 on Build GRCh38
rs761437858 Chr.1: 100965961-100965961 on Build GRCh38
rs1033271937 Chr.1: 100966054-100966054 on Build GRCh38
rs992689997 Chr.1: 100965974-100965974 on Build GRCh38
rs756685376 Chr.1: 100965882-100965882 on Build GRCh38
rs1022380868 Chr.1: 100966175-100966175 on Build GRCh38
rs993721063 Chr.1: 100965821-100965821 on Build GRCh38
rs1048471123 Chr.1: 100966080-100966080 on Build GRCh38
rs538786032 Chr.1: 100965901-100965901 on Build GRCh38
rs772204050 Chr.1: 100965952-100965952 on Build GRCh38
rs140249674 Chr.1: 100965788-100965788 on Build GRCh38
rs574279252 Chr.1: 100966103-100966103 on Build GRCh38
rs748990400 Chr.1: 100965905-100965905 on Build GRCh38
rs372371505 Chr.1: 100965805-100965805 on Build GRCh38
rs780671235 Chr.1: 100965890-100965890 on Build GRCh38
rs909135085 Chr.1: 100966205-100966205 on Build GRCh38
rs957729706 Chr.1: 100966059-100966059 on Build GRCh38
rs775942532 Chr.1: 100965951-100965951 on Build GRCh38
rs759893779 Chr.1: 100966274-100966274 on Build GRCh38
rs572071321 Chr.1: 100966237-100966237 on Build GRCh38
rs751231588 Chr.1: 100965957-100965957 on Build GRCh38
rs960174608 Chr.1: 100965964-100965964 on Build GRCh38
rs775246370 Chr.1: 100965818-100965818 on Build GRCh38
rs12033919 Chr.1: 100966242-100966242 on Build GRCh38
rs774408048 Chr.1: 100965881-100965881 on Build GRCh38
rs763165358 Chr.1: 100965955-100965955 on Build GRCh38
rs1022763330 Chr.1: 100966236-100966236 on Build GRCh38
rs960003607 Chr.1: 100966225-100966225 on Build GRCh38
rs758098642 Chr.1: 100965873-100965873 on Build GRCh38
rs993116002 Chr.1: 100966226-100966226 on Build GRCh38
rs971502553 Chr.1: 100966109-100966109 on Build GRCh38
rs183144833 Chr.1: 100965921-100965921 on Build GRCh38
rs763602102 Chr.1: 100965877-100965877 on Build GRCh38
rs367629804 Chr.1: 100965859-100965859 on Build GRCh38
rs929233770 Chr.1: 100965868-100965868 on Build GRCh38
rs760106806 Chr.1: 100965861-100965861 on Build GRCh38
rs1039219735 Chr.1: 100966098-100966098 on Build GRCh38
rs777344806 Chr.1: 100965839-100965839 on Build GRCh38
rs920323825 Chr.1: 100966249-100966249 on Build GRCh38
rs113502196 Chr.1: 100966225-100966225 on Build GRCh38
rs748359985 Chr.1: 100965924-100965924 on Build GRCh38
rs186800868 Chr.1: 100966202-100966202 on Build GRCh38
rs902539523 Chr.1: 100966188-100966188 on Build GRCh38
rs769638568 Chr.1: 100965806-100965806 on Build GRCh38
rs771193089 Chr.1: 100965932-100965932 on Build GRCh38
rs766998204 Chr.1: 100965838-100965838 on Build GRCh38
rs1038894962 Chr.1: 100966256-100966259 on Build GRCh38
rs890699281 Chr.1: 100966095-100966095 on Build GRCh38
rs773366172 Chr.1: 100965831-100965831 on Build GRCh38
rs781045297 Chr.1: 100965804-100965804 on Build GRCh38
rs543287606 Chr.1: 100966142-100966142 on Build GRCh38
rs557457206 Chr.1: 100966135-100966138 on Build GRCh38
rs757205756 Chr.1: 100965786-100965786 on Build GRCh38

View Full Product Details