Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101236671-101237169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S1PR1
Entrez Gene ID
1901
Gene Name
sphingosine-1-phosphate receptor 1
Gene Aliases
CD363, CHEDG1, D1S3362, ECGF1, EDG-1, EDG1, S1P1
UniGene
Hs.154210
Gene Chromosome Location
Chr.1: 101236749-101241520 on Build GRCh38
Transcripts
NM_001320730.1 NM_001400.4

SNP Detail

SNP ID Chromosome Location
rs980615500 Chr.1: 101236941-101236941 on Build GRCh38
rs572708090 Chr.1: 101236716-101236716 on Build GRCh38
rs200122634 Chr.1: 101237080-101237080 on Build GRCh38
rs369998565 Chr.1: 101237023-101237023 on Build GRCh38
rs919625847 Chr.1: 101237006-101237006 on Build GRCh38
rs983050207 Chr.1: 101236735-101236735 on Build GRCh38
rs932674340 Chr.1: 101236852-101236852 on Build GRCh38
rs952546713 Chr.1: 101236722-101236722 on Build GRCh38
rs994528131 Chr.1: 101236963-101236963 on Build GRCh38
rs199540243 Chr.1: 101237052-101237052 on Build GRCh38
rs1008118915 Chr.1: 101236874-101236874 on Build GRCh38
rs765246247 Chr.1: 101236991-101236992 on Build GRCh38
rs926059597 Chr.1: 101236895-101236895 on Build GRCh38
rs201315127 Chr.1: 101237046-101237046 on Build GRCh38
rs200711236 Chr.1: 101237043-101237043 on Build GRCh38
rs917556296 Chr.1: 101236730-101236730 on Build GRCh38
rs200886101 Chr.1: 101237084-101237084 on Build GRCh38
rs540252941 Chr.1: 101236753-101236755 on Build GRCh38
rs931090808 Chr.1: 101236847-101236847 on Build GRCh38
rs199710944 Chr.1: 101237040-101237040 on Build GRCh38
rs761390265 Chr.1: 101236730-101236733 on Build GRCh38
rs1029351051 Chr.1: 101237042-101237042 on Build GRCh38
rs931060530 Chr.1: 101237010-101237010 on Build GRCh38
rs975024303 Chr.1: 101236843-101236843 on Build GRCh38
rs569770064 Chr.1: 101237009-101237009 on Build GRCh38
rs573961720 Chr.1: 101236831-101236832 on Build GRCh38
rs146490475 Chr.1: 101236803-101236803 on Build GRCh38
rs777331630 Chr.1: 101236969-101236969 on Build GRCh38
rs4987248 Chr.1: 101236990-101236990 on Build GRCh38
rs988895925 Chr.1: 101237083-101237083 on Build GRCh38
rs200656819 Chr.1: 101237069-101237069 on Build GRCh38
rs904548881 Chr.1: 101237140-101237140 on Build GRCh38
rs1053224679 Chr.1: 101236927-101236927 on Build GRCh38
rs187615587 Chr.1: 101236939-101236939 on Build GRCh38
rs201015942 Chr.1: 101237143-101237143 on Build GRCh38
rs201366822 Chr.1: 101236862-101236862 on Build GRCh38
rs375564918 Chr.1: 101236940-101236940 on Build GRCh38
rs1021607089 Chr.1: 101237131-101237131 on Build GRCh38
rs201650910 Chr.1: 101236989-101236989 on Build GRCh38
rs200252006 Chr.1: 101237022-101237022 on Build GRCh38
rs1010248094 Chr.1: 101236928-101236928 on Build GRCh38
rs1056027422 Chr.1: 101237138-101237138 on Build GRCh38
rs1004598569 Chr.1: 101237004-101237004 on Build GRCh38
rs201172051 Chr.1: 101236992-101236992 on Build GRCh38
rs1027332783 Chr.1: 101236719-101236719 on Build GRCh38
rs199824181 Chr.1: 101236851-101236851 on Build GRCh38
rs898253583 Chr.1: 101236844-101236844 on Build GRCh38
rs529885099 Chr.1: 101236985-101236985 on Build GRCh38
rs3765684 Chr.1: 101237032-101237032 on Build GRCh38
rs558339190 Chr.1: 101236812-101236812 on Build GRCh38
rs375482957 Chr.1: 101236727-101236729 on Build GRCh38
rs1034513201 Chr.1: 101236948-101236948 on Build GRCh38
rs201766402 Chr.1: 101237090-101237090 on Build GRCh38
rs1027300698 Chr.1: 101236987-101236987 on Build GRCh38
rs962139049 Chr.1: 101236849-101236849 on Build GRCh38
rs562333104 Chr.1: 101236922-101236922 on Build GRCh38
rs202079706 Chr.1: 101237076-101237076 on Build GRCh38
rs549859367 Chr.1: 101236859-101236859 on Build GRCh38
rs1001601173 Chr.1: 101237142-101237142 on Build GRCh38
rs1040947955 Chr.1: 101237050-101237050 on Build GRCh38
rs200027206 Chr.1: 101237063-101237063 on Build GRCh38
rs934717476 Chr.1: 101236909-101236909 on Build GRCh38
rs543936054 Chr.1: 101236912-101236912 on Build GRCh38
rs939709568 Chr.1: 101236741-101236741 on Build GRCh38
rs1018790205 Chr.1: 101237008-101237008 on Build GRCh38
rs780714868 Chr.1: 101237054-101237054 on Build GRCh38
rs199901447 Chr.1: 101237100-101237100 on Build GRCh38
rs201816028 Chr.1: 101237057-101237057 on Build GRCh38
rs202090953 Chr.1: 101236880-101236880 on Build GRCh38
rs200787075 Chr.1: 101236946-101236946 on Build GRCh38
rs200034816 Chr.1: 101236891-101236891 on Build GRCh38
rs200789386 Chr.1: 101237053-101237053 on Build GRCh38
rs891761789 Chr.1: 101236926-101236926 on Build GRCh38
rs202188906 Chr.1: 101237012-101237012 on Build GRCh38
rs993072007 Chr.1: 101236702-101236702 on Build GRCh38
rs937346604 Chr.1: 101237124-101237124 on Build GRCh38
rs869219354 Chr.1: 101236752-101236755 on Build GRCh38
rs917333563 Chr.1: 101237086-101237086 on Build GRCh38
rs781188725 Chr.1: 101236727-101236730 on Build GRCh38
rs3835397 Chr.1: 101236992-101236992 on Build GRCh38
rs950292537 Chr.1: 101237108-101237108 on Build GRCh38
rs895933110 Chr.1: 101236930-101236930 on Build GRCh38
rs960499268 Chr.1: 101236962-101236962 on Build GRCh38

View Full Product Details