Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 102882957-102883458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
UniGene
Hs.523446
Gene Chromosome Location
Chr.1: 102876467-103108580 on Build GRCh38
Transcripts
NM_001190709.1 NM_001854.3 NM_080629.2 NM_080630.3 NR_134980.1 XM_017000334.1 XM_017000335.1 XM_017000337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057246 c.4936G>A p.E1646K Chr.1: 102883234-102883234 on Build GRCh38
6120295 c.4966G>T p.E1656* Chr.1: 102883204-102883204 on Build GRCh38
5417239 c.4859G>T p.G1620V Chr.1: 102883311-102883311 on Build GRCh38
4605474 c.4885G>C p.G1629R Chr.1: 102883285-102883285 on Build GRCh38
3470421 c.4867T>C p.W1623R Chr.1: 102883303-102883303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751423255 Chr.1: 102883167-102883167 on Build GRCh38
rs532325392 Chr.1: 102883094-102883094 on Build GRCh38
rs767308373 Chr.1: 102883332-102883332 on Build GRCh38
rs551323025 Chr.1: 102883160-102883160 on Build GRCh38
rs1049955088 Chr.1: 102883410-102883410 on Build GRCh38
rs886044973 Chr.1: 102883264-102883264 on Build GRCh38
rs762566719 Chr.1: 102883315-102883315 on Build GRCh38
rs569884799 Chr.1: 102883162-102883162 on Build GRCh38
rs776122573 Chr.1: 102883219-102883219 on Build GRCh38
rs750288600 Chr.1: 102883341-102883341 on Build GRCh38
rs540713668 Chr.1: 102883135-102883135 on Build GRCh38
rs769627169 Chr.1: 102883308-102883308 on Build GRCh38
rs890864371 Chr.1: 102883422-102883422 on Build GRCh38
rs780916396 Chr.1: 102883201-102883201 on Build GRCh38
rs747421087 Chr.1: 102883274-102883274 on Build GRCh38
rs776445554 Chr.1: 102883148-102883148 on Build GRCh38
rs74108030 Chr.1: 102883097-102883097 on Build GRCh38
rs369265343 Chr.1: 102883235-102883235 on Build GRCh38
rs538741278 Chr.1: 102883406-102883406 on Build GRCh38
rs983620607 Chr.1: 102883281-102883281 on Build GRCh38
rs565111127 Chr.1: 102882989-102882989 on Build GRCh38
rs184088500 Chr.1: 102883221-102883221 on Build GRCh38
rs372504909 Chr.1: 102883284-102883284 on Build GRCh38
rs967969297 Chr.1: 102883405-102883405 on Build GRCh38
rs772849014 Chr.1: 102883340-102883340 on Build GRCh38
rs764144144 Chr.1: 102883163-102883163 on Build GRCh38
rs528476341 Chr.1: 102883119-102883119 on Build GRCh38
rs757107063 Chr.1: 102883180-102883180 on Build GRCh38
rs955962479 Chr.1: 102882986-102882986 on Build GRCh38
rs763472511 Chr.1: 102883320-102883320 on Build GRCh38
rs771436179 Chr.1: 102883285-102883285 on Build GRCh38
rs1463041 Chr.1: 102882983-102882983 on Build GRCh38
rs995074235 Chr.1: 102883188-102883188 on Build GRCh38
rs1037924572 Chr.1: 102883347-102883347 on Build GRCh38
rs779106317 Chr.1: 102883225-102883225 on Build GRCh38
rs979301565 Chr.1: 102883427-102883427 on Build GRCh38
rs942252121 Chr.1: 102883338-102883338 on Build GRCh38
rs377036654 Chr.1: 102883294-102883294 on Build GRCh38
rs74778421 Chr.1: 102883205-102883205 on Build GRCh38
rs533130296 Chr.1: 102883158-102883158 on Build GRCh38
rs757945914 Chr.1: 102883414-102883414 on Build GRCh38
rs752820346 Chr.1: 102883355-102883355 on Build GRCh38
rs769489959 Chr.1: 102883329-102883330 on Build GRCh38
rs937447857 Chr.1: 102883015-102883015 on Build GRCh38
rs774985706 Chr.1: 102883314-102883314 on Build GRCh38
rs146179957 Chr.1: 102883330-102883330 on Build GRCh38
rs772011676 Chr.1: 102883242-102883242 on Build GRCh38
rs565284158 Chr.1: 102883146-102883146 on Build GRCh38
rs143660176 Chr.1: 102883076-102883079 on Build GRCh38
rs1021570840 Chr.1: 102883002-102883002 on Build GRCh38
rs759206962 Chr.1: 102883153-102883153 on Build GRCh38
rs139207425 Chr.1: 102883361-102883361 on Build GRCh38
rs550899555 Chr.1: 102883183-102883183 on Build GRCh38
rs748675627 Chr.1: 102883358-102883358 on Build GRCh38
rs369838465 Chr.1: 102883349-102883349 on Build GRCh38
rs752387950 Chr.1: 102883344-102883344 on Build GRCh38
rs1055861496 Chr.1: 102883073-102883073 on Build GRCh38
rs898111956 Chr.1: 102883130-102883130 on Build GRCh38
rs778371759 Chr.1: 102883253-102883253 on Build GRCh38
rs78368739 Chr.1: 102883297-102883297 on Build GRCh38
rs553008435 Chr.1: 102883354-102883354 on Build GRCh38

View Full Product Details