Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20170436-20170982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGO1
Entrez Gene ID
151648
Gene Name
shugoshin 1
Gene Aliases
CAID, NY-BR-85, SGO, SGOL1
UniGene
Hs.105153
Gene Chromosome Location
Chr.3: 20160593-20188143 on Build GRCh38
Transcripts
NM_001012409.3 NM_001012410.4 NM_001012411.3 NM_001012412.4 NM_001012413.3 NM_001199251.2 NM_001199252.2 NM_001199253.2 NM_001199254.2 NM_001199255.2 NM_001199256.2 NM_001199257.2 NM_138484.4 NR_131179.1 NR_131180.1 XM_011533373.2 XM_011533375.2 XM_011533376.2 XM_011533377.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389589 c.1553G>A p.R518H Chr.3: 20170735-20170735 on Build GRCh38
1043296 c.1520C>A p.S507Y Chr.3: 20170768-20170768 on Build GRCh38
1422144 c.1564_1564+1insA p.R522fs*29 Chr.3: 20170723-20170724 on Build GRCh38
4725480 c.1554T>C p.R518R Chr.3: 20170734-20170734 on Build GRCh38
5623499 c.1473G>A p.S491S Chr.3: 20170815-20170815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13060316 Chr.3: 20170814-20170814 on Build GRCh38
rs943194120 Chr.3: 20170482-20170482 on Build GRCh38
rs768593328 Chr.3: 20170773-20170773 on Build GRCh38
rs764819662 Chr.3: 20170784-20170784 on Build GRCh38
rs989254989 Chr.3: 20170532-20170532 on Build GRCh38
rs768145706 Chr.3: 20170844-20170844 on Build GRCh38
rs888624612 Chr.3: 20170691-20170691 on Build GRCh38
rs752787437 Chr.3: 20170847-20170847 on Build GRCh38
rs551717211 Chr.3: 20170550-20170550 on Build GRCh38
rs201334733 Chr.3: 20170820-20170820 on Build GRCh38
rs766835053 Chr.3: 20170803-20170803 on Build GRCh38
rs958022708 Chr.3: 20170526-20170526 on Build GRCh38
rs755106059 Chr.3: 20170711-20170711 on Build GRCh38
rs749588196 Chr.3: 20170857-20170857 on Build GRCh38
rs114711704 Chr.3: 20170696-20170696 on Build GRCh38
rs746059096 Chr.3: 20170723-20170726 on Build GRCh38
rs769210904 Chr.3: 20170798-20170798 on Build GRCh38
rs186498395 Chr.3: 20170669-20170669 on Build GRCh38
rs924967612 Chr.3: 20170890-20170890 on Build GRCh38
rs200039832 Chr.3: 20170752-20170752 on Build GRCh38
rs936301027 Chr.3: 20170924-20170924 on Build GRCh38
rs761342521 Chr.3: 20170782-20170782 on Build GRCh38
rs747279794 Chr.3: 20170764-20170766 on Build GRCh38
rs761608382 Chr.3: 20170828-20170828 on Build GRCh38
rs761397128 Chr.3: 20170692-20170692 on Build GRCh38
rs550006063 Chr.3: 20170460-20170460 on Build GRCh38
rs923089350 Chr.3: 20170875-20170875 on Build GRCh38
rs775543954 Chr.3: 20170679-20170679 on Build GRCh38
rs775282365 Chr.3: 20170763-20170763 on Build GRCh38
rs534484801 Chr.3: 20170587-20170587 on Build GRCh38
rs964741983 Chr.3: 20170848-20170848 on Build GRCh38
rs759383215 Chr.3: 20170673-20170673 on Build GRCh38
rs376190704 Chr.3: 20170854-20170854 on Build GRCh38
rs190246133 Chr.3: 20170953-20170953 on Build GRCh38
rs999480187 Chr.3: 20170755-20170755 on Build GRCh38
rs796405177 Chr.3: 20170696-20170697 on Build GRCh38
rs771866725 Chr.3: 20170675-20170675 on Build GRCh38
rs112748317 Chr.3: 20170713-20170713 on Build GRCh38
rs910853116 Chr.3: 20170543-20170543 on Build GRCh38
rs77194360 Chr.3: 20170735-20170735 on Build GRCh38
rs778883282 Chr.3: 20170623-20170626 on Build GRCh38
rs978025749 Chr.3: 20170885-20170885 on Build GRCh38
rs752150449 Chr.3: 20170707-20170707 on Build GRCh38
rs746263403 Chr.3: 20170508-20170512 on Build GRCh38
rs1052930 Chr.3: 20170697-20170697 on Build GRCh38
rs963794460 Chr.3: 20170903-20170903 on Build GRCh38
rs1017626637 Chr.3: 20170801-20170801 on Build GRCh38
rs770839706 Chr.3: 20170668-20170668 on Build GRCh38
rs781480888 Chr.3: 20170712-20170712 on Build GRCh38
rs529100077 Chr.3: 20170459-20170459 on Build GRCh38
rs373246191 Chr.3: 20170517-20170517 on Build GRCh38
rs201164840 Chr.3: 20170682-20170682 on Build GRCh38
rs772386209 Chr.3: 20170723-20170724 on Build GRCh38
rs936438467 Chr.3: 20170629-20170629 on Build GRCh38
rs367780520 Chr.3: 20170689-20170689 on Build GRCh38
rs976380754 Chr.3: 20170947-20170947 on Build GRCh38
rs755488659 Chr.3: 20170841-20170841 on Build GRCh38
rs1042449533 Chr.3: 20170541-20170541 on Build GRCh38
rs945278587 Chr.3: 20170522-20170522 on Build GRCh38
rs572780063 Chr.3: 20170883-20170883 on Build GRCh38
rs776052006 Chr.3: 20170779-20170779 on Build GRCh38
rs143399015 Chr.3: 20170511-20170511 on Build GRCh38
rs1040712084 Chr.3: 20170583-20170583 on Build GRCh38
rs73193250 Chr.3: 20170644-20170644 on Build GRCh38
rs748447624 Chr.3: 20170656-20170656 on Build GRCh38
rs745624804 Chr.3: 20170736-20170736 on Build GRCh38
rs772969129 Chr.3: 20170787-20170787 on Build GRCh38
rs750711928 Chr.3: 20170695-20170695 on Build GRCh38
rs201941394 Chr.3: 20170685-20170685 on Build GRCh38
rs753471998 Chr.3: 20170686-20170686 on Build GRCh38
rs957911981 Chr.3: 20170877-20170877 on Build GRCh38
rs976329103 Chr.3: 20170861-20170861 on Build GRCh38
rs756915448 Chr.3: 20170932-20170932 on Build GRCh38
rs780589746 Chr.3: 20170759-20170759 on Build GRCh38
rs756287205 Chr.3: 20170853-20170853 on Build GRCh38
rs776221311 Chr.3: 20170804-20170804 on Build GRCh38
rs748255528 Chr.3: 20170719-20170719 on Build GRCh38
rs1024338614 Chr.3: 20170740-20170740 on Build GRCh38
rs760719227 Chr.3: 20170766-20170766 on Build GRCh38
rs139302138 Chr.3: 20170556-20170556 on Build GRCh38
rs763197949 Chr.3: 20170796-20170796 on Build GRCh38
rs180866174 Chr.3: 20170466-20170466 on Build GRCh38

View Full Product Details