Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 21751215-21751721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF385D
Entrez Gene ID
79750
Gene Name
zinc finger protein 385D
Gene Aliases
ZNF659
UniGene
Hs.21026
Gene Chromosome Location
Chr.3: 21418419-22372641 on Build GRCh38
Transcripts
NM_024697.2 XM_011534122.1 XM_011534123.2 XM_011534124.2 XM_017007191.1 XM_017007192.1 XM_017007193.1 XM_017007194.1 XM_017007195.1 XM_017007196.1 XM_017007198.1 XM_017007199.1 XM_017007203.1

SNP Detail

SNP ID Chromosome Location
rs924724875 Chr.3: 21751467-21751467 on Build GRCh38
rs993141784 Chr.3: 21751538-21751538 on Build GRCh38
rs567449441 Chr.3: 21751314-21751314 on Build GRCh38
rs34599430 Chr.3: 21751509-21751509 on Build GRCh38
rs991660517 Chr.3: 21751298-21751298 on Build GRCh38
rs922590326 Chr.3: 21751653-21751653 on Build GRCh38
rs747907946 Chr.3: 21751277-21751277 on Build GRCh38
rs866847713 Chr.3: 21751457-21751457 on Build GRCh38
rs1006275941 Chr.3: 21751267-21751267 on Build GRCh38
rs545260814 Chr.3: 21751460-21751460 on Build GRCh38
rs550189173 Chr.3: 21751352-21751352 on Build GRCh38
rs879883691 Chr.3: 21751246-21751246 on Build GRCh38
rs762132014 Chr.3: 21751358-21751358 on Build GRCh38
rs569055125 Chr.3: 21751362-21751362 on Build GRCh38
rs1013157153 Chr.3: 21751589-21751589 on Build GRCh38
rs890759049 Chr.3: 21751436-21751436 on Build GRCh38
rs1031715610 Chr.3: 21751631-21751631 on Build GRCh38
rs986007669 Chr.3: 21751689-21751689 on Build GRCh38
rs754406715 Chr.3: 21751317-21751317 on Build GRCh38
rs531881350 Chr.3: 21751639-21751639 on Build GRCh38
rs897573646 Chr.3: 21751531-21751531 on Build GRCh38
rs561783627 Chr.3: 21751646-21751646 on Build GRCh38
rs193090539 Chr.3: 21751524-21751524 on Build GRCh38
rs923167445 Chr.3: 21751483-21751483 on Build GRCh38
rs561037895 Chr.3: 21751624-21751624 on Build GRCh38
rs976722670 Chr.3: 21751480-21751480 on Build GRCh38
rs993602131 Chr.3: 21751251-21751251 on Build GRCh38
rs114512374 Chr.3: 21751396-21751396 on Build GRCh38
rs888866354 Chr.3: 21751253-21751253 on Build GRCh38
rs960163908 Chr.3: 21751297-21751297 on Build GRCh38
rs769540739 Chr.3: 21751293-21751293 on Build GRCh38
rs1028665380 Chr.3: 21751250-21751250 on Build GRCh38
rs749566669 Chr.3: 21751474-21751474 on Build GRCh38
rs952124502 Chr.3: 21751278-21751278 on Build GRCh38
rs549036433 Chr.3: 21751291-21751291 on Build GRCh38
rs960299336 Chr.3: 21751421-21751421 on Build GRCh38
rs972321375 Chr.3: 21751288-21751288 on Build GRCh38
rs534001258 Chr.3: 21751407-21751407 on Build GRCh38
rs898076957 Chr.3: 21751452-21751452 on Build GRCh38
rs947508566 Chr.3: 21751323-21751323 on Build GRCh38
rs1035906263 Chr.3: 21751413-21751413 on Build GRCh38
rs560433239 Chr.3: 21751477-21751477 on Build GRCh38
rs912232840 Chr.3: 21751612-21751612 on Build GRCh38
rs1050238602 Chr.3: 21751364-21751364 on Build GRCh38
rs979284435 Chr.3: 21751324-21751324 on Build GRCh38
rs1013062927 Chr.3: 21751433-21751433 on Build GRCh38
rs1056833232 Chr.3: 21751543-21751543 on Build GRCh38
rs895513269 Chr.3: 21751584-21751584 on Build GRCh38
rs1033615819 Chr.3: 21751316-21751316 on Build GRCh38
rs774528276 Chr.3: 21751528-21751528 on Build GRCh38
rs910365310 Chr.3: 21751502-21751502 on Build GRCh38
rs543857849 Chr.3: 21751414-21751414 on Build GRCh38
rs979282294 Chr.3: 21751521-21751521 on Build GRCh38
rs945241737 Chr.3: 21751371-21751371 on Build GRCh38
rs897485254 Chr.3: 21751401-21751401 on Build GRCh38
rs562454796 Chr.3: 21751416-21751416 on Build GRCh38
rs941895725 Chr.3: 21751508-21751508 on Build GRCh38
rs887911072 Chr.3: 21751683-21751683 on Build GRCh38
rs191637163 Chr.3: 21751328-21751328 on Build GRCh38
rs183918407 Chr.3: 21751363-21751363 on Build GRCh38
rs915975992 Chr.3: 21751308-21751308 on Build GRCh38
rs932404583 Chr.3: 21751583-21751583 on Build GRCh38
rs780695210 Chr.3: 21751637-21751637 on Build GRCh38
rs910491865 Chr.3: 21751691-21751691 on Build GRCh38
rs901776161 Chr.3: 21751359-21751359 on Build GRCh38
rs775930532 Chr.3: 21751666-21751666 on Build GRCh38
rs933252060 Chr.3: 21751494-21751494 on Build GRCh38
rs549199589 Chr.3: 21751621-21751621 on Build GRCh38
rs759705810 Chr.3: 21751608-21751608 on Build GRCh38
rs934697482 Chr.3: 21751344-21751344 on Build GRCh38
rs993580344 Chr.3: 21751403-21751403 on Build GRCh38
rs1050329722 Chr.3: 21751496-21751496 on Build GRCh38
rs369498668 Chr.3: 21751245-21751254 on Build GRCh38
rs113074090 Chr.3: 21751501-21751501 on Build GRCh38
rs1040988496 Chr.3: 21751442-21751442 on Build GRCh38
rs7623361 Chr.3: 21751274-21751274 on Build GRCh38
rs1023262580 Chr.3: 21751601-21751601 on Build GRCh38
rs555482250 Chr.3: 21751409-21751409 on Build GRCh38
rs189218752 Chr.3: 21751455-21751455 on Build GRCh38
rs1036694803 Chr.3: 21751623-21751623 on Build GRCh38
rs7634827 Chr.3: 21751390-21751390 on Build GRCh38
rs997338080 Chr.3: 21751245-21751245 on Build GRCh38
rs944985741 Chr.3: 21751440-21751440 on Build GRCh38
rs573717244 Chr.3: 21751412-21751412 on Build GRCh38
rs888835051 Chr.3: 21751365-21751365 on Build GRCh38
rs976691793 Chr.3: 21751650-21751650 on Build GRCh38
rs960950271 Chr.3: 21751500-21751500 on Build GRCh38
rs978867488 Chr.3: 21751465-21751465 on Build GRCh38
rs985174760 Chr.3: 21751590-21751590 on Build GRCh38
rs1049443163 Chr.3: 21751418-21751418 on Build GRCh38
rs956338047 Chr.3: 21751632-21751632 on Build GRCh38
rs752135851 Chr.3: 21751648-21751649 on Build GRCh38

View Full Product Details