Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23944863-23945365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1D2
Entrez Gene ID
9975
Gene Name
nuclear receptor subfamily 1 group D member 2
Gene Aliases
BD73, EAR-1R, RVR
UniGene
Hs.37288
Gene Chromosome Location
Chr.3: 23945260-23980618 on Build GRCh38
Transcripts
NM_005126.4 NR_110524.1

SNP Detail

SNP ID Chromosome Location
rs528459990 Chr.3: 23945136-23945136 on Build GRCh38
rs56317749 Chr.3: 23945256-23945256 on Build GRCh38
rs530804316 Chr.3: 23945007-23945007 on Build GRCh38
rs374610835 Chr.3: 23945228-23945228 on Build GRCh38
rs1053461115 Chr.3: 23945029-23945029 on Build GRCh38
rs546457030 Chr.3: 23945085-23945085 on Build GRCh38
rs1049949295 Chr.3: 23945023-23945023 on Build GRCh38
rs369272943 Chr.3: 23945207-23945207 on Build GRCh38
rs1039485472 Chr.3: 23945307-23945307 on Build GRCh38
rs193249552 Chr.3: 23945138-23945138 on Build GRCh38
rs923816615 Chr.3: 23945242-23945242 on Build GRCh38
rs564359446 Chr.3: 23945129-23945129 on Build GRCh38
rs950960612 Chr.3: 23945320-23945320 on Build GRCh38
rs925995057 Chr.3: 23944989-23944989 on Build GRCh38
rs970306325 Chr.3: 23945125-23945125 on Build GRCh38
rs979271003 Chr.3: 23944982-23944982 on Build GRCh38
rs575820363 Chr.3: 23945084-23945084 on Build GRCh38
rs1038493738 Chr.3: 23945291-23945291 on Build GRCh38
rs1014116019 Chr.3: 23944932-23944932 on Build GRCh38
rs184000561 Chr.3: 23945329-23945329 on Build GRCh38
rs145516978 Chr.3: 23945153-23945154 on Build GRCh38
rs900948788 Chr.3: 23945309-23945309 on Build GRCh38
rs942134452 Chr.3: 23944969-23944969 on Build GRCh38
rs569009224 Chr.3: 23945192-23945192 on Build GRCh38
rs1007034217 Chr.3: 23945301-23945301 on Build GRCh38
rs956028711 Chr.3: 23944996-23944996 on Build GRCh38
rs370432478 Chr.3: 23945164-23945164 on Build GRCh38
rs950969916 Chr.3: 23945099-23945099 on Build GRCh38
rs963789658 Chr.3: 23945122-23945122 on Build GRCh38
rs949264053 Chr.3: 23944983-23944983 on Build GRCh38
rs1014603112 Chr.3: 23945105-23945105 on Build GRCh38
rs956101724 Chr.3: 23945137-23945137 on Build GRCh38
rs542016275 Chr.3: 23945079-23945079 on Build GRCh38
rs575089183 Chr.3: 23945068-23945068 on Build GRCh38
rs866474496 Chr.3: 23944884-23944884 on Build GRCh38
rs1047371792 Chr.3: 23945096-23945096 on Build GRCh38
rs950689624 Chr.3: 23944894-23944894 on Build GRCh38
rs947753279 Chr.3: 23945087-23945105 on Build GRCh38
rs1046283381 Chr.3: 23945177-23945177 on Build GRCh38
rs550471766 Chr.3: 23945182-23945182 on Build GRCh38
rs893514897 Chr.3: 23945043-23945043 on Build GRCh38
rs934224207 Chr.3: 23945017-23945017 on Build GRCh38
rs1030464820 Chr.3: 23945318-23945318 on Build GRCh38
rs907171039 Chr.3: 23945010-23945010 on Build GRCh38
rs115855981 Chr.3: 23945165-23945165 on Build GRCh38
rs909249151 Chr.3: 23944968-23944969 on Build GRCh38
rs983432965 Chr.3: 23944925-23944925 on Build GRCh38
rs988810023 Chr.3: 23945008-23945008 on Build GRCh38
rs911392708 Chr.3: 23945041-23945041 on Build GRCh38
rs997599762 Chr.3: 23945314-23945314 on Build GRCh38
rs983822745 Chr.3: 23945101-23945101 on Build GRCh38
rs28510558 Chr.3: 23944975-23944975 on Build GRCh38
rs909248036 Chr.3: 23945115-23945117 on Build GRCh38
rs935246294 Chr.3: 23945247-23945247 on Build GRCh38
rs1023199465 Chr.3: 23944960-23944960 on Build GRCh38
rs941504557 Chr.3: 23945059-23945059 on Build GRCh38
rs1055356201 Chr.3: 23944974-23944974 on Build GRCh38
rs551642237 Chr.3: 23945286-23945286 on Build GRCh38
rs61176987 Chr.3: 23945208-23945208 on Build GRCh38
rs529066722 Chr.3: 23945146-23945146 on Build GRCh38
rs1001957324 Chr.3: 23945139-23945139 on Build GRCh38
rs911438830 Chr.3: 23945285-23945285 on Build GRCh38
rs999169435 Chr.3: 23945016-23945016 on Build GRCh38
rs561802465 Chr.3: 23945144-23945144 on Build GRCh38
rs1053856329 Chr.3: 23945248-23945248 on Build GRCh38
rs557650329 Chr.3: 23945082-23945082 on Build GRCh38
rs922883894 Chr.3: 23945011-23945011 on Build GRCh38
rs555498136 Chr.3: 23945036-23945036 on Build GRCh38
rs902446928 Chr.3: 23945009-23945013 on Build GRCh38
rs959938595 Chr.3: 23944887-23944887 on Build GRCh38
rs986123377 Chr.3: 23945279-23945279 on Build GRCh38
rs1030515678 Chr.3: 23945095-23945095 on Build GRCh38
rs895758679 Chr.3: 23945104-23945104 on Build GRCh38
rs1033136074 Chr.3: 23945131-23945131 on Build GRCh38
rs1022854700 Chr.3: 23945116-23945116 on Build GRCh38

View Full Product Details