Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25718790-25719339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGLY1
Entrez Gene ID
55768
Gene Name
N-glycanase 1
Gene Aliases
CDDG, CDG1V, PNG1, PNGase
UniGene
Hs.368960
Gene Chromosome Location
Chr.3: 25718944-25790039 on Build GRCh38
Transcripts
NM_001145293.1 NM_001145294.1 NM_001145295.1 NM_018297.3 XM_005265316.1 XM_005265317.1 XM_011533944.1

SNP Detail

SNP ID Chromosome Location
rs1033452228 Chr.3: 25719032-25719032 on Build GRCh38
rs994229868 Chr.3: 25719016-25719016 on Build GRCh38
rs879807736 Chr.3: 25719004-25719004 on Build GRCh38
rs1038522433 Chr.3: 25719002-25719002 on Build GRCh38
rs1037513604 Chr.3: 25718872-25718872 on Build GRCh38
rs1003331600 Chr.3: 25719023-25719023 on Build GRCh38
rs965762194 Chr.3: 25719120-25719120 on Build GRCh38
rs1018714391 Chr.3: 25719101-25719101 on Build GRCh38
rs898634210 Chr.3: 25719248-25719248 on Build GRCh38
rs1038960851 Chr.3: 25719245-25719245 on Build GRCh38
rs557530300 Chr.3: 25719144-25719144 on Build GRCh38
rs749010041 Chr.3: 25719275-25719275 on Build GRCh38
rs554695948 Chr.3: 25719099-25719099 on Build GRCh38
rs76503860 Chr.3: 25719223-25719223 on Build GRCh38
rs188241587 Chr.3: 25719131-25719131 on Build GRCh38
rs1008230071 Chr.3: 25719042-25719045 on Build GRCh38
rs755711297 Chr.3: 25719045-25719045 on Build GRCh38
rs572044910 Chr.3: 25719304-25719304 on Build GRCh38
rs984774961 Chr.3: 25719234-25719234 on Build GRCh38
rs1056600172 Chr.3: 25718961-25718961 on Build GRCh38
rs1056047310 Chr.3: 25719184-25719184 on Build GRCh38
rs1004337537 Chr.3: 25718822-25718822 on Build GRCh38
rs193176664 Chr.3: 25719135-25719135 on Build GRCh38
rs915058959 Chr.3: 25719069-25719069 on Build GRCh38
rs948311860 Chr.3: 25719204-25719204 on Build GRCh38
rs1036952692 Chr.3: 25719302-25719302 on Build GRCh38
rs184665286 Chr.3: 25719289-25719289 on Build GRCh38
rs1021985782 Chr.3: 25719058-25719058 on Build GRCh38
rs199527543 Chr.3: 25718922-25718922 on Build GRCh38
rs376817718 Chr.3: 25719107-25719107 on Build GRCh38
rs977779430 Chr.3: 25719097-25719097 on Build GRCh38
rs577516465 Chr.3: 25719190-25719190 on Build GRCh38
rs552259759 Chr.3: 25718972-25718972 on Build GRCh38
rs964645744 Chr.3: 25719060-25719060 on Build GRCh38
rs77392761 Chr.3: 25719096-25719096 on Build GRCh38
rs748858038 Chr.3: 25719067-25719067 on Build GRCh38
rs946470727 Chr.3: 25718850-25718850 on Build GRCh38
rs976930531 Chr.3: 25719130-25719130 on Build GRCh38
rs1012861132 Chr.3: 25718981-25718984 on Build GRCh38
rs372672237 Chr.3: 25719067-25719071 on Build GRCh38
rs752218968 Chr.3: 25718875-25718875 on Build GRCh38
rs79425415 Chr.3: 25719222-25719222 on Build GRCh38
rs915003563 Chr.3: 25718847-25718847 on Build GRCh38
rs770629842 Chr.3: 25719306-25719306 on Build GRCh38
rs891209060 Chr.3: 25719308-25719308 on Build GRCh38
rs184088528 Chr.3: 25718997-25718997 on Build GRCh38
rs895395651 Chr.3: 25718977-25718977 on Build GRCh38
rs556031820 Chr.3: 25719166-25719169 on Build GRCh38
rs1011229717 Chr.3: 25719057-25719057 on Build GRCh38
rs916844284 Chr.3: 25719196-25719196 on Build GRCh38
rs1042056983 Chr.3: 25718871-25718871 on Build GRCh38
rs372521145 Chr.3: 25718937-25718937 on Build GRCh38
rs545085476 Chr.3: 25719247-25719247 on Build GRCh38
rs909916611 Chr.3: 25719254-25719254 on Build GRCh38
rs566219030 Chr.3: 25718960-25718964 on Build GRCh38
rs769165883 Chr.3: 25719129-25719129 on Build GRCh38
rs939615327 Chr.3: 25718895-25718895 on Build GRCh38
rs966776508 Chr.3: 25719076-25719076 on Build GRCh38

View Full Product Details