Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25789842-25790342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGLY1
Entrez Gene ID
55768
Gene Name
N-glycanase 1
Gene Aliases
CDDG, CDG1V, PNG1, PNGase
UniGene
Hs.368960
Gene Chromosome Location
Chr.3: 25718944-25790039 on Build GRCh38
Transcripts
NM_001145294.1 NM_001145391.1 NM_017897.2 NR_026937.1 XM_006713216.3 XM_006713217.3 XM_017006713.1 XM_017006714.1

Gene Symbol
OXSM
Entrez Gene ID
54995
Gene Name
3-oxoacyl-ACP synthase, mitochondrial
Gene Aliases
FASN2D, KASI, KS
UniGene
Hs.55781
Gene Chromosome Location
Chr.3: 25783661-25794534 on Build GRCh38
Transcripts
NM_001145294.1 NM_001145391.1 NM_017897.2 NR_026937.1 XM_006713216.3 XM_006713217.3 XM_017006713.1 XM_017006714.1

SNP Detail

SNP ID Chromosome Location
rs902373922 Chr.3: 25789951-25789951 on Build GRCh38
rs550498526 Chr.3: 25790177-25790177 on Build GRCh38
rs756879471 Chr.3: 25789877-25789877 on Build GRCh38
rs757901930 Chr.3: 25790019-25790019 on Build GRCh38
rs148981591 Chr.3: 25790133-25790133 on Build GRCh38
rs571573768 Chr.3: 25789950-25789950 on Build GRCh38
rs879134148 Chr.3: 25790011-25790011 on Build GRCh38
rs879671493 Chr.3: 25790224-25790224 on Build GRCh38
rs925059292 Chr.3: 25790099-25790099 on Build GRCh38
rs143057630 Chr.3: 25790162-25790162 on Build GRCh38
rs894989793 Chr.3: 25790297-25790297 on Build GRCh38
rs1041681096 Chr.3: 25790042-25790042 on Build GRCh38
rs1044678436 Chr.3: 25789985-25789985 on Build GRCh38
rs539878359 Chr.3: 25790079-25790079 on Build GRCh38
rs565267554 Chr.3: 25790123-25790123 on Build GRCh38
rs990973486 Chr.3: 25789975-25789975 on Build GRCh38
rs1051241557 Chr.3: 25790115-25790115 on Build GRCh38
rs951387835 Chr.3: 25790068-25790068 on Build GRCh38
rs911402949 Chr.3: 25790106-25790107 on Build GRCh38
rs41285079 Chr.3: 25790022-25790022 on Build GRCh38
rs969176369 Chr.3: 25790168-25790168 on Build GRCh38
rs115286377 Chr.3: 25789978-25789978 on Build GRCh38
rs1054502861 Chr.3: 25789919-25789919 on Build GRCh38
rs946028403 Chr.3: 25790024-25790024 on Build GRCh38
rs138602260 Chr.3: 25790185-25790185 on Build GRCh38
rs41285081 Chr.3: 25790103-25790103 on Build GRCh38
rs758734505 Chr.3: 25789893-25789893 on Build GRCh38
rs987787963 Chr.3: 25790226-25790226 on Build GRCh38
rs1051248980 Chr.3: 25789878-25789878 on Build GRCh38
rs1020800264 Chr.3: 25789946-25789946 on Build GRCh38
rs951289947 Chr.3: 25790152-25790152 on Build GRCh38
rs746302307 Chr.3: 25789882-25789882 on Build GRCh38
rs184003285 Chr.3: 25790041-25790041 on Build GRCh38
rs1003354346 Chr.3: 25789960-25789960 on Build GRCh38
rs371120025 Chr.3: 25790021-25790021 on Build GRCh38
rs1053756521 Chr.3: 25789908-25789908 on Build GRCh38
rs754825988 Chr.3: 25789890-25789890 on Build GRCh38
rs1024887800 Chr.3: 25790166-25790166 on Build GRCh38
rs980485721 Chr.3: 25790091-25790091 on Build GRCh38
rs947924355 Chr.3: 25790084-25790084 on Build GRCh38
rs987003614 Chr.3: 25790014-25790014 on Build GRCh38
rs749930349 Chr.3: 25790015-25790015 on Build GRCh38
rs754218849 Chr.3: 25789968-25789968 on Build GRCh38
rs778526036 Chr.3: 25789881-25789881 on Build GRCh38
rs1010832013 Chr.3: 25789936-25789936 on Build GRCh38
rs191935507 Chr.3: 25789920-25789920 on Build GRCh38
rs903521296 Chr.3: 25789990-25789990 on Build GRCh38
rs1007101669 Chr.3: 25790128-25790128 on Build GRCh38
rs149861362 Chr.3: 25790111-25790111 on Build GRCh38
rs959186431 Chr.3: 25789971-25789971 on Build GRCh38
rs879200762 Chr.3: 25789981-25789981 on Build GRCh38
rs991981814 Chr.3: 25790072-25790072 on Build GRCh38
rs554487596 Chr.3: 25789986-25789986 on Build GRCh38
rs780587506 Chr.3: 25789899-25789899 on Build GRCh38
rs368273574 Chr.3: 25789934-25789934 on Build GRCh38
rs559994263 Chr.3: 25790081-25790081 on Build GRCh38
rs995490785 Chr.3: 25790136-25790136 on Build GRCh38
rs145362063 Chr.3: 25789998-25789998 on Build GRCh38
rs567678101 Chr.3: 25790159-25790159 on Build GRCh38
rs747465435 Chr.3: 25789912-25789912 on Build GRCh38
rs531419952 Chr.3: 25790245-25790245 on Build GRCh38
rs573573311 Chr.3: 25790086-25790086 on Build GRCh38
rs552500579 Chr.3: 25790101-25790101 on Build GRCh38
rs918817635 Chr.3: 25790000-25790000 on Build GRCh38
rs112800537 Chr.3: 25790125-25790125 on Build GRCh38
rs1026084048 Chr.3: 25790145-25790145 on Build GRCh38
rs879509829 Chr.3: 25790109-25790109 on Build GRCh38
rs147738119 Chr.3: 25790045-25790045 on Build GRCh38
rs551840163 Chr.3: 25790122-25790122 on Build GRCh38
rs536965124 Chr.3: 25790004-25790004 on Build GRCh38
rs1010377137 Chr.3: 25790009-25790009 on Build GRCh38
rs962699560 Chr.3: 25790033-25790033 on Build GRCh38
rs557614930 Chr.3: 25790049-25790049 on Build GRCh38
rs370445586 Chr.3: 25790251-25790251 on Build GRCh38
rs974007811 Chr.3: 25789964-25789964 on Build GRCh38
rs745950189 Chr.3: 25790254-25790254 on Build GRCh38
rs369325985 Chr.3: 25789926-25789926 on Build GRCh38
rs577557680 Chr.3: 25790067-25790067 on Build GRCh38
rs781086315 Chr.3: 25790078-25790078 on Build GRCh38
rs1006985493 Chr.3: 25790020-25790020 on Build GRCh38
rs918536087 Chr.3: 25789965-25789965 on Build GRCh38
rs777929368 Chr.3: 25790210-25790210 on Build GRCh38
rs980601785 Chr.3: 25790201-25790201 on Build GRCh38
rs755476524 Chr.3: 25790131-25790131 on Build GRCh38
rs985437616 Chr.3: 25789930-25789930 on Build GRCh38
rs142618134 Chr.3: 25790108-25790108 on Build GRCh38
rs1034801940 Chr.3: 25789970-25789970 on Build GRCh38
rs955455649 Chr.3: 25789903-25789903 on Build GRCh38
rs555087286 Chr.3: 25790274-25790274 on Build GRCh38
rs1056265892 Chr.3: 25790048-25790048 on Build GRCh38
rs947824898 Chr.3: 25789982-25789982 on Build GRCh38
rs917751158 Chr.3: 25790075-25790075 on Build GRCh38
rs764322840 Chr.3: 25789935-25789935 on Build GRCh38
rs964531890 Chr.3: 25790107-25790109 on Build GRCh38
rs892073605 Chr.3: 25790005-25790005 on Build GRCh38
rs993865025 Chr.3: 25790092-25790092 on Build GRCh38
rs376165311 Chr.3: 25790057-25790078 on Build GRCh38

View Full Product Details