Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25794375-25794889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXSM
Entrez Gene ID
54995
Gene Name
3-oxoacyl-ACP synthase, mitochondrial
Gene Aliases
FASN2D, KASI, KS
UniGene
Hs.55781
Gene Chromosome Location
Chr.3: 25783661-25794534 on Build GRCh38
Transcripts
NM_001145391.1 NM_017897.2 NR_026937.1 XM_006713216.3 XM_006713217.3 XM_017006713.1 XM_017006714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4710637 c.1268A>G p.N423S Chr.3: 25794382-25794382 on Build GRCh38
7128164 c.1305G>C p.E435D Chr.3: 25794419-25794419 on Build GRCh38
7128163 c.1296G>T p.W432C Chr.3: 25794410-25794410 on Build GRCh38
125092 c.1328A>G p.N443S Chr.3: 25794442-25794442 on Build GRCh38
1186217 c.1365T>G p.C455W Chr.3: 25794479-25794479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940432080 Chr.3: 25794790-25794790 on Build GRCh38
rs772885075 Chr.3: 25794472-25794472 on Build GRCh38
rs758217012 Chr.3: 25794406-25794406 on Build GRCh38
rs776917837 Chr.3: 25794470-25794470 on Build GRCh38
rs550612600 Chr.3: 25794771-25794771 on Build GRCh38
rs757480318 Chr.3: 25794521-25794521 on Build GRCh38
rs1057076981 Chr.3: 25794529-25794529 on Build GRCh38
rs922949191 Chr.3: 25794823-25794823 on Build GRCh38
rs9855280 Chr.3: 25794772-25794772 on Build GRCh38
rs747709870 Chr.3: 25794442-25794442 on Build GRCh38
rs953672806 Chr.3: 25794463-25794463 on Build GRCh38
rs976444327 Chr.3: 25794407-25794407 on Build GRCh38
rs958604497 Chr.3: 25794757-25794757 on Build GRCh38
rs9851096 Chr.3: 25794492-25794492 on Build GRCh38
rs764955585 Chr.3: 25794429-25794429 on Build GRCh38
rs752823647 Chr.3: 25794637-25794637 on Build GRCh38
rs9855105 Chr.3: 25794691-25794691 on Build GRCh38
rs571935334 Chr.3: 25794674-25794674 on Build GRCh38
rs192973334 Chr.3: 25794763-25794763 on Build GRCh38
rs761975536 Chr.3: 25794526-25794527 on Build GRCh38
rs544600707 Chr.3: 25794604-25794604 on Build GRCh38
rs769249202 Chr.3: 25794445-25794445 on Build GRCh38
rs535569407 Chr.3: 25794851-25794851 on Build GRCh38
rs779194093 Chr.3: 25794526-25794526 on Build GRCh38
rs376248677 Chr.3: 25794487-25794487 on Build GRCh38
rs749429120 Chr.3: 25794404-25794404 on Build GRCh38
rs140533100 Chr.3: 25794692-25794692 on Build GRCh38
rs972103445 Chr.3: 25794494-25794494 on Build GRCh38
rs1036442589 Chr.3: 25794580-25794580 on Build GRCh38
rs183033687 Chr.3: 25794602-25794602 on Build GRCh38
rs753206281 Chr.3: 25794505-25794505 on Build GRCh38
rs933026136 Chr.3: 25794831-25794831 on Build GRCh38
rs953723084 Chr.3: 25794666-25794666 on Build GRCh38
rs764200999 Chr.3: 25794509-25794509 on Build GRCh38
rs997943697 Chr.3: 25794828-25794828 on Build GRCh38
rs934454101 Chr.3: 25794777-25794777 on Build GRCh38
rs1001016353 Chr.3: 25794615-25794615 on Build GRCh38
rs189519597 Chr.3: 25794839-25794839 on Build GRCh38
rs956824330 Chr.3: 25794664-25794664 on Build GRCh38
rs558284237 Chr.3: 25794641-25794641 on Build GRCh38
rs986665180 Chr.3: 25794477-25794477 on Build GRCh38
rs781134895 Chr.3: 25794536-25794536 on Build GRCh38
rs1052844801 Chr.3: 25794614-25794614 on Build GRCh38
rs998713347 Chr.3: 25794612-25794612 on Build GRCh38
rs576160451 Chr.3: 25794502-25794502 on Build GRCh38
rs184637814 Chr.3: 25794774-25794774 on Build GRCh38
rs992752524 Chr.3: 25794740-25794740 on Build GRCh38
rs9831075 Chr.3: 25794532-25794532 on Build GRCh38
rs374473365 Chr.3: 25794401-25794401 on Build GRCh38
rs925892987 Chr.3: 25794775-25794775 on Build GRCh38
rs61737038 Chr.3: 25794491-25794491 on Build GRCh38
rs919318019 Chr.3: 25794719-25794719 on Build GRCh38
rs900883978 Chr.3: 25794806-25794806 on Build GRCh38
rs561555508 Chr.3: 25794761-25794761 on Build GRCh38
rs370940750 Chr.3: 25794460-25794460 on Build GRCh38
rs948670904 Chr.3: 25794587-25794587 on Build GRCh38
rs9834955 Chr.3: 25794597-25794597 on Build GRCh38
rs998190496 Chr.3: 25794596-25794596 on Build GRCh38
rs760869142 Chr.3: 25794508-25794508 on Build GRCh38
rs746808625 Chr.3: 25794423-25794423 on Build GRCh38
rs1027995616 Chr.3: 25794659-25794659 on Build GRCh38
rs771272145 Chr.3: 25794738-25794738 on Build GRCh38
rs377612765 Chr.3: 25794403-25794403 on Build GRCh38
rs370581832 Chr.3: 25794562-25794562 on Build GRCh38
rs997829170 Chr.3: 25794653-25794653 on Build GRCh38
rs541878572 Chr.3: 25794753-25794753 on Build GRCh38
rs768648089 Chr.3: 25794426-25794426 on Build GRCh38
rs535486052 Chr.3: 25794575-25794575 on Build GRCh38
rs757133255 Chr.3: 25794414-25794414 on Build GRCh38
rs762449341 Chr.3: 25794475-25794475 on Build GRCh38
rs528068366 Chr.3: 25794716-25794716 on Build GRCh38
rs779658140 Chr.3: 25794421-25794421 on Build GRCh38
rs940420501 Chr.3: 25794559-25794559 on Build GRCh38
rs555704339 Chr.3: 25794855-25794855 on Build GRCh38
rs970723769 Chr.3: 25794680-25794680 on Build GRCh38
rs769074724 Chr.3: 25794822-25794822 on Build GRCh38
rs754102632 Chr.3: 25794514-25794514 on Build GRCh38
rs900936376 Chr.3: 25794633-25794633 on Build GRCh38
rs546668212 Chr.3: 25794794-25794794 on Build GRCh38
rs187322405 Chr.3: 25794679-25794679 on Build GRCh38
rs200044949 Chr.3: 25794534-25794534 on Build GRCh38

View Full Product Details