Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32449320-32449815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM7
Entrez Gene ID
112616
Gene Name
CKLF like MARVEL transmembrane domain containing 7
Gene Aliases
CKLFSF7
UniGene
Hs.440494
Gene Chromosome Location
Chr.3: 32391671-32455528 on Build GRCh38
Transcripts
NM_138410.3 NM_181472.2 XM_011533319.2 XM_017005646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1730062 c.427G>A p.G143R Chr.3: 32449547-32449547 on Build GRCh38
1485092 c.414C>T p.S138S Chr.3: 32449534-32449534 on Build GRCh38
1043651 c.432G>T p.A144A Chr.3: 32449552-32449552 on Build GRCh38
5934726 c.370C>T p.L124F Chr.3: 32449490-32449490 on Build GRCh38
1422398 c.421G>A p.V141I Chr.3: 32449541-32449541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897543552 Chr.3: 32449388-32449388 on Build GRCh38
rs754987313 Chr.3: 32449484-32449484 on Build GRCh38
rs898763934 Chr.3: 32449712-32449712 on Build GRCh38
rs916190697 Chr.3: 32449638-32449638 on Build GRCh38
rs751659931 Chr.3: 32449475-32449475 on Build GRCh38
rs746579801 Chr.3: 32449534-32449534 on Build GRCh38
rs937529454 Chr.3: 32449693-32449693 on Build GRCh38
rs994490887 Chr.3: 32449474-32449474 on Build GRCh38
rs762278617 Chr.3: 32449432-32449432 on Build GRCh38
rs772081995 Chr.3: 32449409-32449409 on Build GRCh38
rs974597747 Chr.3: 32449588-32449588 on Build GRCh38
rs772170038 Chr.3: 32449530-32449530 on Build GRCh38
rs1027284876 Chr.3: 32449401-32449401 on Build GRCh38
rs988826246 Chr.3: 32449633-32449633 on Build GRCh38
rs752028307 Chr.3: 32449579-32449579 on Build GRCh38
rs930204650 Chr.3: 32449726-32449726 on Build GRCh38
rs778868737 Chr.3: 32449525-32449525 on Build GRCh38
rs748310038 Chr.3: 32449495-32449495 on Build GRCh38
rs750727116 Chr.3: 32449449-32449449 on Build GRCh38
rs375912380 Chr.3: 32449551-32449551 on Build GRCh38
rs748235298 Chr.3: 32449783-32449783 on Build GRCh38
rs1049319025 Chr.3: 32449691-32449691 on Build GRCh38
rs950117250 Chr.3: 32449664-32449664 on Build GRCh38
rs758910208 Chr.3: 32449467-32449467 on Build GRCh38
rs767477658 Chr.3: 32449601-32449601 on Build GRCh38
rs780093363 Chr.3: 32449532-32449532 on Build GRCh38
rs2304593 Chr.3: 32449592-32449592 on Build GRCh38
rs200478443 Chr.3: 32449416-32449416 on Build GRCh38
rs373771542 Chr.3: 32449546-32449546 on Build GRCh38
rs1055953649 Chr.3: 32449603-32449603 on Build GRCh38
rs965850425 Chr.3: 32449446-32449446 on Build GRCh38
rs975970267 Chr.3: 32449451-32449451 on Build GRCh38
rs745596835 Chr.3: 32449526-32449526 on Build GRCh38
rs1038524163 Chr.3: 32449705-32449705 on Build GRCh38
rs913212935 Chr.3: 32449660-32449660 on Build GRCh38
rs765515515 Chr.3: 32449447-32449447 on Build GRCh38
rs754110333 Chr.3: 32449351-32449351 on Build GRCh38
rs763253078 Chr.3: 32449558-32449558 on Build GRCh38
rs553220180 Chr.3: 32449768-32449768 on Build GRCh38
rs149834090 Chr.3: 32449400-32449400 on Build GRCh38
rs575580084 Chr.3: 32449364-32449364 on Build GRCh38
rs766830014 Chr.3: 32449469-32449469 on Build GRCh38
rs200156694 Chr.3: 32449417-32449417 on Build GRCh38
rs1017520879 Chr.3: 32449734-32449734 on Build GRCh38
rs897989697 Chr.3: 32449666-32449666 on Build GRCh38
rs761359854 Chr.3: 32449547-32449547 on Build GRCh38
rs957262889 Chr.3: 32449599-32449599 on Build GRCh38
rs199711756 Chr.3: 32449578-32449578 on Build GRCh38
rs1007477705 Chr.3: 32449405-32449405 on Build GRCh38
rs2304594 Chr.3: 32449402-32449402 on Build GRCh38
rs532551099 Chr.3: 32449421-32449421 on Build GRCh38
rs781514375 Chr.3: 32449486-32449486 on Build GRCh38
rs1047387502 Chr.3: 32449766-32449766 on Build GRCh38
rs536686927 Chr.3: 32449616-32449616 on Build GRCh38
rs950085921 Chr.3: 32449350-32449350 on Build GRCh38
rs950489001 Chr.3: 32449488-32449488 on Build GRCh38
rs188774595 Chr.3: 32449382-32449382 on Build GRCh38
rs191189841 Chr.3: 32449669-32449669 on Build GRCh38
rs369659470 Chr.3: 32449552-32449552 on Build GRCh38
rs776126941 Chr.3: 32449427-32449427 on Build GRCh38
rs113870200 Chr.3: 32449719-32449719 on Build GRCh38
rs950313582 Chr.3: 32449650-32449650 on Build GRCh38
rs768021987 Chr.3: 32449545-32449545 on Build GRCh38
rs182240234 Chr.3: 32449689-32449689 on Build GRCh38
rs534891118 Chr.3: 32449745-32449745 on Build GRCh38

View Full Product Details