Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32685061-32685552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT10
Entrez Gene ID
25904
Gene Name
CCR4-NOT transcription complex subunit 10
Gene Aliases
-
UniGene
Hs.444851
Gene Chromosome Location
Chr.3: 32685145-32773875 on Build GRCh38
Transcripts
NM_001256741.1 NM_015442.2 NR_046352.1 XM_017006109.1 XM_017006110.1 XM_017006111.1 XM_017006112.1

SNP Detail

SNP ID Chromosome Location
rs964311771 Chr.3: 32685377-32685377 on Build GRCh38
rs1025602805 Chr.3: 32685288-32685288 on Build GRCh38
rs929547150 Chr.3: 32685347-32685347 on Build GRCh38
rs901564906 Chr.3: 32685444-32685444 on Build GRCh38
rs1045636516 Chr.3: 32685251-32685251 on Build GRCh38
rs1017037628 Chr.3: 32685160-32685160 on Build GRCh38
rs925707032 Chr.3: 32685215-32685215 on Build GRCh38
rs925096895 Chr.3: 32685280-32685280 on Build GRCh38
rs965748010 Chr.3: 32685198-32685198 on Build GRCh38
rs201048537 Chr.3: 32685244-32685244 on Build GRCh38
rs949938787 Chr.3: 32685250-32685250 on Build GRCh38
rs367803890 Chr.3: 32685467-32685467 on Build GRCh38
rs935776698 Chr.3: 32685218-32685218 on Build GRCh38
rs898103033 Chr.3: 32685341-32685341 on Build GRCh38
rs988974914 Chr.3: 32685394-32685394 on Build GRCh38
rs530900901 Chr.3: 32685166-32685166 on Build GRCh38
rs1017319769 Chr.3: 32685440-32685440 on Build GRCh38
rs190569971 Chr.3: 32685328-32685328 on Build GRCh38
rs925578535 Chr.3: 32685091-32685091 on Build GRCh38
rs761881240 Chr.3: 32685477-32685477 on Build GRCh38
rs567331007 Chr.3: 32685282-32685282 on Build GRCh38
rs555463578 Chr.3: 32685482-32685482 on Build GRCh38
rs936055767 Chr.3: 32685105-32685105 on Build GRCh38
rs546910969 Chr.3: 32685318-32685318 on Build GRCh38
rs111289773 Chr.3: 32685165-32685165 on Build GRCh38
rs569777572 Chr.3: 32685364-32685364 on Build GRCh38
rs151040461 Chr.3: 32685241-32685242 on Build GRCh38
rs528432427 Chr.3: 32685313-32685313 on Build GRCh38
rs544855167 Chr.3: 32685155-32685155 on Build GRCh38
rs1007257413 Chr.3: 32685178-32685178 on Build GRCh38
rs957140242 Chr.3: 32685393-32685393 on Build GRCh38
rs763128754 Chr.3: 32685357-32685357 on Build GRCh38
rs974495096 Chr.3: 32685088-32685088 on Build GRCh38
rs981097066 Chr.3: 32685418-32685418 on Build GRCh38
rs766217872 Chr.3: 32685480-32685480 on Build GRCh38
rs1001028633 Chr.3: 32685378-32685378 on Build GRCh38
rs898174030 Chr.3: 32685257-32685257 on Build GRCh38
rs113437644 Chr.3: 32685266-32685266 on Build GRCh38
rs539252739 Chr.3: 32685334-32685334 on Build GRCh38
rs930041841 Chr.3: 32685533-32685533 on Build GRCh38
rs1032955484 Chr.3: 32685382-32685382 on Build GRCh38
rs950003882 Chr.3: 32685138-32685138 on Build GRCh38
rs1045501589 Chr.3: 32685143-32685143 on Build GRCh38
rs563839511 Chr.3: 32685163-32685163 on Build GRCh38
rs533185748 Chr.3: 32685084-32685084 on Build GRCh38
rs967010692 Chr.3: 32685259-32685264 on Build GRCh38
rs964218633 Chr.3: 32685205-32685205 on Build GRCh38
rs557261744 Chr.3: 32685359-32685359 on Build GRCh38
rs905713828 Chr.3: 32685146-32685146 on Build GRCh38
rs1018417173 Chr.3: 32685200-32685200 on Build GRCh38
rs1053221010 Chr.3: 32685117-32685117 on Build GRCh38
rs559654483 Chr.3: 32685083-32685083 on Build GRCh38
rs950094670 Chr.3: 32685323-32685323 on Build GRCh38
rs549405304 Chr.3: 32685187-32685187 on Build GRCh38
rs367620776 Chr.3: 32685157-32685157 on Build GRCh38
rs993701831 Chr.3: 32685165-32685165 on Build GRCh38
rs182909795 Chr.3: 32685460-32685460 on Build GRCh38
rs987052658 Chr.3: 32685197-32685197 on Build GRCh38
rs988486045 Chr.3: 32685222-32685222 on Build GRCh38
rs974466942 Chr.3: 32685212-32685212 on Build GRCh38
rs905682188 Chr.3: 32685255-32685255 on Build GRCh38
rs898574171 Chr.3: 32685281-32685281 on Build GRCh38
rs912872342 Chr.3: 32685402-32685402 on Build GRCh38
rs765512899 Chr.3: 32685169-32685169 on Build GRCh38
rs1012569676 Chr.3: 32685491-32685491 on Build GRCh38
rs891697109 Chr.3: 32685128-32685128 on Build GRCh38
rs1014634056 Chr.3: 32685153-32685153 on Build GRCh38
rs751356590 Chr.3: 32685494-32685494 on Build GRCh38
rs937295909 Chr.3: 32685449-32685449 on Build GRCh38
rs919758518 Chr.3: 32685470-32685470 on Build GRCh38
rs764289777 Chr.3: 32685421-32685421 on Build GRCh38
rs1017264521 Chr.3: 32685193-32685193 on Build GRCh38
rs971056056 Chr.3: 32685404-32685404 on Build GRCh38
rs1026101026 Chr.3: 32685167-32685167 on Build GRCh38
rs935698647 Chr.3: 32685102-32685102 on Build GRCh38
rs915949382 Chr.3: 32685317-32685317 on Build GRCh38
rs7620163 Chr.3: 32685401-32685401 on Build GRCh38
rs1007643511 Chr.3: 32685416-32685416 on Build GRCh38
rs912893039 Chr.3: 32685235-32685235 on Build GRCh38

View Full Product Details