Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33400154-33400442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL2
Entrez Gene ID
25827
Gene Name
F-box and leucine rich repeat protein 2
Gene Aliases
FBL2, FBL3
UniGene
Hs.475872
Gene Chromosome Location
Chr.3: 33277422-33422371 on Build GRCh38
Transcripts
NM_001128160.1 NM_001128161.1 NM_014517.4

Gene Symbol
UBP1
Entrez Gene ID
7342
Gene Name
upstream binding protein 1 (LBP-1a)
Gene Aliases
LBP-1B, LBP-1a, LBP1A, LBP1B
UniGene
Hs.729120
Gene Chromosome Location
Chr.3: 33388336-33440405 on Build GRCh38
Transcripts
NM_001128160.1 NM_001128161.1 NM_014517.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633124 c.1099T>G p.S367A Chr.3: 33400270-33400270 on Build GRCh38
4924640 c.1105A>G p.T369A Chr.3: 33400264-33400264 on Build GRCh38
4451159 c.1088A>T p.Q363L Chr.3: 33400281-33400281 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778346234 Chr.3: 33400264-33400264 on Build GRCh38
rs146301974 Chr.3: 33400372-33400372 on Build GRCh38
rs534641104 Chr.3: 33400343-33400343 on Build GRCh38
rs747569491 Chr.3: 33400370-33400370 on Build GRCh38
rs147751266 Chr.3: 33400262-33400262 on Build GRCh38
rs754922328 Chr.3: 33400199-33400199 on Build GRCh38
rs755927335 Chr.3: 33400251-33400251 on Build GRCh38
rs775504659 Chr.3: 33400290-33400291 on Build GRCh38
rs760528313 Chr.3: 33400319-33400322 on Build GRCh38
rs891490902 Chr.3: 33400360-33400360 on Build GRCh38
rs904275370 Chr.3: 33400223-33400223 on Build GRCh38
rs769032827 Chr.3: 33400313-33400313 on Build GRCh38
rs1024574689 Chr.3: 33400376-33400376 on Build GRCh38
rs777499187 Chr.3: 33400260-33400260 on Build GRCh38
rs913263263 Chr.3: 33400417-33400417 on Build GRCh38
rs776050805 Chr.3: 33400286-33400286 on Build GRCh38
rs767570798 Chr.3: 33400214-33400214 on Build GRCh38
rs571183138 Chr.3: 33400359-33400359 on Build GRCh38
rs747611043 Chr.3: 33400384-33400384 on Build GRCh38
rs750550507 Chr.3: 33400184-33400184 on Build GRCh38
rs747535331 Chr.3: 33400303-33400303 on Build GRCh38
rs556651302 Chr.3: 33400388-33400388 on Build GRCh38
rs752380867 Chr.3: 33400244-33400244 on Build GRCh38
rs772510728 Chr.3: 33400273-33400273 on Build GRCh38
rs767160728 Chr.3: 33400182-33400183 on Build GRCh38
rs1007484196 Chr.3: 33400371-33400372 on Build GRCh38
rs189110180 Chr.3: 33400349-33400349 on Build GRCh38
rs745414199 Chr.3: 33400267-33400267 on Build GRCh38
rs377033790 Chr.3: 33400329-33400329 on Build GRCh38

View Full Product Details