Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36484791-36485312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAC
Entrez Gene ID
6769
Gene Name
SH3 and cysteine rich domain
Gene Aliases
STAC1
UniGene
Hs.56045
Gene Chromosome Location
Chr.3: 36380455-36548007 on Build GRCh38
Transcripts
NM_001292049.1 NM_003149.2 XM_011534036.2 XM_011534037.2 XM_011534038.2 XM_017007083.1 XM_017007084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936234 c.562A>T p.M188L Chr.3: 36485049-36485049 on Build GRCh38
6468855 c.492A>C p.P164P Chr.3: 36484979-36484979 on Build GRCh38
6791383 c.510C>A p.Y170* Chr.3: 36484997-36484997 on Build GRCh38
1043816 c.502C>T p.R168W Chr.3: 36484989-36484989 on Build GRCh38
5791378 c.506G>A p.R169H Chr.3: 36484993-36484993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764252513 Chr.3: 36485088-36485088 on Build GRCh38
rs764788554 Chr.3: 36485096-36485133 on Build GRCh38
rs947189221 Chr.3: 36485081-36485081 on Build GRCh38
rs768267819 Chr.3: 36485033-36485033 on Build GRCh38
rs1016697821 Chr.3: 36485261-36485261 on Build GRCh38
rs775060513 Chr.3: 36484952-36484952 on Build GRCh38
rs41276519 Chr.3: 36485276-36485276 on Build GRCh38
rs867789333 Chr.3: 36485015-36485015 on Build GRCh38
rs1057408830 Chr.3: 36485159-36485159 on Build GRCh38
rs1015599608 Chr.3: 36485200-36485200 on Build GRCh38
rs975864114 Chr.3: 36485257-36485257 on Build GRCh38
rs975520090 Chr.3: 36485277-36485277 on Build GRCh38
rs371990171 Chr.3: 36484842-36484842 on Build GRCh38
rs903353335 Chr.3: 36485132-36485132 on Build GRCh38
rs753033585 Chr.3: 36485078-36485078 on Build GRCh38
rs1027382883 Chr.3: 36484897-36484897 on Build GRCh38
rs368661092 Chr.3: 36485024-36485024 on Build GRCh38
rs756084428 Chr.3: 36485085-36485085 on Build GRCh38
rs952821040 Chr.3: 36485191-36485191 on Build GRCh38
rs7627803 Chr.3: 36484961-36484961 on Build GRCh38
rs541074547 Chr.3: 36484830-36484830 on Build GRCh38
rs1013574845 Chr.3: 36485167-36485167 on Build GRCh38
rs1041484001 Chr.3: 36485083-36485083 on Build GRCh38
rs377362858 Chr.3: 36484920-36484920 on Build GRCh38
rs894752504 Chr.3: 36485161-36485161 on Build GRCh38
rs1003006516 Chr.3: 36485149-36485149 on Build GRCh38
rs1051920712 Chr.3: 36484984-36484984 on Build GRCh38
rs750311311 Chr.3: 36485109-36485109 on Build GRCh38
rs1003163924 Chr.3: 36484876-36484876 on Build GRCh38
rs557504462 Chr.3: 36485146-36485146 on Build GRCh38
rs371869689 Chr.3: 36485076-36485076 on Build GRCh38
rs756277656 Chr.3: 36485007-36485007 on Build GRCh38
rs76557393 Chr.3: 36484852-36484852 on Build GRCh38
rs371786551 Chr.3: 36484967-36484967 on Build GRCh38
rs776401223 Chr.3: 36485037-36485037 on Build GRCh38
rs750804985 Chr.3: 36485104-36485104 on Build GRCh38
rs1012898105 Chr.3: 36484895-36484895 on Build GRCh38
rs200639094 Chr.3: 36485009-36485009 on Build GRCh38
rs761433029 Chr.3: 36484978-36484978 on Build GRCh38
rs142474647 Chr.3: 36484990-36484990 on Build GRCh38
rs769350072 Chr.3: 36485056-36485056 on Build GRCh38
rs566022574 Chr.3: 36485020-36485020 on Build GRCh38
rs552789257 Chr.3: 36484883-36484883 on Build GRCh38
rs771083134 Chr.3: 36484841-36484841 on Build GRCh38
rs74975922 Chr.3: 36484817-36484817 on Build GRCh38
rs961867293 Chr.3: 36485221-36485221 on Build GRCh38
rs763573396 Chr.3: 36485057-36485057 on Build GRCh38
rs778960489 Chr.3: 36484939-36484939 on Build GRCh38
rs952856881 Chr.3: 36484918-36484918 on Build GRCh38
rs570067392 Chr.3: 36485077-36485077 on Build GRCh38
rs760025141 Chr.3: 36485072-36485072 on Build GRCh38
rs766859384 Chr.3: 36484992-36484992 on Build GRCh38
rs746625339 Chr.3: 36485029-36485029 on Build GRCh38
rs747606645 Chr.3: 36485054-36485054 on Build GRCh38
rs866302876 Chr.3: 36485224-36485224 on Build GRCh38
rs1027023202 Chr.3: 36485179-36485179 on Build GRCh38
rs751004980 Chr.3: 36484989-36484989 on Build GRCh38
rs752699876 Chr.3: 36485006-36485006 on Build GRCh38
rs535475292 Chr.3: 36484995-36484995 on Build GRCh38
rs199583942 Chr.3: 36484988-36484988 on Build GRCh38
rs781514326 Chr.3: 36485000-36485000 on Build GRCh38
rs764068918 Chr.3: 36485227-36485227 on Build GRCh38
rs958973471 Chr.3: 36484894-36484894 on Build GRCh38
rs539278555 Chr.3: 36485068-36485068 on Build GRCh38
rs756729001 Chr.3: 36485059-36485060 on Build GRCh38
rs745640865 Chr.3: 36484946-36484946 on Build GRCh38
rs951147413 Chr.3: 36484862-36484862 on Build GRCh38
rs982822584 Chr.3: 36484919-36484919 on Build GRCh38
rs758131873 Chr.3: 36485021-36485021 on Build GRCh38
rs1004256643 Chr.3: 36485195-36485195 on Build GRCh38
rs765762372 Chr.3: 36485067-36485067 on Build GRCh38
rs1003457647 Chr.3: 36485154-36485155 on Build GRCh38
rs1005368924 Chr.3: 36485252-36485252 on Build GRCh38
rs141427741 Chr.3: 36484993-36484993 on Build GRCh38
rs192857646 Chr.3: 36484965-36484965 on Build GRCh38
rs771925257 Chr.3: 36484950-36484950 on Build GRCh38

View Full Product Details