Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37998827-37999330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VILL
Entrez Gene ID
50853
Gene Name
villin like
Gene Aliases
-
UniGene
Hs.103665
Gene Chromosome Location
Chr.3: 37987974-38007188 on Build GRCh38
Transcripts
NM_015873.3 XM_005265191.3 XM_005265192.4 XM_006713184.3 XM_011533770.2 XM_011533771.1 XM_011533773.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415369 c.1034G>A p.R345Q Chr.3: 37999003-37999003 on Build GRCh38
4601149 c.1062C>A p.N354K Chr.3: 37999031-37999031 on Build GRCh38
1683603 c.979_980insCCA p.T326_N327insT Chr.3: 37998948-37998949 on Build GRCh38
5696607 c.1081+2T>G p.? Chr.3: 37999052-37999052 on Build GRCh38
328713 c.974_975insACC p.Y325>* Chr.3: 37998943-37998944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879267767 Chr.3: 37998854-37998854 on Build GRCh38
rs1002897515 Chr.3: 37999100-37999100 on Build GRCh38
rs763915663 Chr.3: 37999301-37999301 on Build GRCh38
rs554089273 Chr.3: 37998922-37998922 on Build GRCh38
rs986671949 Chr.3: 37999166-37999166 on Build GRCh38
rs963148936 Chr.3: 37999055-37999055 on Build GRCh38
rs201408304 Chr.3: 37999294-37999294 on Build GRCh38
rs368748196 Chr.3: 37999060-37999060 on Build GRCh38
rs955646956 Chr.3: 37999174-37999174 on Build GRCh38
rs768996246 Chr.3: 37999063-37999063 on Build GRCh38
rs377611133 Chr.3: 37998974-37998974 on Build GRCh38
rs201859326 Chr.3: 37999031-37999031 on Build GRCh38
rs143137431 Chr.3: 37998943-37998944 on Build GRCh38
rs892084530 Chr.3: 37999145-37999145 on Build GRCh38
rs781191239 Chr.3: 37998956-37998956 on Build GRCh38
rs754031263 Chr.3: 37999014-37999014 on Build GRCh38
rs750513890 Chr.3: 37999026-37999026 on Build GRCh38
rs140527821 Chr.3: 37998972-37998972 on Build GRCh38
rs772558181 Chr.3: 37998995-37998995 on Build GRCh38
rs367971860 Chr.3: 37999301-37999301 on Build GRCh38
rs767807085 Chr.3: 37998904-37998904 on Build GRCh38
rs903623626 Chr.3: 37998881-37998881 on Build GRCh38
rs1021848478 Chr.3: 37999159-37999159 on Build GRCh38
rs774874979 Chr.3: 37998894-37998894 on Build GRCh38
rs774726124 Chr.3: 37998983-37998983 on Build GRCh38
rs769551941 Chr.3: 37998866-37998866 on Build GRCh38
rs763085266 Chr.3: 37999079-37999079 on Build GRCh38
rs953334898 Chr.3: 37999141-37999155 on Build GRCh38
rs890752947 Chr.3: 37998851-37998851 on Build GRCh38
rs781244242 Chr.3: 37999032-37999032 on Build GRCh38
rs974199164 Chr.3: 37999056-37999056 on Build GRCh38
rs748905788 Chr.3: 37999047-37999047 on Build GRCh38
rs780134429 Chr.3: 37999043-37999044 on Build GRCh38
rs1044471865 Chr.3: 37999085-37999085 on Build GRCh38
rs764257095 Chr.3: 37999004-37999004 on Build GRCh38
rs374387975 Chr.3: 37999290-37999290 on Build GRCh38
rs765350822 Chr.3: 37999066-37999066 on Build GRCh38
rs113261216 Chr.3: 37999135-37999135 on Build GRCh38
rs745364900 Chr.3: 37999058-37999058 on Build GRCh38
rs763532415 Chr.3: 37999070-37999089 on Build GRCh38
rs368878844 Chr.3: 37998948-37998949 on Build GRCh38
rs751119600 Chr.3: 37999292-37999293 on Build GRCh38
rs543954088 Chr.3: 37999126-37999126 on Build GRCh38
rs767582899 Chr.3: 37999082-37999082 on Build GRCh38
rs770277328 Chr.3: 37999065-37999070 on Build GRCh38
rs974659426 Chr.3: 37999250-37999250 on Build GRCh38
rs907271685 Chr.3: 37999089-37999089 on Build GRCh38
rs543366359 Chr.3: 37999293-37999293 on Build GRCh38
rs760999277 Chr.3: 37998998-37998998 on Build GRCh38
rs747925771 Chr.3: 37998957-37998957 on Build GRCh38
rs772644067 Chr.3: 37998870-37998870 on Build GRCh38
rs35288883 Chr.3: 37998933-37998934 on Build GRCh38
rs762294878 Chr.3: 37999057-37999076 on Build GRCh38
rs951920039 Chr.3: 37999280-37999280 on Build GRCh38
rs527791817 Chr.3: 37999300-37999300 on Build GRCh38
rs772611118 Chr.3: 37999059-37999059 on Build GRCh38
rs1039425635 Chr.3: 37998865-37998865 on Build GRCh38
rs768924719 Chr.3: 37999052-37999053 on Build GRCh38
rs747016116 Chr.3: 37999047-37999048 on Build GRCh38
rs759909935 Chr.3: 37998895-37998895 on Build GRCh38
rs371011172 Chr.3: 37998996-37998996 on Build GRCh38
rs752625921 Chr.3: 37999291-37999291 on Build GRCh38
rs35699577 Chr.3: 37998938-37998939 on Build GRCh38
rs199499422 Chr.3: 37999046-37999046 on Build GRCh38
rs770417972 Chr.3: 37998980-37998980 on Build GRCh38
rs893340125 Chr.3: 37999075-37999075 on Build GRCh38
rs761052403 Chr.3: 37999061-37999061 on Build GRCh38
rs372255809 Chr.3: 37999295-37999295 on Build GRCh38
rs548752403 Chr.3: 37999293-37999294 on Build GRCh38
rs367728851 Chr.3: 37999297-37999297 on Build GRCh38
rs766823686 Chr.3: 37998874-37998874 on Build GRCh38
rs752570500 Chr.3: 37999035-37999035 on Build GRCh38
rs748773755 Chr.3: 37998975-37998975 on Build GRCh38
rs11720984 Chr.3: 37999150-37999150 on Build GRCh38
rs764280163 Chr.3: 37998911-37998911 on Build GRCh38
rs948521689 Chr.3: 37999080-37999080 on Build GRCh38
rs200529311 Chr.3: 37999298-37999298 on Build GRCh38
rs111287866 Chr.3: 37999163-37999163 on Build GRCh38
rs574467623 Chr.3: 37999142-37999142 on Build GRCh38
rs766953257 Chr.3: 37999291-37999291 on Build GRCh38
rs1046014675 Chr.3: 37999309-37999309 on Build GRCh38
rs543566434 Chr.3: 37998915-37998915 on Build GRCh38
rs760103623 Chr.3: 37998992-37998992 on Build GRCh38
rs9882803 Chr.3: 37999296-37999296 on Build GRCh38
rs370937155 Chr.3: 37999310-37999310 on Build GRCh38
rs763136327 Chr.3: 37999299-37999299 on Build GRCh38
rs746896524 Chr.3: 37998938-37998938 on Build GRCh38
rs757285875 Chr.3: 37998949-37998949 on Build GRCh38
rs912518687 Chr.3: 37999181-37999181 on Build GRCh38
rs919762525 Chr.3: 37999052-37999052 on Build GRCh38
rs762491187 Chr.3: 37998873-37998873 on Build GRCh38
rs1000355847 Chr.3: 37998886-37998886 on Build GRCh38
rs920420075 Chr.3: 37999277-37999277 on Build GRCh38
rs750571116 Chr.3: 37999078-37999078 on Build GRCh38
rs559921239 Chr.3: 37999254-37999254 on Build GRCh38
rs765420513 Chr.3: 37999024-37999024 on Build GRCh38
rs1045816212 Chr.3: 37999003-37999003 on Build GRCh38
rs770486756 Chr.3: 37999049-37999049 on Build GRCh38
rs776851978 Chr.3: 37999065-37999065 on Build GRCh38
rs558581897 Chr.3: 37999057-37999057 on Build GRCh38
rs1008195913 Chr.3: 37999199-37999199 on Build GRCh38
rs755922307 Chr.3: 37999038-37999038 on Build GRCh38
rs776657201 Chr.3: 37999057-37999061 on Build GRCh38
rs753906141 Chr.3: 37998913-37998913 on Build GRCh38
rs756386762 Chr.3: 37998905-37998905 on Build GRCh38
rs1053513454 Chr.3: 37999133-37999133 on Build GRCh38
rs761801263 Chr.3: 37999021-37999021 on Build GRCh38
rs1018244206 Chr.3: 37999203-37999203 on Build GRCh38
rs942351407 Chr.3: 37999232-37999232 on Build GRCh38
rs773894881 Chr.3: 37999066-37999067 on Build GRCh38
rs141779050 Chr.3: 37998965-37998965 on Build GRCh38
rs971383050 Chr.3: 37998986-37998986 on Build GRCh38

View Full Product Details