Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38707176-38707678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN10A
Entrez Gene ID
6336
Gene Name
sodium voltage-gated channel alpha subunit 10
Gene Aliases
FEPS2, Nav1.8, PN3, SNS, hPN3
UniGene
Hs.250443
Gene Chromosome Location
Chr.3: 38697110-38794010 on Build GRCh38
Transcripts
NM_001293306.2 NM_001293307.2 NM_006514.3 XM_005265371.3 XM_011533993.2 XM_011533994.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
149361 c.4323A>G p.K1441K Chr.3: 38707342-38707342 on Build GRCh38
315009 c.4312G>C p.E1438Q Chr.3: 38707353-38707353 on Build GRCh38
6403251 c.4385T>C p.L1462P Chr.3: 38707280-38707280 on Build GRCh38
3823809 c.4334C>T p.A1445V Chr.3: 38707331-38707331 on Build GRCh38
5545746 c.4338G>A p.M1446I Chr.3: 38707327-38707327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013375023 Chr.3: 38707607-38707607 on Build GRCh38
rs369399424 Chr.3: 38707286-38707286 on Build GRCh38
rs557095386 Chr.3: 38707617-38707617 on Build GRCh38
rs370545434 Chr.3: 38707544-38707544 on Build GRCh38
rs762855694 Chr.3: 38707258-38707258 on Build GRCh38
rs747108606 Chr.3: 38707320-38707320 on Build GRCh38
rs369198406 Chr.3: 38707236-38707236 on Build GRCh38
rs148504955 Chr.3: 38707456-38707456 on Build GRCh38
rs6599248 Chr.3: 38707535-38707535 on Build GRCh38
rs1018251595 Chr.3: 38707458-38707458 on Build GRCh38
rs148619598 Chr.3: 38707287-38707287 on Build GRCh38
rs773975660 Chr.3: 38707234-38707234 on Build GRCh38
rs976789275 Chr.3: 38707481-38707481 on Build GRCh38
rs898671289 Chr.3: 38707271-38707271 on Build GRCh38
rs374834768 Chr.3: 38707396-38707396 on Build GRCh38
rs760479501 Chr.3: 38707359-38707365 on Build GRCh38
rs952203161 Chr.3: 38707436-38707436 on Build GRCh38
rs767087921 Chr.3: 38707278-38707278 on Build GRCh38
rs372714131 Chr.3: 38707644-38707644 on Build GRCh38
rs922307155 Chr.3: 38707297-38707297 on Build GRCh38
rs1049108247 Chr.3: 38707646-38707646 on Build GRCh38
rs986770847 Chr.3: 38707247-38707247 on Build GRCh38
rs564222571 Chr.3: 38707368-38707368 on Build GRCh38
rs867476480 Chr.3: 38707327-38707327 on Build GRCh38
rs757721860 Chr.3: 38707377-38707377 on Build GRCh38
rs759198338 Chr.3: 38707269-38707269 on Build GRCh38
rs202131976 Chr.3: 38707317-38707317 on Build GRCh38
rs191925752 Chr.3: 38707408-38707408 on Build GRCh38
rs1044908535 Chr.3: 38707549-38707549 on Build GRCh38
rs370311310 Chr.3: 38707419-38707419 on Build GRCh38
rs926499449 Chr.3: 38707575-38707575 on Build GRCh38
rs773243150 Chr.3: 38707333-38707333 on Build GRCh38
rs952253500 Chr.3: 38707207-38707207 on Build GRCh38
rs774223726 Chr.3: 38707413-38707413 on Build GRCh38
rs767179565 Chr.3: 38707362-38707362 on Build GRCh38
rs750188485 Chr.3: 38707430-38707430 on Build GRCh38
rs772430907 Chr.3: 38707306-38707306 on Build GRCh38
rs780387548 Chr.3: 38707311-38707311 on Build GRCh38
rs922253247 Chr.3: 38707499-38707499 on Build GRCh38
rs890028585 Chr.3: 38707219-38707219 on Build GRCh38
rs758791887 Chr.3: 38707386-38707386 on Build GRCh38
rs1023332527 Chr.3: 38707398-38707398 on Build GRCh38
rs757704367 Chr.3: 38707289-38707289 on Build GRCh38
rs766057731 Chr.3: 38707259-38707259 on Build GRCh38
rs956398444 Chr.3: 38707522-38707522 on Build GRCh38
rs566023334 Chr.3: 38707475-38707475 on Build GRCh38
rs541475904 Chr.3: 38707641-38707641 on Build GRCh38
rs112208946 Chr.3: 38707594-38707594 on Build GRCh38
rs765776366 Chr.3: 38707378-38707378 on Build GRCh38
rs760268052 Chr.3: 38707371-38707371 on Build GRCh38
rs759286277 Chr.3: 38707361-38707361 on Build GRCh38
rs915072822 Chr.3: 38707354-38707354 on Build GRCh38
rs373801645 Chr.3: 38707418-38707418 on Build GRCh38
rs535925070 Chr.3: 38707429-38707429 on Build GRCh38
rs78376096 Chr.3: 38707548-38707548 on Build GRCh38
rs760773412 Chr.3: 38707248-38707248 on Build GRCh38
rs6790627 Chr.3: 38707342-38707342 on Build GRCh38
rs202063632 Chr.3: 38707392-38707392 on Build GRCh38
rs779410370 Chr.3: 38707293-38707293 on Build GRCh38
rs898638606 Chr.3: 38707570-38707570 on Build GRCh38
rs934126804 Chr.3: 38707488-38707488 on Build GRCh38
rs750853186 Chr.3: 38707380-38707380 on Build GRCh38
rs969040601 Chr.3: 38707625-38707625 on Build GRCh38
rs370610736 Chr.3: 38707353-38707353 on Build GRCh38
rs772974795 Chr.3: 38707252-38707252 on Build GRCh38
rs775728693 Chr.3: 38707246-38707246 on Build GRCh38
rs772337925 Chr.3: 38707245-38707245 on Build GRCh38
rs910834522 Chr.3: 38707273-38707273 on Build GRCh38
rs373337849 Chr.3: 38707249-38707249 on Build GRCh38
rs76987412 Chr.3: 38707497-38707497 on Build GRCh38
rs573916611 Chr.3: 38707640-38707640 on Build GRCh38
rs561620672 Chr.3: 38707645-38707645 on Build GRCh38
rs376437999 Chr.3: 38707285-38707285 on Build GRCh38
rs771173856 Chr.3: 38707235-38707235 on Build GRCh38
rs746180135 Chr.3: 38707299-38707299 on Build GRCh38
rs751281512 Chr.3: 38707265-38707265 on Build GRCh38
rs1047255690 Chr.3: 38707583-38707583 on Build GRCh38
rs762798134 Chr.3: 38707338-38707338 on Build GRCh38
rs775472222 Chr.3: 38707296-38707314 on Build GRCh38
rs746081425 Chr.3: 38707241-38707241 on Build GRCh38
rs376080236 Chr.3: 38707399-38707399 on Build GRCh38
rs775186985 Chr.3: 38707422-38707422 on Build GRCh38
rs754423728 Chr.3: 38707376-38707376 on Build GRCh38
rs527290934 Chr.3: 38707329-38707329 on Build GRCh38
rs771005418 Chr.3: 38707543-38707543 on Build GRCh38
rs763504898 Chr.3: 38707375-38707375 on Build GRCh38
rs755934945 Chr.3: 38707455-38707455 on Build GRCh38
rs549987875 Chr.3: 38707407-38707407 on Build GRCh38

View Full Product Details