Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39280168-39280674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
UniGene
Hs.78913
Gene Chromosome Location
Chr.3: 39263494-39281735 on Build GRCh38
Transcripts
NM_001171171.1 NM_001171172.1 NM_001171174.1

SNP Detail

SNP ID Chromosome Location
rs927580232 Chr.3: 39280241-39280241 on Build GRCh38
rs989461359 Chr.3: 39280236-39280236 on Build GRCh38
rs871610 Chr.3: 39280376-39280376 on Build GRCh38
rs533569982 Chr.3: 39280377-39280377 on Build GRCh38
rs34571589 Chr.3: 39280263-39280263 on Build GRCh38
rs943193516 Chr.3: 39280612-39280612 on Build GRCh38
rs141811866 Chr.3: 39280265-39280265 on Build GRCh38
rs911943477 Chr.3: 39280546-39280546 on Build GRCh38
rs1020332289 Chr.3: 39280428-39280428 on Build GRCh38
rs772382630 Chr.3: 39280454-39280454 on Build GRCh38
rs970887622 Chr.3: 39280409-39280409 on Build GRCh38
rs570183126 Chr.3: 39280271-39280271 on Build GRCh38
rs996401080 Chr.3: 39280364-39280364 on Build GRCh38
rs937481983 Chr.3: 39280246-39280246 on Build GRCh38
rs113215386 Chr.3: 39280456-39280456 on Build GRCh38
rs748478927 Chr.3: 39280406-39280406 on Build GRCh38
rs987574643 Chr.3: 39280509-39280509 on Build GRCh38
rs996145204 Chr.3: 39280648-39280648 on Build GRCh38
rs1047486162 Chr.3: 39280508-39280508 on Build GRCh38
rs900442467 Chr.3: 39280361-39280361 on Build GRCh38
rs1052116227 Chr.3: 39280652-39280652 on Build GRCh38
rs1013986136 Chr.3: 39280383-39280383 on Build GRCh38
rs1002288800 Chr.3: 39280427-39280427 on Build GRCh38
rs927517224 Chr.3: 39280447-39280447 on Build GRCh38
rs36230798 Chr.3: 39280291-39280291 on Build GRCh38
rs35500272 Chr.3: 39280219-39280219 on Build GRCh38
rs147711841 Chr.3: 39280597-39280597 on Build GRCh38
rs1005904907 Chr.3: 39280357-39280357 on Build GRCh38
rs1034353416 Chr.3: 39280432-39280432 on Build GRCh38
rs1038903072 Chr.3: 39280633-39280633 on Build GRCh38
rs907644442 Chr.3: 39280575-39280575 on Build GRCh38
rs181634450 Chr.3: 39280489-39280489 on Build GRCh38
rs778253346 Chr.3: 39280469-39280469 on Build GRCh38
rs947391942 Chr.3: 39280211-39280211 on Build GRCh38
rs978802228 Chr.3: 39280443-39280443 on Build GRCh38
rs190464586 Chr.3: 39280220-39280220 on Build GRCh38
rs941659755 Chr.3: 39280606-39280606 on Build GRCh38
rs9813187 Chr.3: 39280279-39280279 on Build GRCh38
rs900412186 Chr.3: 39280621-39280621 on Build GRCh38
rs899049887 Chr.3: 39280347-39280347 on Build GRCh38
rs959015402 Chr.3: 39280452-39280452 on Build GRCh38
rs562547347 Chr.3: 39280598-39280598 on Build GRCh38
rs562796444 Chr.3: 39280634-39280658 on Build GRCh38
rs1038001009 Chr.3: 39280359-39280359 on Build GRCh38
rs954788376 Chr.3: 39280386-39280386 on Build GRCh38
rs1037403074 Chr.3: 39280611-39280611 on Build GRCh38
rs968660475 Chr.3: 39280440-39280440 on Build GRCh38
rs36230797 Chr.3: 39280314-39280314 on Build GRCh38

View Full Product Details