Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40487107-40487607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF619
Entrez Gene ID
285267
Gene Name
zinc finger protein 619
Gene Aliases
-
UniGene
Hs.407159
Gene Chromosome Location
Chr.3: 40477113-40490237 on Build GRCh38
Transcripts
NM_001145082.2 NM_001145083.1 NM_001145093.2 NR_026746.2 XM_006713112.2 XM_006713113.3 XM_011533607.1 XM_011533608.1 XM_017006225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6831822 c.1043G>T p.R348I Chr.3: 40487433-40487433 on Build GRCh38
3823844 c.948C>T p.I316I Chr.3: 40487506-40487506 on Build GRCh38
5829697 c.987A>C p.E329D Chr.3: 40487377-40487377 on Build GRCh38
1422984 c.973G>A p.E325K Chr.3: 40487531-40487531 on Build GRCh38
5556566 c.1037A>T p.H346L Chr.3: 40487595-40487595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776754875 Chr.3: 40487442-40487442 on Build GRCh38
rs143973123 Chr.3: 40487517-40487517 on Build GRCh38
rs139506802 Chr.3: 40487202-40487202 on Build GRCh38
rs146200388 Chr.3: 40487481-40487481 on Build GRCh38
rs936963452 Chr.3: 40487168-40487168 on Build GRCh38
rs758913680 Chr.3: 40487333-40487333 on Build GRCh38
rs779158381 Chr.3: 40487218-40487218 on Build GRCh38
rs758892326 Chr.3: 40487249-40487249 on Build GRCh38
rs776449714 Chr.3: 40487150-40487150 on Build GRCh38
rs760704632 Chr.3: 40487302-40487302 on Build GRCh38
rs141930111 Chr.3: 40487182-40487182 on Build GRCh38
rs765423437 Chr.3: 40487447-40487447 on Build GRCh38
rs764145216 Chr.3: 40487437-40487437 on Build GRCh38
rs370494897 Chr.3: 40487446-40487446 on Build GRCh38
rs368408720 Chr.3: 40487441-40487441 on Build GRCh38
rs753975021 Chr.3: 40487133-40487134 on Build GRCh38
rs372214247 Chr.3: 40487201-40487201 on Build GRCh38
rs755642277 Chr.3: 40487498-40487498 on Build GRCh38
rs754401840 Chr.3: 40487341-40487341 on Build GRCh38
rs761888195 Chr.3: 40487136-40487136 on Build GRCh38
rs766867565 Chr.3: 40487485-40487485 on Build GRCh38
rs752815641 Chr.3: 40487181-40487181 on Build GRCh38
rs780818740 Chr.3: 40487510-40487510 on Build GRCh38
rs907573795 Chr.3: 40487444-40487444 on Build GRCh38
rs748988679 Chr.3: 40487420-40487420 on Build GRCh38
rs868241531 Chr.3: 40487542-40487542 on Build GRCh38
rs201742151 Chr.3: 40487279-40487279 on Build GRCh38
rs74898403 Chr.3: 40487325-40487325 on Build GRCh38
rs764947871 Chr.3: 40487170-40487170 on Build GRCh38
rs759482596 Chr.3: 40487434-40487434 on Build GRCh38
rs372260574 Chr.3: 40487212-40487212 on Build GRCh38
rs752102729 Chr.3: 40487495-40487495 on Build GRCh38
rs780910779 Chr.3: 40487381-40487381 on Build GRCh38
rs141035606 Chr.3: 40487321-40487321 on Build GRCh38
rs374238547 Chr.3: 40487424-40487424 on Build GRCh38
rs201857902 Chr.3: 40487317-40487317 on Build GRCh38
rs144439722 Chr.3: 40487271-40487271 on Build GRCh38
rs866192697 Chr.3: 40487360-40487360 on Build GRCh38
rs772033534 Chr.3: 40487546-40487546 on Build GRCh38
rs138484130 Chr.3: 40487179-40487179 on Build GRCh38
rs146501133 Chr.3: 40487316-40487316 on Build GRCh38
rs769925282 Chr.3: 40487570-40487570 on Build GRCh38
rs752199384 Chr.3: 40487516-40487516 on Build GRCh38
rs762918923 Chr.3: 40487287-40487287 on Build GRCh38
rs760395198 Chr.3: 40487134-40487134 on Build GRCh38
rs750692749 Chr.3: 40487448-40487448 on Build GRCh38
rs773252909 Chr.3: 40487574-40487574 on Build GRCh38
rs576609937 Chr.3: 40487423-40487423 on Build GRCh38
rs572927429 Chr.3: 40487346-40487346 on Build GRCh38
rs138519036 Chr.3: 40487371-40487371 on Build GRCh38
rs747819613 Chr.3: 40487275-40487275 on Build GRCh38
rs748973097 Chr.3: 40487530-40487530 on Build GRCh38
rs184060445 Chr.3: 40487223-40487223 on Build GRCh38
rs527647687 Chr.3: 40487553-40487553 on Build GRCh38
rs76445796 Chr.3: 40487300-40487300 on Build GRCh38
rs1028714400 Chr.3: 40487395-40487395 on Build GRCh38
rs769372724 Chr.3: 40487419-40487419 on Build GRCh38
rs149623452 Chr.3: 40487373-40487373 on Build GRCh38
rs371071262 Chr.3: 40487382-40487382 on Build GRCh38
rs777250266 Chr.3: 40487526-40487526 on Build GRCh38
rs761664919 Chr.3: 40487156-40487156 on Build GRCh38
rs765921392 Chr.3: 40487541-40487541 on Build GRCh38
rs764163695 Chr.3: 40487313-40487313 on Build GRCh38
rs747525540 Chr.3: 40487352-40487352 on Build GRCh38
rs765262394 Chr.3: 40487198-40487198 on Build GRCh38
rs754483892 Chr.3: 40487355-40487355 on Build GRCh38
rs61740939 Chr.3: 40487555-40487555 on Build GRCh38
rs754088006 Chr.3: 40487209-40487209 on Build GRCh38
rs766278099 Chr.3: 40487291-40487291 on Build GRCh38
rs141182407 Chr.3: 40487175-40487175 on Build GRCh38
rs568095913 Chr.3: 40487379-40487379 on Build GRCh38
rs148232227 Chr.3: 40487476-40487476 on Build GRCh38
rs768460125 Chr.3: 40487142-40487142 on Build GRCh38
rs377170323 Chr.3: 40487255-40487255 on Build GRCh38
rs769619600 Chr.3: 40487276-40487276 on Build GRCh38
rs141502283 Chr.3: 40487337-40487337 on Build GRCh38
rs562250771 Chr.3: 40487545-40487545 on Build GRCh38
rs201349894 Chr.3: 40487256-40487256 on Build GRCh38

View Full Product Details