Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44584886-44585328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF660
Entrez Gene ID
285349
Gene Name
zinc finger protein 660
Gene Aliases
-
UniGene
Hs.733134
Gene Chromosome Location
Chr.3: 44584847-44599699 on Build GRCh38
Transcripts
NM_173658.2 XM_006713117.3 XM_011533622.2

SNP Detail

SNP ID Chromosome Location
rs892061711 Chr.3: 44585019-44585019 on Build GRCh38
rs948536493 Chr.3: 44584938-44584938 on Build GRCh38
rs996400352 Chr.3: 44585066-44585066 on Build GRCh38
rs147353774 Chr.3: 44585066-44585066 on Build GRCh38
rs1019769765 Chr.3: 44585296-44585296 on Build GRCh38
rs948565493 Chr.3: 44585171-44585171 on Build GRCh38
rs1020815599 Chr.3: 44585106-44585106 on Build GRCh38
rs1005490716 Chr.3: 44585023-44585023 on Build GRCh38
rs886883216 Chr.3: 44585259-44585259 on Build GRCh38
rs965503165 Chr.3: 44585297-44585297 on Build GRCh38
rs553693820 Chr.3: 44585281-44585281 on Build GRCh38
rs1028918728 Chr.3: 44585076-44585076 on Build GRCh38
rs991406583 Chr.3: 44585180-44585180 on Build GRCh38
rs568452751 Chr.3: 44585249-44585249 on Build GRCh38
rs937260210 Chr.3: 44584921-44584921 on Build GRCh38
rs528908891 Chr.3: 44585041-44585041 on Build GRCh38
rs985617812 Chr.3: 44585062-44585062 on Build GRCh38
rs768691427 Chr.3: 44584947-44584947 on Build GRCh38
rs937206457 Chr.3: 44585177-44585177 on Build GRCh38
rs928408774 Chr.3: 44585185-44585185 on Build GRCh38
rs1038248662 Chr.3: 44585306-44585306 on Build GRCh38
rs925788838 Chr.3: 44585175-44585175 on Build GRCh38
rs1052860643 Chr.3: 44585236-44585236 on Build GRCh38
rs575132958 Chr.3: 44584976-44584976 on Build GRCh38
rs925883258 Chr.3: 44584916-44584916 on Build GRCh38
rs549625145 Chr.3: 44584939-44584939 on Build GRCh38
rs1013972939 Chr.3: 44585001-44585001 on Build GRCh38
rs1007065546 Chr.3: 44585295-44585295 on Build GRCh38
rs1047292009 Chr.3: 44585002-44585002 on Build GRCh38
rs371538907 Chr.3: 44584972-44584972 on Build GRCh38
rs931314090 Chr.3: 44585159-44585159 on Build GRCh38
rs1048897089 Chr.3: 44584914-44584914 on Build GRCh38
rs1046799121 Chr.3: 44585252-44585252 on Build GRCh38
rs540135531 Chr.3: 44585120-44585120 on Build GRCh38
rs772559246 Chr.3: 44585237-44585237 on Build GRCh38
rs1042963549 Chr.3: 44584913-44584913 on Build GRCh38
rs954233556 Chr.3: 44585054-44585054 on Build GRCh38
rs1055438800 Chr.3: 44584956-44584956 on Build GRCh38
rs1016498089 Chr.3: 44585030-44585030 on Build GRCh38
rs541021550 Chr.3: 44585000-44585000 on Build GRCh38
rs916915764 Chr.3: 44585244-44585244 on Build GRCh38
rs886602818 Chr.3: 44585018-44585018 on Build GRCh38
rs965414736 Chr.3: 44585098-44585098 on Build GRCh38
rs987564123 Chr.3: 44585091-44585091 on Build GRCh38
rs534214869 Chr.3: 44585258-44585258 on Build GRCh38
rs978976591 Chr.3: 44585136-44585136 on Build GRCh38
rs985134463 Chr.3: 44585161-44585161 on Build GRCh38
rs938404293 Chr.3: 44585192-44585193 on Build GRCh38
rs189991558 Chr.3: 44585241-44585241 on Build GRCh38
rs954713722 Chr.3: 44585086-44585086 on Build GRCh38
rs909128915 Chr.3: 44585162-44585162 on Build GRCh38
rs941137445 Chr.3: 44585262-44585262 on Build GRCh38

View Full Product Details