Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44642180-44642694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF197
Entrez Gene ID
10168
Gene Name
zinc finger protein 197
Gene Aliases
D3S1363E, P18, VHLaK, ZKSCAN9, ZNF166, ZNF20, ZSCAN41
UniGene
Hs.157035
Gene Chromosome Location
Chr.3: 44625017-44648471 on Build GRCh38
Transcripts
XM_005264783.4 XM_017005495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1328219 c.1178C>G p.T393S Chr.3: 44642308-44642308 on Build GRCh38
5542085 c.1149C>T p.S383S Chr.3: 44642279-44642279 on Build GRCh38
6359185 c.1053G>C p.L351F Chr.3: 44642183-44642183 on Build GRCh38
308466 c.1339_1342delATCC p.I447fs*22 Chr.3: 44642469-44642472 on Build GRCh38
4460351 c.1162C>T p.H388Y Chr.3: 44642292-44642292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181483016 Chr.3: 44642554-44642554 on Build GRCh38
rs138953408 Chr.3: 44642389-44642389 on Build GRCh38
rs1036139007 Chr.3: 44642559-44642559 on Build GRCh38
rs769895593 Chr.3: 44642247-44642247 on Build GRCh38
rs775947229 Chr.3: 44642382-44642382 on Build GRCh38
rs368799335 Chr.3: 44642359-44642359 on Build GRCh38
rs771788668 Chr.3: 44642639-44642639 on Build GRCh38
rs752688610 Chr.3: 44642509-44642510 on Build GRCh38
rs1049048879 Chr.3: 44642591-44642591 on Build GRCh38
rs560022507 Chr.3: 44642668-44642668 on Build GRCh38
rs553813230 Chr.3: 44642539-44642539 on Build GRCh38
rs868506952 Chr.3: 44642599-44642599 on Build GRCh38
rs769259973 Chr.3: 44642496-44642496 on Build GRCh38
rs987103608 Chr.3: 44642343-44642343 on Build GRCh38
rs758393897 Chr.3: 44642220-44642220 on Build GRCh38
rs747167691 Chr.3: 44642597-44642597 on Build GRCh38
rs777335621 Chr.3: 44642621-44642621 on Build GRCh38
rs770307390 Chr.3: 44642619-44642619 on Build GRCh38
rs767276884 Chr.3: 44642538-44642538 on Build GRCh38
rs767294346 Chr.3: 44642469-44642472 on Build GRCh38
rs759190855 Chr.3: 44642637-44642637 on Build GRCh38
rs772508103 Chr.3: 44642291-44642291 on Build GRCh38
rs189212632 Chr.3: 44642295-44642295 on Build GRCh38
rs769211603 Chr.3: 44642504-44642504 on Build GRCh38
rs374520793 Chr.3: 44642211-44642211 on Build GRCh38
rs764790052 Chr.3: 44642311-44642311 on Build GRCh38
rs555744380 Chr.3: 44642551-44642551 on Build GRCh38
rs147994654 Chr.3: 44642297-44642297 on Build GRCh38
rs767344568 Chr.3: 44642444-44642444 on Build GRCh38
rs748619472 Chr.3: 44642501-44642501 on Build GRCh38
rs751328495 Chr.3: 44642407-44642407 on Build GRCh38
rs766335478 Chr.3: 44642565-44642565 on Build GRCh38
rs748655237 Chr.3: 44642618-44642618 on Build GRCh38
rs780604348 Chr.3: 44642492-44642492 on Build GRCh38
rs777015161 Chr.3: 44642497-44642497 on Build GRCh38
rs774846187 Chr.3: 44642278-44642278 on Build GRCh38
rs759364073 Chr.3: 44642311-44642317 on Build GRCh38
rs772742391 Chr.3: 44642401-44642401 on Build GRCh38
rs142871732 Chr.3: 44642404-44642404 on Build GRCh38
rs773997943 Chr.3: 44642635-44642635 on Build GRCh38
rs746888796 Chr.3: 44642245-44642245 on Build GRCh38
rs764744151 Chr.3: 44642391-44642391 on Build GRCh38
rs920769226 Chr.3: 44642371-44642371 on Build GRCh38
rs774680649 Chr.3: 44642600-44642600 on Build GRCh38
rs777699976 Chr.3: 44642599-44642601 on Build GRCh38
rs775331017 Chr.3: 44642644-44642644 on Build GRCh38
rs759806648 Chr.3: 44642287-44642287 on Build GRCh38
rs754891577 Chr.3: 44642354-44642354 on Build GRCh38
rs766019741 Chr.3: 44642516-44642516 on Build GRCh38
rs748787727 Chr.3: 44642625-44642625 on Build GRCh38
rs762637868 Chr.3: 44642514-44642514 on Build GRCh38
rs781577689 Chr.3: 44642609-44642609 on Build GRCh38
rs148523845 Chr.3: 44642491-44642491 on Build GRCh38
rs142476243 Chr.3: 44642296-44642296 on Build GRCh38
rs780405930 Chr.3: 44642586-44642586 on Build GRCh38
rs747374137 Chr.3: 44642495-44642495 on Build GRCh38
rs779950958 Chr.3: 44642237-44642237 on Build GRCh38
rs758199608 Chr.3: 44642323-44642323 on Build GRCh38
rs200130371 Chr.3: 44642383-44642383 on Build GRCh38
rs757289486 Chr.3: 44642474-44642474 on Build GRCh38
rs1045689798 Chr.3: 44642429-44642429 on Build GRCh38
rs774190033 Chr.3: 44642526-44642526 on Build GRCh38
rs772690637 Chr.3: 44642508-44642508 on Build GRCh38
rs777668872 Chr.3: 44642251-44642251 on Build GRCh38
rs770370021 Chr.3: 44642633-44642633 on Build GRCh38
rs139142373 Chr.3: 44642486-44642486 on Build GRCh38
rs779787494 Chr.3: 44642325-44642325 on Build GRCh38
rs754980379 Chr.3: 44642440-44642440 on Build GRCh38
rs760567181 Chr.3: 44642647-44642647 on Build GRCh38
rs758759580 Chr.3: 44642569-44642569 on Build GRCh38
rs35133599 Chr.3: 44642519-44642519 on Build GRCh38
rs756019945 Chr.3: 44642547-44642548 on Build GRCh38
rs761566023 Chr.3: 44642307-44642307 on Build GRCh38
rs769012958 Chr.3: 44642385-44642385 on Build GRCh38
rs566249568 Chr.3: 44642363-44642363 on Build GRCh38
rs769866425 Chr.3: 44642310-44642310 on Build GRCh38
rs750287565 Chr.3: 44642320-44642320 on Build GRCh38
rs759341212 Chr.3: 44642529-44642529 on Build GRCh38
rs746022328 Chr.3: 44642369-44642369 on Build GRCh38
rs764119270 Chr.3: 44642649-44642649 on Build GRCh38
rs772593820 Chr.3: 44642373-44642373 on Build GRCh38
rs371514033 Chr.3: 44642362-44642362 on Build GRCh38
rs750583643 Chr.3: 44642656-44642656 on Build GRCh38
rs749577535 Chr.3: 44642259-44642259 on Build GRCh38
rs1028746271 Chr.3: 44642293-44642293 on Build GRCh38
rs751522832 Chr.3: 44642344-44642344 on Build GRCh38
rs762546916 Chr.3: 44642403-44642403 on Build GRCh38
rs367642791 Chr.3: 44642450-44642450 on Build GRCh38
rs947966717 Chr.3: 44642487-44642487 on Build GRCh38
rs777952737 Chr.3: 44642358-44642358 on Build GRCh38
rs771030826 Chr.3: 44642271-44642271 on Build GRCh38
rs146540372 Chr.3: 44642553-44642553 on Build GRCh38
rs368530015 Chr.3: 44642387-44642387 on Build GRCh38

View Full Product Details