Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45082198-45082706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCP1
Entrez Gene ID
64866
Gene Name
CUB domain containing protein 1
Gene Aliases
CD318, SIMA135, TRASK
UniGene
Hs.476093
Gene Chromosome Location
Chr.3: 45082274-45146422 on Build GRCh38
Transcripts
NM_022842.4 XM_011534024.1 XM_017007070.1

SNP Detail

SNP ID Chromosome Location
rs1031056260 Chr.3: 45082654-45082654 on Build GRCh38
rs970021479 Chr.3: 45082660-45082660 on Build GRCh38
rs1052254471 Chr.3: 45082460-45082460 on Build GRCh38
rs943695980 Chr.3: 45082459-45082459 on Build GRCh38
rs927727369 Chr.3: 45082421-45082421 on Build GRCh38
rs935189666 Chr.3: 45082446-45082446 on Build GRCh38
rs767494605 Chr.3: 45082247-45082247 on Build GRCh38
rs981295086 Chr.3: 45082290-45082290 on Build GRCh38
rs972633027 Chr.3: 45082339-45082339 on Build GRCh38
rs965476011 Chr.3: 45082672-45082672 on Build GRCh38
rs928584405 Chr.3: 45082309-45082309 on Build GRCh38
rs753989936 Chr.3: 45082665-45082665 on Build GRCh38
rs547968498 Chr.3: 45082492-45082492 on Build GRCh38
rs764316118 Chr.3: 45082528-45082528 on Build GRCh38
rs146102015 Chr.3: 45082414-45082414 on Build GRCh38
rs970053120 Chr.3: 45082242-45082242 on Build GRCh38
rs1023018715 Chr.3: 45082659-45082659 on Build GRCh38
rs183676288 Chr.3: 45082605-45082605 on Build GRCh38
rs755863906 Chr.3: 45082437-45082437 on Build GRCh38
rs543888607 Chr.3: 45082465-45082465 on Build GRCh38
rs1040633580 Chr.3: 45082467-45082467 on Build GRCh38
rs376751397 Chr.3: 45082603-45082603 on Build GRCh38
rs972914670 Chr.3: 45082286-45082286 on Build GRCh38
rs931042759 Chr.3: 45082394-45082394 on Build GRCh38
rs1011626064 Chr.3: 45082230-45082230 on Build GRCh38
rs879655895 Chr.3: 45082308-45082308 on Build GRCh38
rs916256257 Chr.3: 45082366-45082366 on Build GRCh38
rs1006886522 Chr.3: 45082634-45082634 on Build GRCh38
rs961485333 Chr.3: 45082310-45082310 on Build GRCh38
rs902114248 Chr.3: 45082471-45082471 on Build GRCh38
rs11550126 Chr.3: 45082610-45082610 on Build GRCh38
rs1053259609 Chr.3: 45082530-45082530 on Build GRCh38
rs931584660 Chr.3: 45082338-45082338 on Build GRCh38
rs368743589 Chr.3: 45082533-45082533 on Build GRCh38
rs752490069 Chr.3: 45082322-45082322 on Build GRCh38
rs1020149318 Chr.3: 45082662-45082662 on Build GRCh38
rs1022884402 Chr.3: 45082235-45082235 on Build GRCh38
rs4634168 Chr.3: 45082681-45082681 on Build GRCh38
rs1002855239 Chr.3: 45082664-45082664 on Build GRCh38
rs192109919 Chr.3: 45082326-45082326 on Build GRCh38
rs1049507587 Chr.3: 45082408-45082408 on Build GRCh38
rs535671768 Chr.3: 45082305-45082305 on Build GRCh38
rs889831919 Chr.3: 45082611-45082611 on Build GRCh38
rs11063 Chr.3: 45082677-45082677 on Build GRCh38
rs867783309 Chr.3: 45082486-45082486 on Build GRCh38
rs984309595 Chr.3: 45082340-45082340 on Build GRCh38
rs1011593337 Chr.3: 45082622-45082622 on Build GRCh38
rs1046610576 Chr.3: 45082415-45082415 on Build GRCh38
rs919660351 Chr.3: 45082390-45082390 on Build GRCh38

View Full Product Details