Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45902295-45902795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR9
Entrez Gene ID
10803
Gene Name
C-C motif chemokine receptor 9
Gene Aliases
CC-CKR-9, CDw199, GPR-9-6, GPR28
UniGene
Hs.225946
Gene Chromosome Location
Chr.3: 45886504-45903177 on Build GRCh38
Transcripts
NM_001256369.1 NM_006641.3 NM_031200.2 XM_011533312.2 NM_001276378.1 NM_001276379.1 XM_017006644.1 XM_017006645.1

Gene Symbol
LZTFL1
Entrez Gene ID
54585
Gene Name
leucine zipper transcription factor like 1
Gene Aliases
BBS17
UniGene
Hs.30824
Gene Chromosome Location
Chr.3: 45823316-45916042 on Build GRCh38
Transcripts
NM_001256369.1 NM_006641.3 NM_031200.2 XM_011533312.2 NM_001276378.1 NM_001276379.1 XM_017006644.1 XM_017006645.1

SNP Detail

SNP ID Chromosome Location
rs200472101 Chr.3: 45902414-45902414 on Build GRCh38
rs200377147 Chr.3: 45902624-45902624 on Build GRCh38
rs144027662 Chr.3: 45902346-45902346 on Build GRCh38
rs201386944 Chr.3: 45902439-45902439 on Build GRCh38
rs942836198 Chr.3: 45902517-45902517 on Build GRCh38
rs199660171 Chr.3: 45902681-45902681 on Build GRCh38
rs200397646 Chr.3: 45902599-45902599 on Build GRCh38
rs200170116 Chr.3: 45902515-45902515 on Build GRCh38
rs200857548 Chr.3: 45902569-45902569 on Build GRCh38
rs1003302526 Chr.3: 45902708-45902708 on Build GRCh38
rs201284136 Chr.3: 45902479-45902479 on Build GRCh38
rs199697942 Chr.3: 45902696-45902696 on Build GRCh38
rs201442777 Chr.3: 45902420-45902420 on Build GRCh38
rs547390449 Chr.3: 45902417-45902417 on Build GRCh38
rs140757811 Chr.3: 45902448-45902448 on Build GRCh38
rs202161202 Chr.3: 45902622-45902622 on Build GRCh38
rs908588026 Chr.3: 45902336-45902336 on Build GRCh38
rs200040585 Chr.3: 45902425-45902425 on Build GRCh38
rs1057439575 Chr.3: 45902348-45902348 on Build GRCh38
rs757827095 Chr.3: 45902755-45902755 on Build GRCh38
rs201894026 Chr.3: 45902729-45902729 on Build GRCh38
rs552101562 Chr.3: 45902687-45902687 on Build GRCh38
rs192083942 Chr.3: 45902719-45902719 on Build GRCh38
rs888386254 Chr.3: 45902635-45902635 on Build GRCh38
rs199800584 Chr.3: 45902756-45902756 on Build GRCh38
rs56817174 Chr.3: 45902433-45902433 on Build GRCh38
rs200630761 Chr.3: 45902713-45902713 on Build GRCh38
rs202014793 Chr.3: 45902578-45902578 on Build GRCh38
rs73830610 Chr.3: 45902327-45902327 on Build GRCh38
rs961756794 Chr.3: 45902738-45902738 on Build GRCh38
rs201588273 Chr.3: 45902381-45902381 on Build GRCh38
rs199642231 Chr.3: 45902371-45902371 on Build GRCh38
rs201519911 Chr.3: 45902659-45902659 on Build GRCh38
rs989076679 Chr.3: 45902397-45902397 on Build GRCh38
rs202110703 Chr.3: 45902328-45902328 on Build GRCh38
rs200633785 Chr.3: 45902767-45902767 on Build GRCh38
rs201238517 Chr.3: 45902341-45902341 on Build GRCh38
rs199535242 Chr.3: 45902389-45902389 on Build GRCh38
rs552933184 Chr.3: 45902772-45902772 on Build GRCh38
rs200919915 Chr.3: 45902372-45902372 on Build GRCh38
rs775796023 Chr.3: 45902500-45902500 on Build GRCh38
rs202085580 Chr.3: 45902493-45902493 on Build GRCh38
rs201549112 Chr.3: 45902621-45902621 on Build GRCh38
rs1041485195 Chr.3: 45902524-45902524 on Build GRCh38
rs532262664 Chr.3: 45902370-45902370 on Build GRCh38
rs892177982 Chr.3: 45902501-45902501 on Build GRCh38
rs188947419 Chr.3: 45902690-45902690 on Build GRCh38
rs548505305 Chr.3: 45902548-45902548 on Build GRCh38
rs200420405 Chr.3: 45902340-45902340 on Build GRCh38
rs201024613 Chr.3: 45902691-45902691 on Build GRCh38
rs537203640 Chr.3: 45902677-45902677 on Build GRCh38
rs866460583 Chr.3: 45902369-45902369 on Build GRCh38
rs1000298349 Chr.3: 45902442-45902442 on Build GRCh38

View Full Product Details