Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46579847-46580351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC2
Entrez Gene ID
79442
Gene Name
leucine rich repeat containing 2
Gene Aliases
-
UniGene
Hs.657345
Gene Chromosome Location
Chr.3: 46515385-46580099 on Build GRCh38
Transcripts
XM_011534110.2 NM_001174136.1 NM_003212.3

Gene Symbol
TDGF1
Entrez Gene ID
6997
Gene Name
teratocarcinoma-derived growth factor 1
Gene Aliases
CR, CRGF, CRIPTO
UniGene
Hs.385870
Gene Chromosome Location
Chr.3: 46574555-46582463 on Build GRCh38
Transcripts
XM_011534110.2 NM_001174136.1 NM_003212.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5486359 c.448G>T p.D150Y Chr.3: 46580060-46580060 on Build GRCh38
6468934 c.336A>G p.K112K Chr.3: 46579851-46579851 on Build GRCh38
1309173 c.442G>A p.G148S Chr.3: 46580054-46580054 on Build GRCh38
6362248 c.339G>T p.E113D Chr.3: 46579951-46579951 on Build GRCh38
1045309 c.350C>A p.S117Y Chr.3: 46579962-46579962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759921933 Chr.3: 46579956-46579956 on Build GRCh38
rs2280413 Chr.3: 46580303-46580303 on Build GRCh38
rs35423054 Chr.3: 46579923-46579923 on Build GRCh38
rs758858378 Chr.3: 46580054-46580055 on Build GRCh38
rs952436226 Chr.3: 46580113-46580114 on Build GRCh38
rs766838145 Chr.3: 46580077-46580078 on Build GRCh38
rs773586696 Chr.3: 46579931-46579931 on Build GRCh38
rs760730510 Chr.3: 46580166-46580166 on Build GRCh38
rs1044851645 Chr.3: 46579888-46579889 on Build GRCh38
rs776758854 Chr.3: 46579930-46579930 on Build GRCh38
rs778799368 Chr.3: 46579898-46579898 on Build GRCh38
rs530873763 Chr.3: 46579990-46579990 on Build GRCh38
rs913802416 Chr.3: 46580191-46580191 on Build GRCh38
rs369887696 Chr.3: 46580040-46580040 on Build GRCh38
rs923543172 Chr.3: 46580117-46580117 on Build GRCh38
rs962525592 Chr.3: 46579951-46579951 on Build GRCh38
rs537100605 Chr.3: 46580204-46580204 on Build GRCh38
rs765298590 Chr.3: 46579976-46579976 on Build GRCh38
rs192154393 Chr.3: 46580163-46580163 on Build GRCh38
rs750746102 Chr.3: 46580052-46580053 on Build GRCh38
rs768691451 Chr.3: 46579884-46579884 on Build GRCh38
rs146889320 Chr.3: 46580028-46580028 on Build GRCh38
rs765529715 Chr.3: 46579957-46579958 on Build GRCh38
rs763661306 Chr.3: 46580038-46580038 on Build GRCh38
rs776242457 Chr.3: 46580048-46580048 on Build GRCh38
rs745687084 Chr.3: 46579906-46579906 on Build GRCh38
rs13075468 Chr.3: 46579960-46579960 on Build GRCh38
rs184761688 Chr.3: 46580112-46580112 on Build GRCh38
rs776993432 Chr.3: 46579889-46579892 on Build GRCh38
rs558413601 Chr.3: 46580296-46580296 on Build GRCh38
rs756132962 Chr.3: 46580014-46580014 on Build GRCh38
rs889192248 Chr.3: 46579977-46579977 on Build GRCh38
rs753863501 Chr.3: 46579893-46579893 on Build GRCh38
rs150809256 Chr.3: 46579943-46579943 on Build GRCh38
rs749850332 Chr.3: 46579942-46579942 on Build GRCh38
rs121909501 Chr.3: 46579986-46579986 on Build GRCh38
rs755667379 Chr.3: 46579881-46579881 on Build GRCh38
rs753862179 Chr.3: 46580066-46580066 on Build GRCh38
rs748468547 Chr.3: 46579993-46579993 on Build GRCh38
rs762031964 Chr.3: 46579924-46579929 on Build GRCh38
rs201516372 Chr.3: 46580061-46580061 on Build GRCh38
rs763689362 Chr.3: 46580242-46580242 on Build GRCh38
rs747367470 Chr.3: 46580110-46580110 on Build GRCh38
rs993984274 Chr.3: 46580278-46580278 on Build GRCh38
rs983793447 Chr.3: 46580115-46580115 on Build GRCh38
rs980480142 Chr.3: 46580119-46580119 on Build GRCh38
rs17852001 Chr.3: 46579954-46579954 on Build GRCh38
rs780402577 Chr.3: 46580107-46580107 on Build GRCh38
rs200408700 Chr.3: 46580102-46580102 on Build GRCh38
rs1037803323 Chr.3: 46580231-46580231 on Build GRCh38
rs879083910 Chr.3: 46579876-46579876 on Build GRCh38
rs977564552 Chr.3: 46580116-46580116 on Build GRCh38
rs777770656 Chr.3: 46580017-46580017 on Build GRCh38
rs199536679 Chr.3: 46580053-46580053 on Build GRCh38
rs759409674 Chr.3: 46580030-46580030 on Build GRCh38
rs149918911 Chr.3: 46580299-46580299 on Build GRCh38
rs200231018 Chr.3: 46580324-46580324 on Build GRCh38
rs749372682 Chr.3: 46580018-46580018 on Build GRCh38
rs186681190 Chr.3: 46579872-46579872 on Build GRCh38
rs760320801 Chr.3: 46580037-46580037 on Build GRCh38
rs753414482 Chr.3: 46579969-46579969 on Build GRCh38
rs755087463 Chr.3: 46579925-46579925 on Build GRCh38
rs757196291 Chr.3: 46579972-46579972 on Build GRCh38
rs748154683 Chr.3: 46579929-46579929 on Build GRCh38
rs771383889 Chr.3: 46580023-46580023 on Build GRCh38
rs369755187 Chr.3: 46580103-46580103 on Build GRCh38
rs767233749 Chr.3: 46579871-46579871 on Build GRCh38
rs753457057 Chr.3: 46580281-46580281 on Build GRCh38
rs200480549 Chr.3: 46579967-46579967 on Build GRCh38
rs941990270 Chr.3: 46580194-46580194 on Build GRCh38
rs781489694 Chr.3: 46579927-46579927 on Build GRCh38
rs559658799 Chr.3: 46579897-46579897 on Build GRCh38
rs767305917 Chr.3: 46580054-46580054 on Build GRCh38
rs763721268 Chr.3: 46579968-46579968 on Build GRCh38
rs552393963 Chr.3: 46580034-46580034 on Build GRCh38
rs751323439 Chr.3: 46580058-46580058 on Build GRCh38
rs777999994 Chr.3: 46580062-46580062 on Build GRCh38
rs774857986 Chr.3: 46580027-46580027 on Build GRCh38
rs761432328 Chr.3: 46580051-46580051 on Build GRCh38
rs867926061 Chr.3: 46579892-46579892 on Build GRCh38
rs1050894493 Chr.3: 46580164-46580164 on Build GRCh38
rs774629008 Chr.3: 46579950-46579950 on Build GRCh38
rs191449374 Chr.3: 46580067-46580067 on Build GRCh38
rs927707221 Chr.3: 46580141-46580141 on Build GRCh38
rs189625844 Chr.3: 46580113-46580113 on Build GRCh38

View Full Product Details