Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47577608-47578098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG5
Entrez Gene ID
10675
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
NGC
UniGene
Hs.45127
Gene Chromosome Location
Chr.3: 47562238-47580240 on Build GRCh38
Transcripts
NM_001206942.1 NM_001206943.1 NM_001206944.1 NM_001206945.1 NM_006574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4882864 c.379C>T p.L127F Chr.3: 47577647-47577647 on Build GRCh38
6636886 c.354C>T p.G118G Chr.3: 47577672-47577672 on Build GRCh38
5502692 c.418C>G p.P140A Chr.3: 47577608-47577608 on Build GRCh38
4118090 c.292A>G p.T98A Chr.3: 47577734-47577734 on Build GRCh38
345534 c.275C>T p.S92L Chr.3: 47577751-47577751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777508677 Chr.3: 47577878-47577878 on Build GRCh38
rs944862828 Chr.3: 47577673-47577673 on Build GRCh38
rs565546480 Chr.3: 47577918-47577918 on Build GRCh38
rs1013823755 Chr.3: 47577932-47577932 on Build GRCh38
rs547331033 Chr.3: 47577884-47577884 on Build GRCh38
rs900237513 Chr.3: 47577751-47577751 on Build GRCh38
rs945003490 Chr.3: 47578019-47578019 on Build GRCh38
rs368745564 Chr.3: 47577835-47577835 on Build GRCh38
rs983574554 Chr.3: 47578000-47578000 on Build GRCh38
rs755673769 Chr.3: 47577738-47577738 on Build GRCh38
rs759864034 Chr.3: 47577847-47577847 on Build GRCh38
rs113329262 Chr.3: 47577933-47577933 on Build GRCh38
rs144574217 Chr.3: 47577648-47577648 on Build GRCh38
rs773415706 Chr.3: 47577957-47577957 on Build GRCh38
rs988862604 Chr.3: 47578008-47578008 on Build GRCh38
rs776769845 Chr.3: 47577798-47577798 on Build GRCh38
rs1030344124 Chr.3: 47577984-47577984 on Build GRCh38
rs755832111 Chr.3: 47577874-47577874 on Build GRCh38
rs780743795 Chr.3: 47577737-47577737 on Build GRCh38
rs758222872 Chr.3: 47577934-47577934 on Build GRCh38
rs536944663 Chr.3: 47577942-47577942 on Build GRCh38
rs937714811 Chr.3: 47577770-47577770 on Build GRCh38
rs1023491201 Chr.3: 47577939-47577939 on Build GRCh38
rs976041243 Chr.3: 47577972-47577972 on Build GRCh38
rs776908099 Chr.3: 47577679-47577679 on Build GRCh38
rs941235305 Chr.3: 47578014-47578014 on Build GRCh38
rs750458179 Chr.3: 47577710-47577710 on Build GRCh38
rs773286841 Chr.3: 47577817-47577817 on Build GRCh38
rs769966583 Chr.3: 47577810-47577810 on Build GRCh38
rs755364149 Chr.3: 47577732-47577732 on Build GRCh38
rs752070836 Chr.3: 47577841-47577841 on Build GRCh38
rs763485103 Chr.3: 47577713-47577713 on Build GRCh38
rs766317802 Chr.3: 47577933-47577934 on Build GRCh38
rs549444682 Chr.3: 47577698-47577698 on Build GRCh38
rs1048857237 Chr.3: 47577808-47577808 on Build GRCh38
rs756822917 Chr.3: 47577887-47577887 on Build GRCh38
rs1001509092 Chr.3: 47577921-47577921 on Build GRCh38
rs865844420 Chr.3: 47578039-47578039 on Build GRCh38
rs374688213 Chr.3: 47577728-47577728 on Build GRCh38
rs868354878 Chr.3: 47578007-47578007 on Build GRCh38
rs1004553866 Chr.3: 47577834-47577834 on Build GRCh38
rs774268770 Chr.3: 47577944-47577945 on Build GRCh38
rs779933497 Chr.3: 47577937-47577937 on Build GRCh38
rs779036249 Chr.3: 47577899-47577899 on Build GRCh38
rs200269902 Chr.3: 47577824-47577824 on Build GRCh38
rs913463526 Chr.3: 47578016-47578016 on Build GRCh38
rs777457180 Chr.3: 47577744-47577744 on Build GRCh38
rs896435109 Chr.3: 47577926-47577926 on Build GRCh38
rs747096349 Chr.3: 47577994-47577994 on Build GRCh38
rs771869340 Chr.3: 47577788-47577788 on Build GRCh38
rs1056027689 Chr.3: 47578047-47578047 on Build GRCh38
rs767945759 Chr.3: 47577849-47577849 on Build GRCh38
rs571293541 Chr.3: 47577714-47577714 on Build GRCh38
rs1021515608 Chr.3: 47577895-47577895 on Build GRCh38
rs980126183 Chr.3: 47577951-47577951 on Build GRCh38
rs759318146 Chr.3: 47577663-47577663 on Build GRCh38
rs765282762 Chr.3: 47577705-47577705 on Build GRCh38
rs548931319 Chr.3: 47577975-47577975 on Build GRCh38
rs771004920 Chr.3: 47577777-47577777 on Build GRCh38
rs770019960 Chr.3: 47577950-47577950 on Build GRCh38
rs957365768 Chr.3: 47577996-47577996 on Build GRCh38
rs1040578256 Chr.3: 47577745-47577745 on Build GRCh38
rs750815387 Chr.3: 47578066-47578066 on Build GRCh38
rs976760084 Chr.3: 47578038-47578038 on Build GRCh38
rs1056140080 Chr.3: 47577814-47577814 on Build GRCh38
rs766936266 Chr.3: 47577837-47577837 on Build GRCh38
rs774291212 Chr.3: 47577660-47577660 on Build GRCh38
rs752961814 Chr.3: 47577862-47577862 on Build GRCh38
rs762121350 Chr.3: 47577807-47577807 on Build GRCh38
rs751975089 Chr.3: 47577729-47577729 on Build GRCh38
rs745797029 Chr.3: 47577910-47577910 on Build GRCh38
rs1038274349 Chr.3: 47577652-47577652 on Build GRCh38
rs913524355 Chr.3: 47577664-47577664 on Build GRCh38
rs781470055 Chr.3: 47577667-47577667 on Build GRCh38
rs745716723 Chr.3: 47577779-47577779 on Build GRCh38
rs748739514 Chr.3: 47577760-47577760 on Build GRCh38

View Full Product Details