Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48268207-48268702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF589
Entrez Gene ID
51385
Gene Name
zinc finger protein 589
Gene Aliases
SZF1
UniGene
Hs.172602
Gene Chromosome Location
Chr.3: 48241022-48275293 on Build GRCh38
Transcripts
NM_016089.2 XM_017006575.1 XM_017006576.1 XM_017006577.1 XM_017006578.1 XM_017006579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864694 c.985G>A p.E329K Chr.3: 48268676-48268676 on Build GRCh38
3594869 c.992C>T p.S331L Chr.3: 48268683-48268683 on Build GRCh38
3365206 c.926G>A p.C309Y Chr.3: 48268617-48268617 on Build GRCh38
5783127 c.815G>C p.G272A Chr.3: 48268506-48268506 on Build GRCh38
346718 c.858A>C p.I286I Chr.3: 48268549-48268549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747046361 Chr.3: 48268371-48268371 on Build GRCh38
rs79893792 Chr.3: 48268636-48268636 on Build GRCh38
rs574615806 Chr.3: 48268339-48268339 on Build GRCh38
rs371210936 Chr.3: 48268512-48268512 on Build GRCh38
rs71323392 Chr.3: 48268565-48268565 on Build GRCh38
rs370843777 Chr.3: 48268599-48268599 on Build GRCh38
rs746944268 Chr.3: 48268496-48268497 on Build GRCh38
rs777897899 Chr.3: 48268572-48268572 on Build GRCh38
rs778947942 Chr.3: 48268498-48268498 on Build GRCh38
rs370174993 Chr.3: 48268366-48268366 on Build GRCh38
rs976476431 Chr.3: 48268388-48268388 on Build GRCh38
rs764833194 Chr.3: 48268352-48268357 on Build GRCh38
rs763775270 Chr.3: 48268517-48268517 on Build GRCh38
rs752300068 Chr.3: 48268457-48268457 on Build GRCh38
rs757861108 Chr.3: 48268479-48268481 on Build GRCh38
rs569288162 Chr.3: 48268603-48268603 on Build GRCh38
rs768867547 Chr.3: 48268263-48268263 on Build GRCh38
rs530424709 Chr.3: 48268507-48268507 on Build GRCh38
rs897789956 Chr.3: 48268526-48268526 on Build GRCh38
rs764977114 Chr.3: 48268622-48268622 on Build GRCh38
rs564454039 Chr.3: 48268539-48268539 on Build GRCh38
rs770707034 Chr.3: 48268493-48268493 on Build GRCh38
rs752286998 Chr.3: 48268343-48268343 on Build GRCh38
rs769952189 Chr.3: 48268484-48268484 on Build GRCh38
rs758841033 Chr.3: 48268471-48268471 on Build GRCh38
rs371658406 Chr.3: 48268445-48268445 on Build GRCh38
rs550793582 Chr.3: 48268598-48268598 on Build GRCh38
rs762842315 Chr.3: 48268303-48268303 on Build GRCh38
rs369009928 Chr.3: 48268455-48268455 on Build GRCh38
rs780941159 Chr.3: 48268570-48268570 on Build GRCh38
rs374895625 Chr.3: 48268525-48268525 on Build GRCh38
rs373422989 Chr.3: 48268588-48268588 on Build GRCh38
rs377678218 Chr.3: 48268589-48268589 on Build GRCh38
rs764137307 Chr.3: 48268655-48268655 on Build GRCh38
rs754968067 Chr.3: 48268510-48268511 on Build GRCh38
rs372985265 Chr.3: 48268478-48268478 on Build GRCh38
rs917227919 Chr.3: 48268627-48268627 on Build GRCh38
rs781097137 Chr.3: 48268477-48268477 on Build GRCh38
rs768410268 Chr.3: 48268602-48268602 on Build GRCh38
rs755852556 Chr.3: 48268348-48268348 on Build GRCh38
rs772077810 Chr.3: 48268503-48268503 on Build GRCh38
rs200772666 Chr.3: 48268296-48268296 on Build GRCh38
rs202092368 Chr.3: 48268454-48268454 on Build GRCh38
rs201451932 Chr.3: 48268623-48268623 on Build GRCh38
rs772947464 Chr.3: 48268420-48268420 on Build GRCh38
rs933493741 Chr.3: 48268444-48268444 on Build GRCh38
rs942062245 Chr.3: 48268514-48268514 on Build GRCh38
rs770794949 Chr.3: 48268450-48268450 on Build GRCh38
rs752665188 Chr.3: 48268566-48268566 on Build GRCh38
rs761612073 Chr.3: 48268537-48268537 on Build GRCh38
rs760205576 Chr.3: 48268464-48268464 on Build GRCh38
rs747976451 Chr.3: 48268587-48268587 on Build GRCh38
rs200096770 Chr.3: 48268413-48268413 on Build GRCh38
rs766331160 Chr.3: 48268332-48268332 on Build GRCh38
rs780634908 Chr.3: 48268475-48268475 on Build GRCh38
rs768265868 Chr.3: 48268601-48268601 on Build GRCh38
rs756192788 Chr.3: 48268570-48268570 on Build GRCh38
rs753958821 Chr.3: 48268659-48268659 on Build GRCh38
rs991493678 Chr.3: 48268399-48268399 on Build GRCh38
rs537398400 Chr.3: 48268255-48268255 on Build GRCh38
rs899454876 Chr.3: 48268233-48268233 on Build GRCh38
rs759961458 Chr.3: 48268629-48268629 on Build GRCh38
rs917261067 Chr.3: 48268403-48268403 on Build GRCh38
rs755404118 Chr.3: 48268559-48268559 on Build GRCh38
rs758927114 Chr.3: 48268362-48268362 on Build GRCh38
rs762756407 Chr.3: 48268424-48268424 on Build GRCh38
rs201248262 Chr.3: 48268574-48268574 on Build GRCh38
rs778540933 Chr.3: 48268584-48268584 on Build GRCh38
rs756673175 Chr.3: 48268647-48268647 on Build GRCh38
rs1011203586 Chr.3: 48268616-48268616 on Build GRCh38
rs771143260 Chr.3: 48268678-48268679 on Build GRCh38
rs748300010 Chr.3: 48268383-48268383 on Build GRCh38
rs779208602 Chr.3: 48268668-48268668 on Build GRCh38
rs377506779 Chr.3: 48268662-48268662 on Build GRCh38
rs11718329 Chr.3: 48268338-48268338 on Build GRCh38
rs1033958950 Chr.3: 48268608-48268608 on Build GRCh38
rs760276637 Chr.3: 48268515-48268515 on Build GRCh38
rs1033587357 Chr.3: 48268242-48268242 on Build GRCh38
rs959300516 Chr.3: 48268609-48268609 on Build GRCh38
rs761522040 Chr.3: 48268604-48268604 on Build GRCh38
rs763863549 Chr.3: 48268467-48268467 on Build GRCh38
rs749903084 Chr.3: 48268374-48268387 on Build GRCh38
rs764931442 Chr.3: 48268271-48268271 on Build GRCh38
rs759369485 Chr.3: 48268500-48268500 on Build GRCh38
rs750761132 Chr.3: 48268468-48268468 on Build GRCh38
rs773035359 Chr.3: 48268292-48268292 on Build GRCh38
rs779976716 Chr.3: 48268487-48268487 on Build GRCh38
rs558895789 Chr.3: 48268264-48268264 on Build GRCh38
rs776626921 Chr.3: 48268637-48268637 on Build GRCh38
rs755561639 Chr.3: 48268476-48268476 on Build GRCh38
rs757524885 Chr.3: 48268576-48268576 on Build GRCh38
rs772747022 Chr.3: 48268241-48268241 on Build GRCh38
rs373593163 Chr.3: 48268382-48268382 on Build GRCh38
rs779389997 Chr.3: 48268354-48268354 on Build GRCh38
rs375870007 Chr.3: 48268353-48268353 on Build GRCh38
rs749014463 Chr.3: 48268490-48268490 on Build GRCh38
rs748286602 Chr.3: 48268481-48268481 on Build GRCh38
rs777981813 Chr.3: 48268489-48268489 on Build GRCh38
rs774259909 Chr.3: 48268499-48268499 on Build GRCh38
rs371119718 Chr.3: 48268256-48268256 on Build GRCh38
rs868389078 Chr.3: 48268640-48268640 on Build GRCh38
rs552108812 Chr.3: 48268538-48268538 on Build GRCh38

View Full Product Details