Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48298335-48298841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME6
Entrez Gene ID
10201
Gene Name
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases
IPIA-ALPHA, NDK 6, NM23-H6
UniGene
Hs.465558
Gene Chromosome Location
Chr.3: 48289369-48302259 on Build GRCh38
Transcripts
NM_001308426.1 NM_001308427.1 NM_001308428.1 NM_001308430.1 NM_001308431.1 NM_001308433.1 NM_001308434.1 NM_001308435.1 NM_005793.4 XM_017005511.1 XM_017005512.1 XM_017005513.1 XM_017005514.1 XM_017005515.1 XM_017005516.1 XM_017005517.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6164920 c.53C>A p.A18D Chr.3: 48298488-48298488 on Build GRCh38
6727358 c.94G>A p.A32T Chr.3: 48298447-48298447 on Build GRCh38
5497048 c.49C>G p.Q17E Chr.3: 48298492-48298492 on Build GRCh38
4118172 c.41G>A p.R14Q Chr.3: 48298500-48298500 on Build GRCh38
1670146 c.20G>T p.S7I Chr.3: 48298521-48298521 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752603465 Chr.3: 48298537-48298537 on Build GRCh38
rs112361174 Chr.3: 48298596-48298596 on Build GRCh38
rs922757164 Chr.3: 48298439-48298439 on Build GRCh38
rs145217729 Chr.3: 48298400-48298400 on Build GRCh38
rs756095614 Chr.3: 48298583-48298583 on Build GRCh38
rs765391620 Chr.3: 48298402-48298402 on Build GRCh38
rs751518004 Chr.3: 48298420-48298420 on Build GRCh38
rs1019804698 Chr.3: 48298717-48298717 on Build GRCh38
rs765420443 Chr.3: 48298521-48298521 on Build GRCh38
rs1032973033 Chr.3: 48298743-48298743 on Build GRCh38
rs750700488 Chr.3: 48298461-48298461 on Build GRCh38
rs772394180 Chr.3: 48298558-48298559 on Build GRCh38
rs896291027 Chr.3: 48298607-48298609 on Build GRCh38
rs140436849 Chr.3: 48298447-48298447 on Build GRCh38
rs755874243 Chr.3: 48298404-48298407 on Build GRCh38
rs766520445 Chr.3: 48298505-48298505 on Build GRCh38
rs766242085 Chr.3: 48298531-48298531 on Build GRCh38
rs375987418 Chr.3: 48298724-48298724 on Build GRCh38
rs749096550 Chr.3: 48298745-48298745 on Build GRCh38
rs376455498 Chr.3: 48298510-48298510 on Build GRCh38
rs1036872479 Chr.3: 48298538-48298538 on Build GRCh38
rs956372296 Chr.3: 48298655-48298655 on Build GRCh38
rs944885480 Chr.3: 48298436-48298436 on Build GRCh38
rs561840595 Chr.3: 48298703-48298703 on Build GRCh38
rs987799140 Chr.3: 48298670-48298670 on Build GRCh38
rs758467668 Chr.3: 48298493-48298493 on Build GRCh38
rs764140127 Chr.3: 48298577-48298577 on Build GRCh38
rs1032225828 Chr.3: 48298632-48298632 on Build GRCh38
rs780049468 Chr.3: 48298407-48298407 on Build GRCh38
rs202073300 Chr.3: 48298560-48298560 on Build GRCh38
rs940675538 Chr.3: 48298529-48298529 on Build GRCh38
rs200196665 Chr.3: 48298520-48298520 on Build GRCh38
rs372251148 Chr.3: 48298454-48298454 on Build GRCh38
rs13085251 Chr.3: 48298555-48298555 on Build GRCh38
rs979171460 Chr.3: 48298365-48298365 on Build GRCh38
rs200771816 Chr.3: 48298522-48298522 on Build GRCh38
rs779121550 Chr.3: 48298404-48298405 on Build GRCh38
rs747473393 Chr.3: 48298512-48298512 on Build GRCh38
rs371517115 Chr.3: 48298561-48298561 on Build GRCh38
rs908806126 Chr.3: 48298471-48298471 on Build GRCh38
rs755002273 Chr.3: 48298445-48298445 on Build GRCh38
rs758259358 Chr.3: 48298405-48298405 on Build GRCh38
rs540495008 Chr.3: 48298631-48298631 on Build GRCh38
rs147691869 Chr.3: 48298448-48298448 on Build GRCh38
rs964039244 Chr.3: 48298774-48298774 on Build GRCh38
rs754897425 Chr.3: 48298539-48298539 on Build GRCh38
rs573092585 Chr.3: 48298622-48298622 on Build GRCh38
rs746069617 Chr.3: 48298516-48298517 on Build GRCh38
rs749006710 Chr.3: 48298500-48298500 on Build GRCh38
rs769433709 Chr.3: 48298453-48298453 on Build GRCh38
rs922107980 Chr.3: 48298761-48298761 on Build GRCh38
rs753893815 Chr.3: 48298401-48298401 on Build GRCh38
rs763313374 Chr.3: 48298525-48298525 on Build GRCh38
rs757178115 Chr.3: 48298757-48298757 on Build GRCh38
rs1000822576 Chr.3: 48298737-48298737 on Build GRCh38
rs1049265733 Chr.3: 48298474-48298474 on Build GRCh38
rs373853526 Chr.3: 48298534-48298534 on Build GRCh38
rs767373381 Chr.3: 48298554-48298554 on Build GRCh38
rs750862307 Chr.3: 48298707-48298707 on Build GRCh38
rs73831565 Chr.3: 48298592-48298592 on Build GRCh38
rs867549210 Chr.3: 48298610-48298610 on Build GRCh38
rs975509419 Chr.3: 48298423-48298423 on Build GRCh38
rs769064177 Chr.3: 48298518-48298518 on Build GRCh38
rs772415195 Chr.3: 48298501-48298501 on Build GRCh38
rs190676458 Chr.3: 48298480-48298480 on Build GRCh38

View Full Product Details