Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48599984-48600488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UQCRC1
Entrez Gene ID
7384
Gene Name
ubiquinol-cytochrome c reductase core protein I
Gene Aliases
D3S3191, QCR1, UQCR1
UniGene
Hs.119251
Gene Chromosome Location
Chr.3: 48598999-48609665 on Build GRCh38
Transcripts
NM_003365.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564560 c.1274_1275CC>TT p.P425L Chr.3: 48600090-48600091 on Build GRCh38
6164899 c.1254G>A p.L418L Chr.3: 48600111-48600111 on Build GRCh38
6817713 c.1263C>T p.G421G Chr.3: 48600102-48600102 on Build GRCh38
242183 c.1265G>A p.R422H Chr.3: 48600100-48600100 on Build GRCh38
5864724 c.1295G>A p.R432Q Chr.3: 48600070-48600070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111768878 Chr.3: 48600153-48600153 on Build GRCh38
rs771354964 Chr.3: 48600114-48600114 on Build GRCh38
rs750440742 Chr.3: 48600080-48600080 on Build GRCh38
rs570940063 Chr.3: 48600163-48600163 on Build GRCh38
rs1047976702 Chr.3: 48600089-48600089 on Build GRCh38
rs768689935 Chr.3: 48600434-48600434 on Build GRCh38
rs536201637 Chr.3: 48600226-48600226 on Build GRCh38
rs531011243 Chr.3: 48600117-48600117 on Build GRCh38
rs142369485 Chr.3: 48600030-48600030 on Build GRCh38
rs376087580 Chr.3: 48600121-48600121 on Build GRCh38
rs976544103 Chr.3: 48600309-48600309 on Build GRCh38
rs778689141 Chr.3: 48600323-48600323 on Build GRCh38
rs760882102 Chr.3: 48600064-48600064 on Build GRCh38
rs865819911 Chr.3: 48600451-48600451 on Build GRCh38
rs1048048319 Chr.3: 48600369-48600369 on Build GRCh38
rs370334893 Chr.3: 48600122-48600122 on Build GRCh38
rs779551181 Chr.3: 48600097-48600097 on Build GRCh38
rs1026476330 Chr.3: 48600220-48600220 on Build GRCh38
rs763330421 Chr.3: 48600026-48600026 on Build GRCh38
rs910655500 Chr.3: 48600275-48600275 on Build GRCh38
rs568762177 Chr.3: 48600218-48600218 on Build GRCh38
rs772577890 Chr.3: 48600453-48600453 on Build GRCh38
rs779267869 Chr.3: 48600435-48600435 on Build GRCh38
rs899402098 Chr.3: 48600211-48600211 on Build GRCh38
rs888023589 Chr.3: 48600388-48600388 on Build GRCh38
rs1021457714 Chr.3: 48600215-48600215 on Build GRCh38
rs747470917 Chr.3: 48600396-48600396 on Build GRCh38
rs780005332 Chr.3: 48600274-48600274 on Build GRCh38
rs1003544448 Chr.3: 48600243-48600243 on Build GRCh38
rs774882687 Chr.3: 48600039-48600039 on Build GRCh38
rs777977428 Chr.3: 48600198-48600198 on Build GRCh38
rs35291424 Chr.3: 48600076-48600077 on Build GRCh38
rs757354563 Chr.3: 48600205-48600205 on Build GRCh38
rs761693806 Chr.3: 48600158-48600158 on Build GRCh38
rs910633419 Chr.3: 48600038-48600038 on Build GRCh38
rs773853914 Chr.3: 48600040-48600044 on Build GRCh38
rs978362081 Chr.3: 48600456-48600456 on Build GRCh38
rs151327221 Chr.3: 48600414-48600414 on Build GRCh38
rs746247555 Chr.3: 48600445-48600445 on Build GRCh38
rs561599217 Chr.3: 48600010-48600010 on Build GRCh38
rs959340454 Chr.3: 48600362-48600362 on Build GRCh38
rs943485757 Chr.3: 48600282-48600282 on Build GRCh38
rs376772981 Chr.3: 48600036-48600036 on Build GRCh38
rs755779479 Chr.3: 48600200-48600200 on Build GRCh38
rs886535239 Chr.3: 48600233-48600233 on Build GRCh38
rs548810979 Chr.3: 48600384-48600384 on Build GRCh38
rs754611181 Chr.3: 48600100-48600100 on Build GRCh38
rs747859059 Chr.3: 48600189-48600189 on Build GRCh38
rs569760734 Chr.3: 48600106-48600106 on Build GRCh38
rs753634982 Chr.3: 48600070-48600070 on Build GRCh38
rs371109527 Chr.3: 48600463-48600463 on Build GRCh38
rs767697688 Chr.3: 48600057-48600057 on Build GRCh38
rs771403679 Chr.3: 48600459-48600459 on Build GRCh38
rs768199715 Chr.3: 48600021-48600021 on Build GRCh38
rs757165759 Chr.3: 48600071-48600071 on Build GRCh38
rs143235444 Chr.3: 48600382-48600382 on Build GRCh38
rs1050755327 Chr.3: 48600143-48600143 on Build GRCh38
rs376280155 Chr.3: 48600126-48600126 on Build GRCh38
rs373520913 Chr.3: 48600041-48600041 on Build GRCh38
rs941722324 Chr.3: 48600397-48600397 on Build GRCh38
rs923443350 Chr.3: 48600354-48600354 on Build GRCh38
rs778960903 Chr.3: 48600075-48600075 on Build GRCh38
rs373119179 Chr.3: 48600090-48600090 on Build GRCh38
rs773526213 Chr.3: 48600022-48600022 on Build GRCh38
rs777032513 Chr.3: 48600147-48600147 on Build GRCh38
rs750191813 Chr.3: 48600161-48600161 on Build GRCh38
rs547224474 Chr.3: 48600165-48600165 on Build GRCh38
rs746893368 Chr.3: 48600302-48600302 on Build GRCh38
rs534403170 Chr.3: 48600386-48600386 on Build GRCh38
rs780875481 Chr.3: 48600101-48600101 on Build GRCh38
rs746531924 Chr.3: 48600098-48600098 on Build GRCh38
rs749871981 Chr.3: 48600048-48600048 on Build GRCh38
rs200945906 Chr.3: 48600164-48600164 on Build GRCh38
rs202154537 Chr.3: 48600159-48600159 on Build GRCh38
rs368507809 Chr.3: 48600128-48600128 on Build GRCh38
rs764438194 Chr.3: 48600067-48600067 on Build GRCh38
rs556975231 Chr.3: 48600297-48600297 on Build GRCh38
rs749308153 Chr.3: 48600201-48600201 on Build GRCh38
rs530620760 Chr.3: 48600370-48600370 on Build GRCh38
rs557543632 Chr.3: 48600462-48600462 on Build GRCh38
rs780822393 Chr.3: 48600174-48600174 on Build GRCh38
rs1022659388 Chr.3: 48600424-48600424 on Build GRCh38
rs540446036 Chr.3: 48600004-48600004 on Build GRCh38
rs758282397 Chr.3: 48600084-48600084 on Build GRCh38
rs959285363 Chr.3: 48600024-48600024 on Build GRCh38

View Full Product Details