Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48782896-48783408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAR2A
Entrez Gene ID
5576
Gene Name
protein kinase cAMP-dependent type II regulatory subunit alpha
Gene Aliases
PKR2, PRKAR2
UniGene
Hs.631923
Gene Chromosome Location
Chr.3: 48744601-48847850 on Build GRCh38
Transcripts
NM_001321982.1 NM_001321983.1 NM_001321989.1 NM_004157.3 XM_005265315.3 XM_011533942.2 XM_011533943.1 XM_017006815.1 XM_017006816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285511 c.517G>A p.G173R Chr.3: 48783011-48783011 on Build GRCh38
1309241 c.448C>A p.Q150K Chr.3: 48783080-48783080 on Build GRCh38
4118335 c.456C>T p.L152L Chr.3: 48783072-48783072 on Build GRCh38
6755717 c.443T>G p.L148R Chr.3: 48783085-48783085 on Build GRCh38
1566739 c.541C>T p.R181W Chr.3: 48782987-48782987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374213032 Chr.3: 48783121-48783121 on Build GRCh38
rs1021610775 Chr.3: 48783010-48783010 on Build GRCh38
rs779126494 Chr.3: 48782949-48782949 on Build GRCh38
rs914424131 Chr.3: 48783259-48783259 on Build GRCh38
rs1043803091 Chr.3: 48783332-48783332 on Build GRCh38
rs560353945 Chr.3: 48783071-48783071 on Build GRCh38
rs763180994 Chr.3: 48783346-48783346 on Build GRCh38
rs757835834 Chr.3: 48783069-48783069 on Build GRCh38
rs773819473 Chr.3: 48783139-48783139 on Build GRCh38
rs569041219 Chr.3: 48783155-48783155 on Build GRCh38
rs762639940 Chr.3: 48783020-48783020 on Build GRCh38
rs374537905 Chr.3: 48782952-48782952 on Build GRCh38
rs778105179 Chr.3: 48782944-48782944 on Build GRCh38
rs201062271 Chr.3: 48782987-48782987 on Build GRCh38
rs566685047 Chr.3: 48783310-48783310 on Build GRCh38
rs201671205 Chr.3: 48783072-48783072 on Build GRCh38
rs762407930 Chr.3: 48783131-48783131 on Build GRCh38
rs550124936 Chr.3: 48783210-48783210 on Build GRCh38
rs928515177 Chr.3: 48783171-48783171 on Build GRCh38
rs140319669 Chr.3: 48782924-48782924 on Build GRCh38
rs930965578 Chr.3: 48783347-48783347 on Build GRCh38
rs780071307 Chr.3: 48783082-48783082 on Build GRCh38
rs370951963 Chr.3: 48783302-48783302 on Build GRCh38
rs768389946 Chr.3: 48783095-48783095 on Build GRCh38
rs111499766 Chr.3: 48782948-48782948 on Build GRCh38
rs981554178 Chr.3: 48783090-48783090 on Build GRCh38
rs34544100 Chr.3: 48783260-48783261 on Build GRCh38
rs772674401 Chr.3: 48783015-48783015 on Build GRCh38
rs746831162 Chr.3: 48783092-48783092 on Build GRCh38
rs908272468 Chr.3: 48783383-48783383 on Build GRCh38
rs760885038 Chr.3: 48783040-48783040 on Build GRCh38
rs947230531 Chr.3: 48783264-48783264 on Build GRCh38
rs754364832 Chr.3: 48783052-48783052 on Build GRCh38
rs910658784 Chr.3: 48783382-48783382 on Build GRCh38
rs754228715 Chr.3: 48783030-48783030 on Build GRCh38
rs534229511 Chr.3: 48783366-48783366 on Build GRCh38
rs551576146 Chr.3: 48783231-48783231 on Build GRCh38
rs988025620 Chr.3: 48783156-48783156 on Build GRCh38
rs988575001 Chr.3: 48783188-48783188 on Build GRCh38
rs760395553 Chr.3: 48783176-48783176 on Build GRCh38
rs549211150 Chr.3: 48783075-48783075 on Build GRCh38
rs367819043 Chr.3: 48782945-48782945 on Build GRCh38
rs371959163 Chr.3: 48783035-48783035 on Build GRCh38
rs768640819 Chr.3: 48782982-48782982 on Build GRCh38
rs368491432 Chr.3: 48783000-48783000 on Build GRCh38
rs201124752 Chr.3: 48783124-48783124 on Build GRCh38
rs904524915 Chr.3: 48783065-48783065 on Build GRCh38
rs770576475 Chr.3: 48783132-48783132 on Build GRCh38
rs982510242 Chr.3: 48783359-48783359 on Build GRCh38
rs182210450 Chr.3: 48783032-48783032 on Build GRCh38
rs577790327 Chr.3: 48782996-48782996 on Build GRCh38
rs746734245 Chr.3: 48782981-48782981 on Build GRCh38
rs973629311 Chr.3: 48783230-48783230 on Build GRCh38
rs780045555 Chr.3: 48782962-48782962 on Build GRCh38
rs770148612 Chr.3: 48783167-48783167 on Build GRCh38
rs773371231 Chr.3: 48783314-48783314 on Build GRCh38
rs772729688 Chr.3: 48783125-48783125 on Build GRCh38
rs919559324 Chr.3: 48783244-48783244 on Build GRCh38
rs776704329 Chr.3: 48783126-48783126 on Build GRCh38
rs955964180 Chr.3: 48783180-48783180 on Build GRCh38
rs764678772 Chr.3: 48783046-48783046 on Build GRCh38
rs772393758 Chr.3: 48782956-48782956 on Build GRCh38
rs187242176 Chr.3: 48783112-48783112 on Build GRCh38
rs191910867 Chr.3: 48783213-48783213 on Build GRCh38

View Full Product Details