Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48790375-48790906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAR2A
Entrez Gene ID
5576
Gene Name
protein kinase cAMP-dependent type II regulatory subunit alpha
Gene Aliases
PKR2, PRKAR2
UniGene
Hs.631923
Gene Chromosome Location
Chr.3: 48744601-48847850 on Build GRCh38
Transcripts
NM_001321982.1 NM_001321983.1 NM_001321989.1 NM_004157.3 XM_005265315.3 XM_011533942.2 XM_011533943.1 XM_017006815.1 XM_017006816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5780938 c.420C>G p.F140L Chr.3: 48790559-48790559 on Build GRCh38
5978631 c.401G>A p.C134Y Chr.3: 48790578-48790578 on Build GRCh38
5521199 c.361C>T p.P121S Chr.3: 48790618-48790618 on Build GRCh38
1328167 c.376C>T p.Q126* Chr.3: 48790603-48790603 on Build GRCh38
1045961 c.406G>T p.D136Y Chr.3: 48790573-48790573 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765793002 Chr.3: 48790534-48790534 on Build GRCh38
rs573758838 Chr.3: 48790836-48790836 on Build GRCh38
rs766465497 Chr.3: 48790545-48790545 on Build GRCh38
rs777026773 Chr.3: 48790508-48790508 on Build GRCh38
rs1044152237 Chr.3: 48790736-48790737 on Build GRCh38
rs900728827 Chr.3: 48790514-48790514 on Build GRCh38
rs764433139 Chr.3: 48790493-48790493 on Build GRCh38
rs979403210 Chr.3: 48790505-48790505 on Build GRCh38
rs944805389 Chr.3: 48790837-48790837 on Build GRCh38
rs755192539 Chr.3: 48790573-48790573 on Build GRCh38
rs929433161 Chr.3: 48790656-48790656 on Build GRCh38
rs762462281 Chr.3: 48790531-48790531 on Build GRCh38
rs763546803 Chr.3: 48790663-48790663 on Build GRCh38
rs888103713 Chr.3: 48790577-48790577 on Build GRCh38
rs750565022 Chr.3: 48790535-48790535 on Build GRCh38
rs762231146 Chr.3: 48790649-48790649 on Build GRCh38
rs941764322 Chr.3: 48790735-48790735 on Build GRCh38
rs755040216 Chr.3: 48790447-48790447 on Build GRCh38
rs192526145 Chr.3: 48790638-48790638 on Build GRCh38
rs1002730502 Chr.3: 48790429-48790429 on Build GRCh38
rs763391493 Chr.3: 48790656-48790657 on Build GRCh38
rs748291376 Chr.3: 48790587-48790587 on Build GRCh38
rs773911011 Chr.3: 48790654-48790654 on Build GRCh38
rs372227280 Chr.3: 48790631-48790631 on Build GRCh38
rs769731769 Chr.3: 48790619-48790622 on Build GRCh38
rs1021422511 Chr.3: 48790482-48790482 on Build GRCh38
rs925907224 Chr.3: 48790827-48790827 on Build GRCh38
rs557474201 Chr.3: 48790777-48790777 on Build GRCh38
rs764502632 Chr.3: 48790489-48790489 on Build GRCh38
rs922100438 Chr.3: 48790863-48790863 on Build GRCh38
rs145200736 Chr.3: 48790402-48790402 on Build GRCh38
rs535678587 Chr.3: 48790461-48790461 on Build GRCh38
rs544440541 Chr.3: 48790862-48790862 on Build GRCh38
rs530832803 Chr.3: 48790652-48790652 on Build GRCh38
rs750758696 Chr.3: 48790882-48790882 on Build GRCh38
rs758492480 Chr.3: 48790538-48790538 on Build GRCh38
rs779101322 Chr.3: 48790572-48790572 on Build GRCh38
rs201060984 Chr.3: 48790708-48790709 on Build GRCh38
rs772392315 Chr.3: 48790773-48790773 on Build GRCh38
rs776884768 Chr.3: 48790641-48790641 on Build GRCh38
rs1057398426 Chr.3: 48790799-48790799 on Build GRCh38
rs1014061970 Chr.3: 48790764-48790764 on Build GRCh38
rs751791908 Chr.3: 48790546-48790546 on Build GRCh38
rs1048055263 Chr.3: 48790547-48790547 on Build GRCh38
rs770407130 Chr.3: 48790626-48790626 on Build GRCh38
rs879094305 Chr.3: 48790592-48790592 on Build GRCh38
rs539701758 Chr.3: 48790771-48790771 on Build GRCh38
rs960091041 Chr.3: 48790758-48790758 on Build GRCh38
rs762251125 Chr.3: 48790801-48790801 on Build GRCh38
rs777619237 Chr.3: 48790606-48790606 on Build GRCh38
rs762145911 Chr.3: 48790501-48790502 on Build GRCh38
rs765850196 Chr.3: 48790651-48790651 on Build GRCh38
rs113030327 Chr.3: 48790580-48790580 on Build GRCh38
rs561857915 Chr.3: 48790834-48790834 on Build GRCh38
rs865908799 Chr.3: 48790455-48790455 on Build GRCh38
rs748812211 Chr.3: 48790624-48790624 on Build GRCh38
rs925959641 Chr.3: 48790439-48790439 on Build GRCh38
rs752119160 Chr.3: 48790673-48790673 on Build GRCh38
rs760317517 Chr.3: 48790446-48790446 on Build GRCh38
rs1010073781 Chr.3: 48790468-48790468 on Build GRCh38
rs149126652 Chr.3: 48790808-48790808 on Build GRCh38
rs766405188 Chr.3: 48790667-48790667 on Build GRCh38
rs756075947 Chr.3: 48790588-48790588 on Build GRCh38
rs948659370 Chr.3: 48790397-48790397 on Build GRCh38
rs1010455383 Chr.3: 48790813-48790813 on Build GRCh38
rs976738602 Chr.3: 48790595-48790595 on Build GRCh38
rs373211422 Chr.3: 48790709-48790710 on Build GRCh38
rs199498651 Chr.3: 48790578-48790578 on Build GRCh38
rs199568216 Chr.3: 48790823-48790823 on Build GRCh38
rs890563870 Chr.3: 48790481-48790481 on Build GRCh38
rs539840920 Chr.3: 48790829-48790829 on Build GRCh38

View Full Product Details