Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49005974-49006470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P4HTM
Entrez Gene ID
54681
Gene Name
prolyl 4-hydroxylase, transmembrane
Gene Aliases
EGLN4, HIFPH4, P4H-TM, PH-4, PH4, PHD4
UniGene
Hs.654944
Gene Chromosome Location
Chr.3: 48989908-49007149 on Build GRCh38
Transcripts
NM_177938.2 NM_177939.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736613 c.1182C>T p.D394D Chr.3: 49006081-49006081 on Build GRCh38
3595148 c.1403G>A p.G468E Chr.3: 49006119-49006119 on Build GRCh38
7382176 c.1213G>C p.D405H Chr.3: 49006112-49006112 on Build GRCh38
4710719 c.1410G>T p.L470L Chr.3: 49006126-49006126 on Build GRCh38
3595149 c.1220G>A p.G407E Chr.3: 49006119-49006119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043862836 Chr.3: 49006345-49006345 on Build GRCh38
rs895264209 Chr.3: 49006012-49006012 on Build GRCh38
rs757519182 Chr.3: 49006402-49006403 on Build GRCh38
rs763413685 Chr.3: 49006071-49006071 on Build GRCh38
rs1008555943 Chr.3: 49006427-49006427 on Build GRCh38
rs371643299 Chr.3: 49006218-49006218 on Build GRCh38
rs374239797 Chr.3: 49006396-49006397 on Build GRCh38
rs7624225 Chr.3: 49006108-49006108 on Build GRCh38
rs746600922 Chr.3: 49006149-49006149 on Build GRCh38
rs988884327 Chr.3: 49006107-49006107 on Build GRCh38
rs367845737 Chr.3: 49006057-49006057 on Build GRCh38
rs371468668 Chr.3: 49006081-49006081 on Build GRCh38
rs755173723 Chr.3: 49006075-49006083 on Build GRCh38
rs778876398 Chr.3: 49006118-49006118 on Build GRCh38
rs770179774 Chr.3: 49006162-49006162 on Build GRCh38
rs902788716 Chr.3: 49006368-49006368 on Build GRCh38
rs1030862817 Chr.3: 49006262-49006262 on Build GRCh38
rs927926797 Chr.3: 49006248-49006248 on Build GRCh38
rs749965454 Chr.3: 49006126-49006126 on Build GRCh38
rs138246626 Chr.3: 49006026-49006026 on Build GRCh38
rs199972576 Chr.3: 49006100-49006100 on Build GRCh38
rs773718180 Chr.3: 49006064-49006064 on Build GRCh38
rs759545561 Chr.3: 49006195-49006195 on Build GRCh38
rs778063783 Chr.3: 49006168-49006168 on Build GRCh38
rs774919797 Chr.3: 49006190-49006190 on Build GRCh38
rs775885940 Chr.3: 49006209-49006209 on Build GRCh38
rs772122473 Chr.3: 49006196-49006196 on Build GRCh38
rs1010735434 Chr.3: 49006342-49006342 on Build GRCh38
rs566437375 Chr.3: 49006025-49006025 on Build GRCh38
rs970612471 Chr.3: 49006054-49006054 on Build GRCh38
rs1023266903 Chr.3: 49006251-49006251 on Build GRCh38
rs758831068 Chr.3: 49006062-49006063 on Build GRCh38
rs539398559 Chr.3: 49006326-49006326 on Build GRCh38
rs376875133 Chr.3: 49006210-49006210 on Build GRCh38
rs372064608 Chr.3: 49006138-49006138 on Build GRCh38
rs998508263 Chr.3: 49006031-49006031 on Build GRCh38
rs946911716 Chr.3: 49006328-49006328 on Build GRCh38
rs748713169 Chr.3: 49006210-49006210 on Build GRCh38
rs143571875 Chr.3: 49006252-49006252 on Build GRCh38
rs911203680 Chr.3: 49006436-49006436 on Build GRCh38
rs890084314 Chr.3: 49006402-49006402 on Build GRCh38
rs939452312 Chr.3: 49006249-49006249 on Build GRCh38
rs895222364 Chr.3: 49006276-49006276 on Build GRCh38
rs386660822 Chr.3: 49005999-49006003 on Build GRCh38
rs188208065 Chr.3: 49006385-49006385 on Build GRCh38
rs541736041 Chr.3: 49006209-49006210 on Build GRCh38
rs771660136 Chr.3: 49006029-49006029 on Build GRCh38
rs1003385408 Chr.3: 49006257-49006257 on Build GRCh38
rs757914897 Chr.3: 49006137-49006137 on Build GRCh38
rs375889763 Chr.3: 49006092-49006092 on Build GRCh38
rs779588385 Chr.3: 49006147-49006147 on Build GRCh38
rs751238636 Chr.3: 49006217-49006217 on Build GRCh38
rs774344716 Chr.3: 49006083-49006083 on Build GRCh38
rs1024014067 Chr.3: 49006027-49006027 on Build GRCh38
rs182639399 Chr.3: 49006381-49006381 on Build GRCh38
rs767700412 Chr.3: 49006091-49006091 on Build GRCh38
rs753849062 Chr.3: 49006105-49006105 on Build GRCh38
rs759632578 Chr.3: 49006088-49006088 on Build GRCh38
rs765142473 Chr.3: 49006213-49006213 on Build GRCh38
rs368732856 Chr.3: 49006093-49006093 on Build GRCh38
rs1017446231 Chr.3: 49006192-49006192 on Build GRCh38
rs201918214 Chr.3: 49006211-49006211 on Build GRCh38
rs750259045 Chr.3: 49006173-49006173 on Build GRCh38
rs771384004 Chr.3: 49006177-49006177 on Build GRCh38
rs200837151 Chr.3: 49006215-49006215 on Build GRCh38
rs761661739 Chr.3: 49006212-49006212 on Build GRCh38
rs780791874 Chr.3: 49006235-49006235 on Build GRCh38
rs374563891 Chr.3: 49006082-49006082 on Build GRCh38
rs375230127 Chr.3: 49006017-49006017 on Build GRCh38
rs985870229 Chr.3: 49006406-49006406 on Build GRCh38
rs779749783 Chr.3: 49006043-49006043 on Build GRCh38
rs757304685 Chr.3: 49006116-49006116 on Build GRCh38
rs546993167 Chr.3: 49006047-49006047 on Build GRCh38
rs764871844 Chr.3: 49006109-49006109 on Build GRCh38
rs1052722398 Chr.3: 49006383-49006383 on Build GRCh38
rs754674889 Chr.3: 49006229-49006229 on Build GRCh38
rs983464063 Chr.3: 49006244-49006244 on Build GRCh38
rs985469053 Chr.3: 49006207-49006207 on Build GRCh38
rs78875983 Chr.3: 49006155-49006155 on Build GRCh38

View Full Product Details