Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49428922-49429418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NICN1
Entrez Gene ID
84276
Gene Name
nicolin 1
Gene Aliases
-
UniGene
Hs.191460
Gene Chromosome Location
Chr.3: 49422333-49429324 on Build GRCh38
Transcripts
NM_032316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7414517 c.43G>T p.A15S Chr.3: 49429197-49429197 on Build GRCh38
1137057 c.15G>A p.L5L Chr.3: 49429225-49429225 on Build GRCh38
5774433 c.97A>G p.I33V Chr.3: 49429143-49429143 on Build GRCh38
6499864 c.129C>G p.F43L Chr.3: 49429111-49429111 on Build GRCh38
4650634 c.19C>T p.P7S Chr.3: 49429221-49429221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989055427 Chr.3: 49429326-49429326 on Build GRCh38
rs772909297 Chr.3: 49429135-49429135 on Build GRCh38
rs754321515 Chr.3: 49429165-49429165 on Build GRCh38
rs141772476 Chr.3: 49429132-49429132 on Build GRCh38
rs371055267 Chr.3: 49429200-49429200 on Build GRCh38
rs776591820 Chr.3: 49429202-49429202 on Build GRCh38
rs766646065 Chr.3: 49429142-49429143 on Build GRCh38
rs376139643 Chr.3: 49429067-49429067 on Build GRCh38
rs1012870967 Chr.3: 49429111-49429111 on Build GRCh38
rs766118469 Chr.3: 49429144-49429144 on Build GRCh38
rs765648502 Chr.3: 49429250-49429250 on Build GRCh38
rs944503499 Chr.3: 49429005-49429005 on Build GRCh38
rs138545966 Chr.3: 49429175-49429175 on Build GRCh38
rs746243043 Chr.3: 49429180-49429180 on Build GRCh38
rs568625671 Chr.3: 49429192-49429192 on Build GRCh38
rs771305193 Chr.3: 49429069-49429069 on Build GRCh38
rs781525372 Chr.3: 49429213-49429213 on Build GRCh38
rs896735323 Chr.3: 49429089-49429089 on Build GRCh38
rs1008453474 Chr.3: 49429160-49429160 on Build GRCh38
rs940927095 Chr.3: 49429079-49429079 on Build GRCh38
rs1056698168 Chr.3: 49429087-49429087 on Build GRCh38
rs970595702 Chr.3: 49428986-49428986 on Build GRCh38
rs780250047 Chr.3: 49429191-49429191 on Build GRCh38
rs748076037 Chr.3: 49429205-49429205 on Build GRCh38
rs749292015 Chr.3: 49429280-49429280 on Build GRCh38
rs777825507 Chr.3: 49429279-49429279 on Build GRCh38
rs770805412 Chr.3: 49428994-49428994 on Build GRCh38
rs766985199 Chr.3: 49429162-49429162 on Build GRCh38
rs564651403 Chr.3: 49429124-49429124 on Build GRCh38
rs916112653 Chr.3: 49428973-49428973 on Build GRCh38
rs751633578 Chr.3: 49429147-49429148 on Build GRCh38
rs144096179 Chr.3: 49429061-49429061 on Build GRCh38
rs544384904 Chr.3: 49429040-49429040 on Build GRCh38
rs982047814 Chr.3: 49429289-49429289 on Build GRCh38
rs35470398 Chr.3: 49429053-49429054 on Build GRCh38
rs762840569 Chr.3: 49429208-49429208 on Build GRCh38
rs373149059 Chr.3: 49429057-49429057 on Build GRCh38
rs922047977 Chr.3: 49429047-49429047 on Build GRCh38
rs770800765 Chr.3: 49429283-49429283 on Build GRCh38
rs754596286 Chr.3: 49429065-49429065 on Build GRCh38
rs952486937 Chr.3: 49428954-49428954 on Build GRCh38
rs759199070 Chr.3: 49429288-49429288 on Build GRCh38
rs975002930 Chr.3: 49429302-49429302 on Build GRCh38
rs988696125 Chr.3: 49429263-49429263 on Build GRCh38
rs575205701 Chr.3: 49429161-49429161 on Build GRCh38
rs955666117 Chr.3: 49429261-49429261 on Build GRCh38
rs746064312 Chr.3: 49429083-49429083 on Build GRCh38
rs988664940 Chr.3: 49428947-49428947 on Build GRCh38
rs952372297 Chr.3: 49429243-49429243 on Build GRCh38
rs984847948 Chr.3: 49429338-49429338 on Build GRCh38
rs747076385 Chr.3: 49429103-49429103 on Build GRCh38
rs756954104 Chr.3: 49429032-49429032 on Build GRCh38
rs11130191 Chr.3: 49429203-49429203 on Build GRCh38
rs768697137 Chr.3: 49429195-49429195 on Build GRCh38
rs762639505 Chr.3: 49429142-49429142 on Build GRCh38
rs780446039 Chr.3: 49429256-49429256 on Build GRCh38
rs1040979365 Chr.3: 49429172-49429172 on Build GRCh38
rs774319932 Chr.3: 49429284-49429284 on Build GRCh38
rs921771229 Chr.3: 49429307-49429307 on Build GRCh38
rs758730331 Chr.3: 49429253-49429253 on Build GRCh38
rs766321646 Chr.3: 49429213-49429213 on Build GRCh38
rs747180776 Chr.3: 49429257-49429257 on Build GRCh38
rs768791212 Chr.3: 49429110-49429110 on Build GRCh38
rs546312930 Chr.3: 49429122-49429122 on Build GRCh38
rs759284971 Chr.3: 49429233-49429233 on Build GRCh38
rs757726620 Chr.3: 49429245-49429245 on Build GRCh38
rs930724249 Chr.3: 49429050-49429050 on Build GRCh38
rs752154466 Chr.3: 49429244-49429244 on Build GRCh38
rs781263968 Chr.3: 49429272-49429272 on Build GRCh38
rs772287234 Chr.3: 49429095-49429095 on Build GRCh38
rs758818324 Chr.3: 49429185-49429185 on Build GRCh38
rs546983508 Chr.3: 49429168-49429168 on Build GRCh38
rs774129151 Chr.3: 49429222-49429222 on Build GRCh38
rs907885248 Chr.3: 49429386-49429386 on Build GRCh38
rs755121327 Chr.3: 49429150-49429151 on Build GRCh38
rs751327956 Chr.3: 49429156-49429156 on Build GRCh38
rs755183254 Chr.3: 49429262-49429262 on Build GRCh38
rs142635342 Chr.3: 49429266-49429267 on Build GRCh38
rs990525983 Chr.3: 49428960-49428960 on Build GRCh38
rs529565084 Chr.3: 49429275-49429275 on Build GRCh38
rs767013283 Chr.3: 49429238-49429238 on Build GRCh38
rs914583933 Chr.3: 49429338-49429338 on Build GRCh38
rs200333545 Chr.3: 49429072-49429072 on Build GRCh38
rs192878512 Chr.3: 49429378-49429378 on Build GRCh38
rs146341285 Chr.3: 49429319-49429319 on Build GRCh38
rs905638462 Chr.3: 49429183-49429183 on Build GRCh38

View Full Product Details