Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49684861-49685348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MST1
Entrez Gene ID
4485
Gene Name
macrophage stimulating 1
Gene Aliases
D3F15S2, DNF15S2, HGFL, MSP, NF15S2
UniGene
Hs.349110
Gene Chromosome Location
Chr.3: 49683947-49689528 on Build GRCh38
Transcripts
NM_020998.3 XM_006713166.1 XM_011533732.1 XM_011533737.2 XM_011533738.2 XM_017006460.1 XM_017006461.1 XM_017006462.1 XM_017006463.1 XM_017006464.1 XM_017006465.1 XM_017006466.1 XM_017006467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7218982 c.1537C>G p.R513G Chr.3: 49685269-49685269 on Build GRCh38
1600014 c.1565G>C p.G522A Chr.3: 49685069-49685069 on Build GRCh38
4149959 c.1565G>A p.G522E Chr.3: 49685069-49685069 on Build GRCh38
2850289 c.1603delC p.R535fs*37 Chr.3: 49685031-49685031 on Build GRCh38
5355208 c.1537C>T p.R513W Chr.3: 49685269-49685269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753640826 Chr.3: 49684904-49684904 on Build GRCh38
rs893976398 Chr.3: 49685256-49685256 on Build GRCh38
rs756580124 Chr.3: 49685071-49685071 on Build GRCh38
rs760682000 Chr.3: 49685129-49685129 on Build GRCh38
rs746379716 Chr.3: 49685220-49685221 on Build GRCh38
rs62262682 Chr.3: 49685328-49685328 on Build GRCh38
rs759572668 Chr.3: 49685002-49685002 on Build GRCh38
rs1057287 Chr.3: 49685314-49685314 on Build GRCh38
rs1053953192 Chr.3: 49685191-49685191 on Build GRCh38
rs764938913 Chr.3: 49685052-49685052 on Build GRCh38
rs2476570 Chr.3: 49685116-49685116 on Build GRCh38
rs746068925 Chr.3: 49685115-49685115 on Build GRCh38
rs916894623 Chr.3: 49685136-49685136 on Build GRCh38
rs773076864 Chr.3: 49685302-49685302 on Build GRCh38
rs71262351 Chr.3: 49685269-49685269 on Build GRCh38
rs3882326 Chr.3: 49685201-49685201 on Build GRCh38
rs750299218 Chr.3: 49685238-49685238 on Build GRCh38
rs561328863 Chr.3: 49685107-49685107 on Build GRCh38
rs572177192 Chr.3: 49685035-49685035 on Build GRCh38
rs768583817 Chr.3: 49685282-49685282 on Build GRCh38
rs749307562 Chr.3: 49685064-49685065 on Build GRCh38
rs756275887 Chr.3: 49684917-49684918 on Build GRCh38
rs776461231 Chr.3: 49685138-49685138 on Build GRCh38
rs3882323 Chr.3: 49685330-49685330 on Build GRCh38
rs542749939 Chr.3: 49685036-49685036 on Build GRCh38
rs780161891 Chr.3: 49685274-49685274 on Build GRCh38
rs552761128 Chr.3: 49684944-49684944 on Build GRCh38
rs771316312 Chr.3: 49685098-49685098 on Build GRCh38
rs372625101 Chr.3: 49685297-49685297 on Build GRCh38
rs749609961 Chr.3: 49685096-49685096 on Build GRCh38
rs778721577 Chr.3: 49684926-49684926 on Build GRCh38
rs769378535 Chr.3: 49684927-49684927 on Build GRCh38
rs377097129 Chr.3: 49685315-49685315 on Build GRCh38
rs2933247 Chr.3: 49685176-49685176 on Build GRCh38
rs771087532 Chr.3: 49684978-49684978 on Build GRCh38
rs779467629 Chr.3: 49684934-49684934 on Build GRCh38
rs565400329 Chr.3: 49685279-49685279 on Build GRCh38
rs778160360 Chr.3: 49685094-49685094 on Build GRCh38
rs748664501 Chr.3: 49684962-49684962 on Build GRCh38
rs779354045 Chr.3: 49685099-49685100 on Build GRCh38
rs769916597 Chr.3: 49685299-49685299 on Build GRCh38
rs757380950 Chr.3: 49685070-49685071 on Build GRCh38
rs750066095 Chr.3: 49685059-49685059 on Build GRCh38
rs920456984 Chr.3: 49685039-49685039 on Build GRCh38
rs761826355 Chr.3: 49685298-49685298 on Build GRCh38
rs778303900 Chr.3: 49685262-49685262 on Build GRCh38
rs576408730 Chr.3: 49685169-49685169 on Build GRCh38
rs774579761 Chr.3: 49684998-49684998 on Build GRCh38
rs201558559 Chr.3: 49685067-49685067 on Build GRCh38
rs902746502 Chr.3: 49685179-49685179 on Build GRCh38
rs749623546 Chr.3: 49684972-49684972 on Build GRCh38
rs779600056 Chr.3: 49685070-49685070 on Build GRCh38
rs765954901 Chr.3: 49685247-49685247 on Build GRCh38
rs757139258 Chr.3: 49685037-49685037 on Build GRCh38
rs534468601 Chr.3: 49684883-49684883 on Build GRCh38
rs775570550 Chr.3: 49685125-49685125 on Build GRCh38
rs756046746 Chr.3: 49684890-49684890 on Build GRCh38
rs200437638 Chr.3: 49685249-49685249 on Build GRCh38
rs765098610 Chr.3: 49685236-49685236 on Build GRCh38
rs772242763 Chr.3: 49685118-49685118 on Build GRCh38
rs767555694 Chr.3: 49685030-49685030 on Build GRCh38
rs766915584 Chr.3: 49685327-49685327 on Build GRCh38
rs987318672 Chr.3: 49684906-49684906 on Build GRCh38
rs760656029 Chr.3: 49685031-49685031 on Build GRCh38
rs149138392 Chr.3: 49685273-49685273 on Build GRCh38
rs754312726 Chr.3: 49685263-49685263 on Build GRCh38
rs935622522 Chr.3: 49685183-49685183 on Build GRCh38
rs150169514 Chr.3: 49685329-49685329 on Build GRCh38
rs747070915 Chr.3: 49685275-49685275 on Build GRCh38
rs142344838 Chr.3: 49685326-49685326 on Build GRCh38
rs772368903 Chr.3: 49685279-49685280 on Build GRCh38
rs3882325 Chr.3: 49685310-49685310 on Build GRCh38
rs201631890 Chr.3: 49685295-49685295 on Build GRCh38
rs778122157 Chr.3: 49685013-49685015 on Build GRCh38
rs1041434662 Chr.3: 49685172-49685172 on Build GRCh38
rs4052589 Chr.3: 49684964-49684964 on Build GRCh38
rs762692684 Chr.3: 49685245-49685245 on Build GRCh38
rs375438838 Chr.3: 49685268-49685268 on Build GRCh38
rs199553610 Chr.3: 49685316-49685316 on Build GRCh38
rs754473083 Chr.3: 49685259-49685259 on Build GRCh38
rs9837520 Chr.3: 49684923-49684923 on Build GRCh38
rs371390810 Chr.3: 49685018-49685018 on Build GRCh38
rs761879370 Chr.3: 49685230-49685230 on Build GRCh38
rs758009791 Chr.3: 49684932-49684932 on Build GRCh38
rs768589445 Chr.3: 49685134-49685134 on Build GRCh38
rs763928830 Chr.3: 49684899-49684899 on Build GRCh38

View Full Product Details