Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50173668-50174161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3F
Entrez Gene ID
6405
Gene Name
semaphorin 3F
Gene Aliases
SEMA-IV, SEMA4, SEMAK
UniGene
Hs.32981
Gene Chromosome Location
Chr.3: 50155045-50189075 on Build GRCh38
Transcripts
NM_001318798.1 NM_001318800.1 NM_004186.4 XM_005265381.4 XM_005265382.4 XM_006713290.3 XM_011533998.2 XM_011534000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1046415 c.198C>T p.H66H Chr.3: 50173878-50173878 on Build GRCh38
4754948 c.136C>G p.H46D Chr.3: 50173816-50173816 on Build GRCh38
1046416 c.198_203delCGACCG p.D67_R68delDR Chr.3: 50173878-50173883 on Build GRCh38
6322640 c.336+9G>T p.? Chr.3: 50174123-50174123 on Build GRCh38
1753292 c.177C>T p.I59I Chr.3: 50173857-50173857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542825031 Chr.3: 50173695-50173695 on Build GRCh38
rs867418847 Chr.3: 50173734-50173734 on Build GRCh38
rs200472848 Chr.3: 50173753-50173753 on Build GRCh38
rs765182452 Chr.3: 50174040-50174040 on Build GRCh38
rs756077250 Chr.3: 50173812-50173812 on Build GRCh38
rs929853541 Chr.3: 50173784-50173784 on Build GRCh38
rs147135053 Chr.3: 50173813-50173813 on Build GRCh38
rs780910563 Chr.3: 50173990-50173990 on Build GRCh38
rs750334812 Chr.3: 50174041-50174041 on Build GRCh38
rs146858928 Chr.3: 50174091-50174091 on Build GRCh38
rs367874618 Chr.3: 50173819-50173819 on Build GRCh38
rs997899343 Chr.3: 50174093-50174093 on Build GRCh38
rs774926819 Chr.3: 50173942-50173942 on Build GRCh38
rs756191868 Chr.3: 50173887-50173887 on Build GRCh38
rs139093472 Chr.3: 50174090-50174090 on Build GRCh38
rs200876993 Chr.3: 50173978-50173987 on Build GRCh38
rs746523630 Chr.3: 50173930-50173930 on Build GRCh38
rs751847501 Chr.3: 50173870-50173870 on Build GRCh38
rs746237265 Chr.3: 50174003-50174003 on Build GRCh38
rs754747484 Chr.3: 50174047-50174047 on Build GRCh38
rs532137565 Chr.3: 50173689-50173689 on Build GRCh38
rs776757060 Chr.3: 50174037-50174037 on Build GRCh38
rs368468478 Chr.3: 50173761-50173761 on Build GRCh38
rs371508526 Chr.3: 50173909-50173909 on Build GRCh38
rs757048607 Chr.3: 50173908-50173908 on Build GRCh38
rs138196965 Chr.3: 50173795-50173795 on Build GRCh38
rs775729500 Chr.3: 50173952-50173952 on Build GRCh38
rs760150530 Chr.3: 50174106-50174106 on Build GRCh38
rs767779495 Chr.3: 50173879-50173879 on Build GRCh38
rs201333607 Chr.3: 50173978-50173978 on Build GRCh38
rs773862039 Chr.3: 50173939-50173939 on Build GRCh38
rs759067419 Chr.3: 50173742-50173742 on Build GRCh38
rs777707294 Chr.3: 50173890-50173890 on Build GRCh38
rs760227157 Chr.3: 50173845-50173845 on Build GRCh38
rs143205421 Chr.3: 50173693-50173693 on Build GRCh38
rs761912679 Chr.3: 50174124-50174124 on Build GRCh38
rs747660442 Chr.3: 50173991-50173991 on Build GRCh38
rs755092522 Chr.3: 50173783-50173783 on Build GRCh38
rs781357100 Chr.3: 50173992-50173992 on Build GRCh38
rs1027865253 Chr.3: 50173743-50173743 on Build GRCh38
rs750432231 Chr.3: 50173838-50173838 on Build GRCh38
rs745455419 Chr.3: 50173835-50173835 on Build GRCh38
rs768172772 Chr.3: 50173935-50173935 on Build GRCh38
rs763442956 Chr.3: 50173853-50173853 on Build GRCh38
rs1005611446 Chr.3: 50174061-50174061 on Build GRCh38
rs770253654 Chr.3: 50173846-50173846 on Build GRCh38
rs752308945 Chr.3: 50174064-50174064 on Build GRCh38
rs749757541 Chr.3: 50173940-50173940 on Build GRCh38
rs991248742 Chr.3: 50174082-50174082 on Build GRCh38
rs548177299 Chr.3: 50173864-50173864 on Build GRCh38
rs374780500 Chr.3: 50173961-50173961 on Build GRCh38
rs764091820 Chr.3: 50173972-50173972 on Build GRCh38
rs779604849 Chr.3: 50173983-50173983 on Build GRCh38
rs745661919 Chr.3: 50173928-50173928 on Build GRCh38
rs376670399 Chr.3: 50174069-50174069 on Build GRCh38
rs986734979 Chr.3: 50173730-50173730 on Build GRCh38
rs771441394 Chr.3: 50173747-50173747 on Build GRCh38
rs1026296238 Chr.3: 50173996-50173996 on Build GRCh38
rs982550289 Chr.3: 50174038-50174038 on Build GRCh38
rs747370824 Chr.3: 50174094-50174094 on Build GRCh38
rs368006800 Chr.3: 50173836-50173836 on Build GRCh38
rs753671263 Chr.3: 50173723-50173723 on Build GRCh38
rs569980926 Chr.3: 50173878-50173878 on Build GRCh38
rs754551257 Chr.3: 50173986-50173986 on Build GRCh38
rs757242592 Chr.3: 50173977-50173977 on Build GRCh38
rs758164070 Chr.3: 50173981-50173981 on Build GRCh38
rs369738643 Chr.3: 50174114-50174114 on Build GRCh38
rs772439222 Chr.3: 50174016-50174016 on Build GRCh38
rs775576962 Chr.3: 50174018-50174018 on Build GRCh38
rs150737136 Chr.3: 50174054-50174054 on Build GRCh38
rs374355347 Chr.3: 50174042-50174042 on Build GRCh38
rs368839186 Chr.3: 50173976-50173976 on Build GRCh38
rs761849508 Chr.3: 50174039-50174039 on Build GRCh38
rs937182132 Chr.3: 50173818-50173818 on Build GRCh38
rs750859168 Chr.3: 50173767-50173767 on Build GRCh38
rs766731625 Chr.3: 50173773-50173773 on Build GRCh38
rs763388968 Chr.3: 50173768-50173768 on Build GRCh38
rs985333083 Chr.3: 50173883-50173883 on Build GRCh38
rs758053222 Chr.3: 50173929-50173929 on Build GRCh38
rs112798318 Chr.3: 50173787-50173787 on Build GRCh38
rs766065309 Chr.3: 50174129-50174149 on Build GRCh38
rs759918248 Chr.3: 50173947-50173947 on Build GRCh38
rs764286271 Chr.3: 50174030-50174030 on Build GRCh38
rs372379422 Chr.3: 50173781-50173781 on Build GRCh38
rs777198413 Chr.3: 50173748-50173748 on Build GRCh38
rs1041428936 Chr.3: 50174100-50174100 on Build GRCh38
rs150847709 Chr.3: 50174081-50174081 on Build GRCh38
rs751272757 Chr.3: 50174128-50174129 on Build GRCh38
rs757181901 Chr.3: 50173737-50173737 on Build GRCh38
rs774920210 Chr.3: 50173839-50173839 on Build GRCh38
rs778564860 Chr.3: 50173826-50173826 on Build GRCh38
rs554250122 Chr.3: 50174077-50174077 on Build GRCh38
rs767677743 Chr.3: 50173950-50173950 on Build GRCh38
rs970735446 Chr.3: 50174013-50174013 on Build GRCh38
rs953551401 Chr.3: 50173727-50173727 on Build GRCh38
rs753733068 Chr.3: 50173891-50173891 on Build GRCh38
rs760892130 Chr.3: 50174027-50174027 on Build GRCh38
rs750224845 Chr.3: 50173979-50173979 on Build GRCh38
rs774981923 Chr.3: 50174000-50174000 on Build GRCh38
rs752763527 Chr.3: 50173882-50173882 on Build GRCh38
rs770660254 Chr.3: 50173821-50173821 on Build GRCh38
rs771554773 Chr.3: 50173941-50173941 on Build GRCh38

View Full Product Details