Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50580706-50581214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEMK1
Entrez Gene ID
51409
Gene Name
HemK methyltransferase family member 1
Gene Aliases
HEMK, MTQ1
UniGene
Hs.517987
Gene Chromosome Location
Chr.3: 50569152-50589050 on Build GRCh38
Transcripts
NM_001317851.1 NM_016173.4 XM_005265218.3 XM_011533806.2 XM_011533807.1 XM_011533808.1 XM_011533809.2 XM_011533810.2 XM_011533812.2 XM_011533813.2 XM_011533814.2

SNP Detail

SNP ID Chromosome Location
rs1020566987 Chr.3: 50580954-50580954 on Build GRCh38
rs990039153 Chr.3: 50580747-50580747 on Build GRCh38
rs955385396 Chr.3: 50581084-50581084 on Build GRCh38
rs892951131 Chr.3: 50580907-50580907 on Build GRCh38
rs922629662 Chr.3: 50580769-50580769 on Build GRCh38
rs1034182525 Chr.3: 50580739-50580739 on Build GRCh38
rs373935758 Chr.3: 50581137-50581137 on Build GRCh38
rs537778954 Chr.3: 50581127-50581127 on Build GRCh38
rs911322840 Chr.3: 50581092-50581092 on Build GRCh38
rs34988368 Chr.3: 50581173-50581174 on Build GRCh38
rs376030389 Chr.3: 50581148-50581148 on Build GRCh38
rs1045393529 Chr.3: 50580885-50580885 on Build GRCh38
rs1020219150 Chr.3: 50580749-50580749 on Build GRCh38
rs941542959 Chr.3: 50580829-50580829 on Build GRCh38
rs906902132 Chr.3: 50580889-50580889 on Build GRCh38
rs559679250 Chr.3: 50580925-50580925 on Build GRCh38
rs906314016 Chr.3: 50581010-50581010 on Build GRCh38
rs770546350 Chr.3: 50581059-50581059 on Build GRCh38
rs563778776 Chr.3: 50580873-50580873 on Build GRCh38
rs757079781 Chr.3: 50580770-50580770 on Build GRCh38
rs1038563080 Chr.3: 50580836-50580836 on Build GRCh38
rs192833117 Chr.3: 50580908-50580908 on Build GRCh38
rs552870159 Chr.3: 50580750-50580750 on Build GRCh38
rs530282025 Chr.3: 50580974-50580974 on Build GRCh38
rs528170867 Chr.3: 50580887-50580887 on Build GRCh38
rs1001869281 Chr.3: 50581036-50581036 on Build GRCh38
rs541673146 Chr.3: 50580777-50580777 on Build GRCh38
rs185015346 Chr.3: 50581109-50581109 on Build GRCh38
rs35121974 Chr.3: 50581074-50581074 on Build GRCh38
rs987592041 Chr.3: 50580767-50580767 on Build GRCh38
rs563569580 Chr.3: 50580794-50580794 on Build GRCh38
rs1034107545 Chr.3: 50580895-50580895 on Build GRCh38
rs943361187 Chr.3: 50580778-50580778 on Build GRCh38
rs967315188 Chr.3: 50580964-50580964 on Build GRCh38
rs1001244843 Chr.3: 50580892-50580892 on Build GRCh38
rs957193936 Chr.3: 50580745-50580745 on Build GRCh38
rs1024715844 Chr.3: 50580975-50580975 on Build GRCh38
rs147675339 Chr.3: 50581149-50581149 on Build GRCh38
rs943458994 Chr.3: 50581160-50581160 on Build GRCh38
rs985478378 Chr.3: 50581089-50581089 on Build GRCh38
rs149017134 Chr.3: 50580764-50580764 on Build GRCh38
rs545353691 Chr.3: 50580833-50580833 on Build GRCh38
rs746686162 Chr.3: 50580924-50580924 on Build GRCh38
rs745725342 Chr.3: 50580858-50580858 on Build GRCh38
rs145340689 Chr.3: 50581085-50581085 on Build GRCh38
rs950561735 Chr.3: 50581129-50581129 on Build GRCh38
rs891981627 Chr.3: 50580937-50580937 on Build GRCh38
rs1046407056 Chr.3: 50581145-50581145 on Build GRCh38
rs780110343 Chr.3: 50581106-50581106 on Build GRCh38
rs552599834 Chr.3: 50581132-50581132 on Build GRCh38
rs190323009 Chr.3: 50580797-50580797 on Build GRCh38
rs930247696 Chr.3: 50580864-50580864 on Build GRCh38

View Full Product Details