Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51633821-51634319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD54L2
Entrez Gene ID
23132
Gene Name
RAD54-like 2 (S. cerevisiae)
Gene Aliases
ARIP4, HSPC325, SRISNF2L
UniGene
Hs.105399
Gene Chromosome Location
Chr.3: 51538683-51668660 on Build GRCh38
Transcripts
NM_001322253.1 NM_001322256.1 NM_001322257.1 NM_015106.3 XM_006713062.3 XM_017005987.1 XM_017005988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7418125 c.1009-1G>T p.? Chr.3: 51633901-51633901 on Build GRCh38
5899950 c.1099C>T p.Q367* Chr.3: 51633992-51633992 on Build GRCh38
6893231 c.1036G>A p.E346K Chr.3: 51633929-51633929 on Build GRCh38
6761979 c.1106G>A p.R369Q Chr.3: 51633999-51633999 on Build GRCh38
7336580 c.1108T>A p.F370I Chr.3: 51634001-51634001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766884755 Chr.3: 51633919-51633919 on Build GRCh38
rs967805024 Chr.3: 51634270-51634270 on Build GRCh38
rs143648072 Chr.3: 51634003-51634003 on Build GRCh38
rs767307034 Chr.3: 51633873-51633874 on Build GRCh38
rs749768111 Chr.3: 51633988-51633988 on Build GRCh38
rs149414610 Chr.3: 51633907-51633907 on Build GRCh38
rs747159383 Chr.3: 51633856-51633856 on Build GRCh38
rs1043217229 Chr.3: 51633878-51633878 on Build GRCh38
rs990436121 Chr.3: 51634201-51634201 on Build GRCh38
rs9844541 Chr.3: 51634139-51634139 on Build GRCh38
rs200135857 Chr.3: 51634043-51634043 on Build GRCh38
rs76085189 Chr.3: 51634086-51634086 on Build GRCh38
rs373159206 Chr.3: 51634087-51634087 on Build GRCh38
rs749976872 Chr.3: 51633928-51633928 on Build GRCh38
rs752584384 Chr.3: 51633964-51633964 on Build GRCh38
rs776014971 Chr.3: 51633983-51633983 on Build GRCh38
rs763601558 Chr.3: 51633899-51633899 on Build GRCh38
rs72953404 Chr.3: 51634282-51634282 on Build GRCh38
rs764218181 Chr.3: 51634133-51634133 on Build GRCh38
rs781475410 Chr.3: 51633969-51633969 on Build GRCh38
rs759210694 Chr.3: 51634044-51634044 on Build GRCh38
rs777931887 Chr.3: 51633854-51633854 on Build GRCh38
rs770352830 Chr.3: 51633886-51633886 on Build GRCh38
rs960397649 Chr.3: 51634181-51634181 on Build GRCh38
rs752494598 Chr.3: 51634065-51634065 on Build GRCh38
rs916359329 Chr.3: 51634231-51634231 on Build GRCh38
rs573595419 Chr.3: 51634089-51634089 on Build GRCh38
rs114438035 Chr.3: 51633966-51633966 on Build GRCh38
rs554579923 Chr.3: 51634104-51634104 on Build GRCh38
rs746129258 Chr.3: 51633974-51633974 on Build GRCh38
rs191008861 Chr.3: 51633954-51633954 on Build GRCh38
rs776106693 Chr.3: 51633896-51633896 on Build GRCh38
rs760216853 Chr.3: 51634016-51634016 on Build GRCh38
rs760178765 Chr.3: 51633936-51633936 on Build GRCh38
rs916326454 Chr.3: 51633848-51633848 on Build GRCh38
rs148593384 Chr.3: 51633951-51633951 on Build GRCh38
rs915681800 Chr.3: 51634055-51634055 on Build GRCh38
rs377016156 Chr.3: 51634023-51634023 on Build GRCh38
rs759968069 Chr.3: 51633877-51633877 on Build GRCh38
rs769209062 Chr.3: 51633993-51633993 on Build GRCh38
rs200783382 Chr.3: 51633967-51633967 on Build GRCh38
rs778730370 Chr.3: 51633960-51633960 on Build GRCh38
rs531798001 Chr.3: 51633866-51633866 on Build GRCh38
rs886815435 Chr.3: 51634110-51634111 on Build GRCh38
rs370238700 Chr.3: 51633873-51633873 on Build GRCh38
rs61101419 Chr.3: 51633916-51633916 on Build GRCh38
rs540869048 Chr.3: 51634125-51634125 on Build GRCh38
rs901403022 Chr.3: 51634155-51634155 on Build GRCh38
rs770258894 Chr.3: 51633976-51633976 on Build GRCh38
rs766068319 Chr.3: 51633938-51633938 on Build GRCh38
rs764976433 Chr.3: 51634063-51634063 on Build GRCh38
rs758277201 Chr.3: 51634081-51634081 on Build GRCh38

View Full Product Details