Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52199003-52199498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAS1
Entrez Gene ID
211
Gene Name
5'-aminolevulinate synthase 1
Gene Aliases
ALAS, ALAS-H, ALAS3, ALASH, MIG4
UniGene
Hs.476308
Gene Chromosome Location
Chr.3: 52198083-52214327 on Build GRCh38
Transcripts
NM_000688.5 NM_001304443.1 NM_001304444.1 NM_199166.2 XM_011533477.1 XM_011533478.2 XM_017005873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1309318 c.41T>G p.V14G Chr.3: 52199282-52199282 on Build GRCh38
7140604 c.60G>C p.Q20H Chr.3: 52199301-52199301 on Build GRCh38
2852575 c.20G>A p.R7H Chr.3: 52199261-52199261 on Build GRCh38
6598090 c.182C>T p.T61I Chr.3: 52199423-52199423 on Build GRCh38
1046818 c.79T>C p.L27L Chr.3: 52199320-52199320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760777659 Chr.3: 52199406-52199406 on Build GRCh38
rs143432678 Chr.3: 52199215-52199215 on Build GRCh38
rs139865127 Chr.3: 52199357-52199357 on Build GRCh38
rs749512727 Chr.3: 52199350-52199350 on Build GRCh38
rs568005929 Chr.3: 52199053-52199053 on Build GRCh38
rs202007599 Chr.3: 52199374-52199374 on Build GRCh38
rs528585181 Chr.3: 52199262-52199262 on Build GRCh38
rs61729068 Chr.3: 52199368-52199368 on Build GRCh38
rs778316387 Chr.3: 52199431-52199431 on Build GRCh38
rs185535407 Chr.3: 52199460-52199460 on Build GRCh38
rs374909536 Chr.3: 52199202-52199202 on Build GRCh38
rs148819009 Chr.3: 52199042-52199042 on Build GRCh38
rs779110321 Chr.3: 52199417-52199417 on Build GRCh38
rs35338461 Chr.3: 52199279-52199279 on Build GRCh38
rs767362905 Chr.3: 52199322-52199322 on Build GRCh38
rs780318962 Chr.3: 52199348-52199348 on Build GRCh38
rs745382348 Chr.3: 52199366-52199366 on Build GRCh38
rs368153991 Chr.3: 52199246-52199246 on Build GRCh38
rs1026479551 Chr.3: 52199160-52199160 on Build GRCh38
rs151190937 Chr.3: 52199390-52199390 on Build GRCh38
rs371816029 Chr.3: 52199459-52199459 on Build GRCh38
rs774896720 Chr.3: 52199207-52199207 on Build GRCh38
rs542849183 Chr.3: 52199131-52199131 on Build GRCh38
rs535433738 Chr.3: 52199150-52199150 on Build GRCh38
rs761093598 Chr.3: 52199230-52199230 on Build GRCh38
rs769934281 Chr.3: 52199186-52199186 on Build GRCh38
rs768454292 Chr.3: 52199377-52199377 on Build GRCh38
rs752999300 Chr.3: 52199428-52199428 on Build GRCh38
rs772498476 Chr.3: 52199358-52199358 on Build GRCh38
rs775075760 Chr.3: 52199283-52199283 on Build GRCh38
rs142401935 Chr.3: 52199259-52199259 on Build GRCh38
rs758970433 Chr.3: 52199275-52199275 on Build GRCh38
rs776120817 Chr.3: 52199229-52199229 on Build GRCh38
rs769075509 Chr.3: 52199261-52199261 on Build GRCh38
rs757455604 Chr.3: 52199468-52199469 on Build GRCh38
rs78520106 Chr.3: 52199035-52199035 on Build GRCh38
rs758758646 Chr.3: 52199430-52199430 on Build GRCh38
rs746150701 Chr.3: 52199183-52199183 on Build GRCh38
rs993321793 Chr.3: 52199037-52199037 on Build GRCh38
rs765911192 Chr.3: 52199223-52199223 on Build GRCh38
rs759176551 Chr.3: 52199243-52199243 on Build GRCh38
rs781372666 Chr.3: 52199258-52199258 on Build GRCh38
rs780151880 Chr.3: 52199260-52199260 on Build GRCh38
rs755148387 Chr.3: 52199415-52199415 on Build GRCh38
rs368742565 Chr.3: 52199285-52199285 on Build GRCh38
rs182395582 Chr.3: 52199364-52199364 on Build GRCh38
rs994104836 Chr.3: 52199156-52199156 on Build GRCh38
rs779391579 Chr.3: 52199454-52199454 on Build GRCh38
rs144906218 Chr.3: 52199289-52199289 on Build GRCh38
rs748813370 Chr.3: 52199455-52199455 on Build GRCh38
rs776164943 Chr.3: 52199271-52199271 on Build GRCh38
rs756204390 Chr.3: 52199341-52199341 on Build GRCh38
rs144022954 Chr.3: 52199434-52199434 on Build GRCh38
rs192989030 Chr.3: 52199204-52199204 on Build GRCh38
rs753933485 Chr.3: 52199410-52199410 on Build GRCh38
rs753520796 Chr.3: 52199256-52199256 on Build GRCh38
rs562566359 Chr.3: 52199440-52199440 on Build GRCh38
rs768669096 Chr.3: 52199440-52199443 on Build GRCh38
rs761715965 Chr.3: 52199378-52199378 on Build GRCh38
rs750392406 Chr.3: 52199405-52199405 on Build GRCh38
rs977918019 Chr.3: 52199030-52199030 on Build GRCh38
rs750459005 Chr.3: 52199325-52199325 on Build GRCh38
rs926277816 Chr.3: 52199236-52199236 on Build GRCh38
rs906632538 Chr.3: 52199190-52199190 on Build GRCh38
rs757391785 Chr.3: 52199257-52199257 on Build GRCh38
rs1022230397 Chr.3: 52199235-52199235 on Build GRCh38
rs889439279 Chr.3: 52199058-52199058 on Build GRCh38
rs775885258 Chr.3: 52199201-52199201 on Build GRCh38
rs527889142 Chr.3: 52199249-52199249 on Build GRCh38
rs774202996 Chr.3: 52199462-52199462 on Build GRCh38
rs150932815 Chr.3: 52199248-52199248 on Build GRCh38
rs61729067 Chr.3: 52199371-52199371 on Build GRCh38
rs762814249 Chr.3: 52199284-52199284 on Build GRCh38
rs374359979 Chr.3: 52199254-52199254 on Build GRCh38
rs1007466272 Chr.3: 52199278-52199278 on Build GRCh38
rs1037917462 Chr.3: 52199133-52199133 on Build GRCh38
rs766417411 Chr.3: 52199409-52199409 on Build GRCh38
rs895199424 Chr.3: 52199424-52199424 on Build GRCh38
rs757222865 Chr.3: 52199303-52199303 on Build GRCh38
rs899379168 Chr.3: 52199141-52199141 on Build GRCh38
rs557897094 Chr.3: 52199265-52199265 on Build GRCh38
rs145966605 Chr.3: 52199268-52199268 on Build GRCh38
rs755309775 Chr.3: 52199351-52199351 on Build GRCh38
rs1018284135 Chr.3: 52199081-52199081 on Build GRCh38
rs150267006 Chr.3: 52199463-52199463 on Build GRCh38
rs964149227 Chr.3: 52199086-52199086 on Build GRCh38
rs1055218741 Chr.3: 52199389-52199389 on Build GRCh38

View Full Product Details