Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52413906-52414413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF7
Entrez Gene ID
51533
Gene Name
PHD finger protein 7
Gene Aliases
HSPC045, HSPC226, NYD-SP6
UniGene
Hs.372719
Gene Chromosome Location
Chr.3: 52410508-52423641 on Build GRCh38
Transcripts
NM_001278221.2 NM_001321126.1 NM_001321127.1 NM_016483.6 XM_011533826.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2853508 c.94G>T p.V32F Chr.3: 52414048-52414048 on Build GRCh38
6745171 c.50C>T p.A17V Chr.3: 52414004-52414004 on Build GRCh38
7155497 c.83C>T p.S28F Chr.3: 52414037-52414037 on Build GRCh38
2853506 c.80C>T p.P27L Chr.3: 52414034-52414034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570228025 Chr.3: 52414332-52414332 on Build GRCh38
rs973504200 Chr.3: 52413956-52413956 on Build GRCh38
rs200440194 Chr.3: 52414034-52414034 on Build GRCh38
rs139713041 Chr.3: 52414000-52414000 on Build GRCh38
rs186541059 Chr.3: 52414299-52414299 on Build GRCh38
rs375124410 Chr.3: 52414118-52414118 on Build GRCh38
rs751175610 Chr.3: 52414086-52414086 on Build GRCh38
rs771306678 Chr.3: 52414224-52414224 on Build GRCh38
rs918169825 Chr.3: 52413969-52413969 on Build GRCh38
rs73072983 Chr.3: 52414035-52414035 on Build GRCh38
rs371415986 Chr.3: 52413987-52413987 on Build GRCh38
rs763871841 Chr.3: 52414083-52414083 on Build GRCh38
rs954806270 Chr.3: 52414077-52414077 on Build GRCh38
rs190197093 Chr.3: 52414055-52414055 on Build GRCh38
rs145451027 Chr.3: 52414069-52414069 on Build GRCh38
rs867675579 Chr.3: 52414148-52414148 on Build GRCh38
rs745866415 Chr.3: 52414003-52414003 on Build GRCh38
rs979550407 Chr.3: 52414134-52414135 on Build GRCh38
rs1013741957 Chr.3: 52414330-52414330 on Build GRCh38
rs757038634 Chr.3: 52413993-52413993 on Build GRCh38
rs904387030 Chr.3: 52414345-52414345 on Build GRCh38
rs762176516 Chr.3: 52413950-52413950 on Build GRCh38
rs1034254956 Chr.3: 52414012-52414012 on Build GRCh38
rs756837553 Chr.3: 52414093-52414093 on Build GRCh38
rs761334062 Chr.3: 52414038-52414038 on Build GRCh38
rs745750279 Chr.3: 52413964-52413964 on Build GRCh38
rs777512443 Chr.3: 52413985-52413985 on Build GRCh38
rs3832250 Chr.3: 52414267-52414267 on Build GRCh38
rs780173307 Chr.3: 52414017-52414017 on Build GRCh38
rs993398598 Chr.3: 52414153-52414153 on Build GRCh38
rs896577827 Chr.3: 52414306-52414306 on Build GRCh38
rs367664585 Chr.3: 52413982-52413982 on Build GRCh38
rs201971017 Chr.3: 52414022-52414022 on Build GRCh38
rs780757232 Chr.3: 52414305-52414305 on Build GRCh38
rs182829207 Chr.3: 52414194-52414194 on Build GRCh38
rs559165371 Chr.3: 52414373-52414373 on Build GRCh38
rs1040620957 Chr.3: 52414145-52414145 on Build GRCh38
rs993132652 Chr.3: 52414073-52414073 on Build GRCh38
rs113084678 Chr.3: 52414103-52414103 on Build GRCh38
rs887811593 Chr.3: 52414199-52414199 on Build GRCh38
rs762827491 Chr.3: 52413965-52413965 on Build GRCh38
rs904930836 Chr.3: 52414027-52414027 on Build GRCh38
rs925296337 Chr.3: 52413930-52413930 on Build GRCh38
rs1017883186 Chr.3: 52414130-52414132 on Build GRCh38
rs750898978 Chr.3: 52413961-52413961 on Build GRCh38
rs767911287 Chr.3: 52413957-52413957 on Build GRCh38
rs149514196 Chr.3: 52414186-52414186 on Build GRCh38
rs138052405 Chr.3: 52414074-52414074 on Build GRCh38
rs747365943 Chr.3: 52414320-52414320 on Build GRCh38
rs750903529 Chr.3: 52414284-52414284 on Build GRCh38
rs1051880521 Chr.3: 52414302-52414302 on Build GRCh38
rs944530718 Chr.3: 52414136-52414136 on Build GRCh38
rs371292768 Chr.3: 52414157-52414157 on Build GRCh38
rs761069427 Chr.3: 52413975-52413975 on Build GRCh38
rs560257760 Chr.3: 52414025-52414025 on Build GRCh38
rs1032604881 Chr.3: 52414382-52414382 on Build GRCh38
rs966951158 Chr.3: 52414357-52414357 on Build GRCh38
rs1017434494 Chr.3: 52414340-52414340 on Build GRCh38
rs949699625 Chr.3: 52414208-52414208 on Build GRCh38
rs935565167 Chr.3: 52413933-52413933 on Build GRCh38
rs773421228 Chr.3: 52414037-52414037 on Build GRCh38
rs538862468 Chr.3: 52414337-52414337 on Build GRCh38
rs1021157495 Chr.3: 52414344-52414344 on Build GRCh38
rs536330367 Chr.3: 52414298-52414298 on Build GRCh38
rs764883733 Chr.3: 52413978-52413978 on Build GRCh38
rs561185718 Chr.3: 52414310-52414310 on Build GRCh38
rs879889445 Chr.3: 52414338-52414338 on Build GRCh38
rs375153940 Chr.3: 52414051-52414051 on Build GRCh38
rs926825197 Chr.3: 52414266-52414267 on Build GRCh38
rs769687551 Chr.3: 52414011-52414011 on Build GRCh38
rs1050150421 Chr.3: 52414119-52414119 on Build GRCh38
rs766341138 Chr.3: 52413981-52413981 on Build GRCh38
rs1045219564 Chr.3: 52414260-52414260 on Build GRCh38
rs1037380621 Chr.3: 52414267-52414268 on Build GRCh38
rs571458546 Chr.3: 52414132-52414132 on Build GRCh38
rs1052720129 Chr.3: 52413955-52413955 on Build GRCh38

View Full Product Details