Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108695624-108696133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF38B
Entrez Gene ID
55119
Gene Name
pre-mRNA processing factor 38B
Gene Aliases
NET1
UniGene
Hs.342307
Gene Chromosome Location
Chr.1: 108692292-108701803 on Build GRCh38
Transcripts
NM_018061.2 NR_037185.1 XM_011541687.2 XM_017001596.1 XM_017001597.1 XM_017001598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4517755 c.410_411CC>TT p.T137I Chr.1: 108696107-108696108 on Build GRCh38
1332310 c.282G>A p.T94T Chr.1: 108695707-108695707 on Build GRCh38
6507585 c.333G>A p.G111G Chr.1: 108695758-108695758 on Build GRCh38
1688201 c.410C>T p.T137I Chr.1: 108696107-108696107 on Build GRCh38
1747581 c.435G>C p.V145V Chr.1: 108696132-108696132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768091839 Chr.1: 108695784-108695784 on Build GRCh38
rs755700721 Chr.1: 108695728-108695728 on Build GRCh38
rs188372092 Chr.1: 108696016-108696016 on Build GRCh38
rs752168102 Chr.1: 108696000-108696000 on Build GRCh38
rs765396046 Chr.1: 108695677-108695677 on Build GRCh38
rs1052685873 Chr.1: 108695822-108695822 on Build GRCh38
rs764728526 Chr.1: 108695694-108695694 on Build GRCh38
rs771967199 Chr.1: 108695800-108695800 on Build GRCh38
rs368061409 Chr.1: 108695692-108695692 on Build GRCh38
rs140358618 Chr.1: 108695707-108695707 on Build GRCh38
rs752797820 Chr.1: 108695689-108695689 on Build GRCh38
rs767162665 Chr.1: 108695666-108695666 on Build GRCh38
rs760925921 Chr.1: 108695892-108695892 on Build GRCh38
rs143538277 Chr.1: 108695809-108695809 on Build GRCh38
rs914254655 Chr.1: 108696054-108696054 on Build GRCh38
rs566135720 Chr.1: 108695853-108695853 on Build GRCh38
rs528613348 Chr.1: 108695885-108695885 on Build GRCh38
rs778393003 Chr.1: 108696036-108696036 on Build GRCh38
rs373088534 Chr.1: 108695746-108695746 on Build GRCh38
rs768607377 Chr.1: 108695654-108695654 on Build GRCh38
rs953934819 Chr.1: 108695953-108695953 on Build GRCh38
rs769758722 Chr.1: 108696037-108696038 on Build GRCh38
rs775814607 Chr.1: 108695819-108695819 on Build GRCh38
rs946999297 Chr.1: 108695836-108695836 on Build GRCh38
rs370201352 Chr.1: 108695808-108695808 on Build GRCh38
rs183393363 Chr.1: 108695790-108695790 on Build GRCh38
rs761626914 Chr.1: 108695814-108695816 on Build GRCh38
rs781671370 Chr.1: 108695966-108695966 on Build GRCh38
rs759670190 Chr.1: 108695673-108695673 on Build GRCh38
rs898371110 Chr.1: 108695924-108695924 on Build GRCh38
rs373034626 Chr.1: 108695858-108695858 on Build GRCh38
rs753435148 Chr.1: 108696030-108696030 on Build GRCh38
rs752045275 Chr.1: 108695695-108695695 on Build GRCh38
rs1056502186 Chr.1: 108695879-108695879 on Build GRCh38
rs202150869 Chr.1: 108696029-108696029 on Build GRCh38
rs750776863 Chr.1: 108696027-108696027 on Build GRCh38
rs748153821 Chr.1: 108696090-108696090 on Build GRCh38
rs757676938 Chr.1: 108696013-108696013 on Build GRCh38
rs201667827 Chr.1: 108695710-108695710 on Build GRCh38
rs1031669817 Chr.1: 108695884-108695884 on Build GRCh38
rs754544028 Chr.1: 108696035-108696035 on Build GRCh38
rs551761460 Chr.1: 108695986-108695986 on Build GRCh38
rs921122274 Chr.1: 108695706-108695706 on Build GRCh38
rs771878804 Chr.1: 108696093-108696093 on Build GRCh38
rs776971677 Chr.1: 108695671-108695671 on Build GRCh38
rs150976030 Chr.1: 108695834-108695834 on Build GRCh38
rs200293137 Chr.1: 108695999-108695999 on Build GRCh38
rs765575026 Chr.1: 108695811-108695811 on Build GRCh38
rs748832609 Chr.1: 108695764-108695764 on Build GRCh38
rs774232905 Chr.1: 108695655-108695655 on Build GRCh38
rs995456616 Chr.1: 108695942-108695942 on Build GRCh38
rs544756593 Chr.1: 108695850-108695850 on Build GRCh38
rs1015549154 Chr.1: 108695921-108695921 on Build GRCh38
rs1004207029 Chr.1: 108695918-108695918 on Build GRCh38
rs894734530 Chr.1: 108695798-108695798 on Build GRCh38
rs1028558209 Chr.1: 108695945-108695945 on Build GRCh38
rs761676275 Chr.1: 108695663-108695663 on Build GRCh38
rs745576150 Chr.1: 108695711-108695711 on Build GRCh38
rs774288320 Chr.1: 108695785-108695785 on Build GRCh38

View Full Product Details