Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109483838-109484328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
UniGene
Hs.118248
Gene Chromosome Location
Chr.1: 109483904-109492804 on Build GRCh38
Transcripts
NM_153340.4 XM_005270445.3 XM_005270446.2 XM_011540638.2 XM_011540639.2 XM_011540641.2 XM_011540644.2 XM_011540645.1 XM_011540646.2 XM_017000267.1 XM_017000268.1 XM_017000269.1 XM_017000270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5555108 c.78A>G p.G26G Chr.1: 109484031-109484031 on Build GRCh38
4404970 c.112C>T p.L38L Chr.1: 109484065-109484065 on Build GRCh38
4142106 c.53T>G p.V18G Chr.1: 109484006-109484006 on Build GRCh38
5558620 c.42G>C p.L14L Chr.1: 109483995-109483995 on Build GRCh38
3862118 c.51G>A p.R17R Chr.1: 109484004-109484004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768262182 Chr.1: 109483999-109483999 on Build GRCh38
rs926264516 Chr.1: 109483921-109483921 on Build GRCh38
rs535193596 Chr.1: 109483877-109483877 on Build GRCh38
rs903967468 Chr.1: 109484216-109484216 on Build GRCh38
rs557996994 Chr.1: 109484133-109484133 on Build GRCh38
rs763383569 Chr.1: 109484097-109484097 on Build GRCh38
rs757565388 Chr.1: 109483987-109483987 on Build GRCh38
rs765628680 Chr.1: 109484012-109484012 on Build GRCh38
rs759917945 Chr.1: 109484010-109484010 on Build GRCh38
rs747751536 Chr.1: 109484007-109484007 on Build GRCh38
rs775898963 Chr.1: 109484091-109484091 on Build GRCh38
rs899939359 Chr.1: 109483986-109483986 on Build GRCh38
rs945293725 Chr.1: 109483971-109483971 on Build GRCh38
rs111528173 Chr.1: 109484180-109484180 on Build GRCh38
rs959112488 Chr.1: 109484125-109484125 on Build GRCh38
rs548718185 Chr.1: 109483880-109483880 on Build GRCh38
rs777514291 Chr.1: 109484077-109484077 on Build GRCh38
rs1048884188 Chr.1: 109484004-109484004 on Build GRCh38
rs932765771 Chr.1: 109484274-109484274 on Build GRCh38
rs767532532 Chr.1: 109484126-109484126 on Build GRCh38
rs867358274 Chr.1: 109484099-109484099 on Build GRCh38
rs989569350 Chr.1: 109483940-109483940 on Build GRCh38
rs761579893 Chr.1: 109483942-109483942 on Build GRCh38
rs373780424 Chr.1: 109484215-109484215 on Build GRCh38
rs761039483 Chr.1: 109484130-109484130 on Build GRCh38
rs766644418 Chr.1: 109483946-109483946 on Build GRCh38
rs996765106 Chr.1: 109484002-109484002 on Build GRCh38
rs770128816 Chr.1: 109484090-109484090 on Build GRCh38
rs752429555 Chr.1: 109483908-109483932 on Build GRCh38
rs756063490 Chr.1: 109483992-109483992 on Build GRCh38
rs376553327 Chr.1: 109484072-109484072 on Build GRCh38
rs780168906 Chr.1: 109483994-109483994 on Build GRCh38
rs187088889 Chr.1: 109483952-109483952 on Build GRCh38
rs777120649 Chr.1: 109483949-109483949 on Build GRCh38
rs752727370 Chr.1: 109483976-109483976 on Build GRCh38
rs771576473 Chr.1: 109484073-109484073 on Build GRCh38
rs1036671749 Chr.1: 109483981-109483981 on Build GRCh38
rs767799921 Chr.1: 109484053-109484053 on Build GRCh38
rs776804349 Chr.1: 109484009-109484009 on Build GRCh38
rs981881981 Chr.1: 109483915-109483915 on Build GRCh38
rs966648910 Chr.1: 109484134-109484134 on Build GRCh38
rs1000033955 Chr.1: 109484232-109484232 on Build GRCh38
rs746711126 Chr.1: 109484080-109484080 on Build GRCh38
rs755036907 Chr.1: 109484061-109484061 on Build GRCh38
rs1016212837 Chr.1: 109483899-109483899 on Build GRCh38
rs574019407 Chr.1: 109484275-109484275 on Build GRCh38
rs751658741 Chr.1: 109484045-109484045 on Build GRCh38
rs376463281 Chr.1: 109484022-109484022 on Build GRCh38
rs962467585 Chr.1: 109484307-109484307 on Build GRCh38
rs537776582 Chr.1: 109484239-109484239 on Build GRCh38
rs982938425 Chr.1: 109483868-109483868 on Build GRCh38
rs749281664 Chr.1: 109483995-109483995 on Build GRCh38
rs970463166 Chr.1: 109483900-109483900 on Build GRCh38
rs369600058 Chr.1: 109484056-109484056 on Build GRCh38
rs780047451 Chr.1: 109484058-109484058 on Build GRCh38
rs768386528 Chr.1: 109483932-109483932 on Build GRCh38
rs534900400 Chr.1: 109483953-109483953 on Build GRCh38
rs966976206 Chr.1: 109484209-109484209 on Build GRCh38
rs771760910 Chr.1: 109484008-109484008 on Build GRCh38
rs764281475 Chr.1: 109483982-109483982 on Build GRCh38
rs1012122081 Chr.1: 109484101-109484101 on Build GRCh38
rs573491 Chr.1: 109484269-109484269 on Build GRCh38
rs369274008 Chr.1: 109484237-109484237 on Build GRCh38
rs113663439 Chr.1: 109484205-109484205 on Build GRCh38
rs1011447370 Chr.1: 109483904-109483904 on Build GRCh38
rs761980831 Chr.1: 109484119-109484119 on Build GRCh38
rs755605943 Chr.1: 109483920-109483944 on Build GRCh38
rs1030979865 Chr.1: 109484233-109484233 on Build GRCh38
rs765747540 Chr.1: 109483962-109483962 on Build GRCh38
rs997713868 Chr.1: 109484270-109484270 on Build GRCh38
rs543141158 Chr.1: 109484280-109484280 on Build GRCh38
rs780780207 Chr.1: 109483988-109483988 on Build GRCh38
rs944512029 Chr.1: 109484218-109484218 on Build GRCh38
rs1038848057 Chr.1: 109484221-109484221 on Build GRCh38
rs909383412 Chr.1: 109484288-109484288 on Build GRCh38
rs978435560 Chr.1: 109484213-109484213 on Build GRCh38
rs565890431 Chr.1: 109483890-109483890 on Build GRCh38
rs111520338 Chr.1: 109484202-109484202 on Build GRCh38
rs750201060 Chr.1: 109484055-109484055 on Build GRCh38
rs373236674 Chr.1: 109484065-109484065 on Build GRCh38
rs773847982 Chr.1: 109483941-109483941 on Build GRCh38
rs775952170 Chr.1: 109484016-109484016 on Build GRCh38
rs200167248 Chr.1: 109484006-109484006 on Build GRCh38
rs745686290 Chr.1: 109483989-109483989 on Build GRCh38
rs750654860 Chr.1: 109484128-109484128 on Build GRCh38
rs764076726 Chr.1: 109484043-109484043 on Build GRCh38
rs1056403078 Chr.1: 109484067-109484067 on Build GRCh38
rs758439174 Chr.1: 109483979-109483979 on Build GRCh38
rs937633229 Chr.1: 109483923-109483923 on Build GRCh38
rs757359791 Chr.1: 109484052-109484052 on Build GRCh38
rs769215142 Chr.1: 109484117-109484117 on Build GRCh38
rs751716452 Chr.1: 109483985-109483985 on Build GRCh38
rs774242870 Chr.1: 109484118-109484118 on Build GRCh38
rs115762613 Chr.1: 109484281-109484281 on Build GRCh38
rs559147878 Chr.1: 109483863-109483863 on Build GRCh38

View Full Product Details