Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109509409-109509904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
UniGene
Hs.726479
Gene Chromosome Location
Chr.1: 109504178-109509742 on Build GRCh38
Transcripts
NM_020703.2 XM_011541812.2

SNP Detail

SNP ID Chromosome Location
rs544262917 Chr.1: 109509714-109509714 on Build GRCh38
rs924867013 Chr.1: 109509880-109509880 on Build GRCh38
rs1022220101 Chr.1: 109509617-109509617 on Build GRCh38
rs928494169 Chr.1: 109509495-109509495 on Build GRCh38
rs958891154 Chr.1: 109509428-109509428 on Build GRCh38
rs571195100 Chr.1: 109509453-109509453 on Build GRCh38
rs976419924 Chr.1: 109509865-109509865 on Build GRCh38
rs998603198 Chr.1: 109509855-109509855 on Build GRCh38
rs932467144 Chr.1: 109509689-109509689 on Build GRCh38
rs1040952457 Chr.1: 109509488-109509488 on Build GRCh38
rs1043477991 Chr.1: 109509875-109509875 on Build GRCh38
rs556952652 Chr.1: 109509544-109509544 on Build GRCh38
rs962298668 Chr.1: 109509443-109509443 on Build GRCh38
rs899661558 Chr.1: 109509497-109509497 on Build GRCh38
rs909707427 Chr.1: 109509714-109509715 on Build GRCh38
rs965043933 Chr.1: 109509638-109509638 on Build GRCh38
rs1006724732 Chr.1: 109509529-109509529 on Build GRCh38
rs924249776 Chr.1: 109509635-109509635 on Build GRCh38
rs1036884808 Chr.1: 109509724-109509724 on Build GRCh38
rs960105115 Chr.1: 109509502-109509502 on Build GRCh38
rs1013118927 Chr.1: 109509612-109509612 on Build GRCh38
rs977591928 Chr.1: 109509623-109509623 on Build GRCh38
rs1057386708 Chr.1: 109509798-109509798 on Build GRCh38
rs1021770471 Chr.1: 109509841-109509841 on Build GRCh38
rs2570972 Chr.1: 109509517-109509517 on Build GRCh38
rs775383898 Chr.1: 109509874-109509874 on Build GRCh38
rs991630484 Chr.1: 109509438-109509438 on Build GRCh38
rs1036343226 Chr.1: 109509546-109509546 on Build GRCh38
rs897843714 Chr.1: 109509556-109509556 on Build GRCh38
rs561373291 Chr.1: 109509717-109509717 on Build GRCh38
rs531297869 Chr.1: 109509585-109509585 on Build GRCh38
rs924273801 Chr.1: 109509458-109509458 on Build GRCh38
rs897813219 Chr.1: 109509743-109509743 on Build GRCh38
rs1002916564 Chr.1: 109509770-109509770 on Build GRCh38
rs553560897 Chr.1: 109509622-109509622 on Build GRCh38
rs1035971495 Chr.1: 109509593-109509593 on Build GRCh38
rs548211587 Chr.1: 109509675-109509675 on Build GRCh38
rs976619682 Chr.1: 109509657-109509657 on Build GRCh38
rs958858797 Chr.1: 109509611-109509611 on Build GRCh38
rs546690974 Chr.1: 109509861-109509861 on Build GRCh38
rs573488756 Chr.1: 109509624-109509624 on Build GRCh38
rs971875555 Chr.1: 109509484-109509484 on Build GRCh38
rs1035332724 Chr.1: 109509800-109509800 on Build GRCh38
rs573646611 Chr.1: 109509570-109509570 on Build GRCh38
rs771949634 Chr.1: 109509867-109509867 on Build GRCh38
rs182292044 Chr.1: 109509686-109509686 on Build GRCh38
rs942100004 Chr.1: 109509720-109509720 on Build GRCh38
rs969303889 Chr.1: 109509842-109509842 on Build GRCh38
rs894696215 Chr.1: 109509823-109509823 on Build GRCh38
rs991538667 Chr.1: 109509509-109509509 on Build GRCh38
rs530133605 Chr.1: 109509830-109509830 on Build GRCh38
rs575919799 Chr.1: 109509698-109509698 on Build GRCh38

View Full Product Details