Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58078639-58079148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNB
Entrez Gene ID
2317
Gene Name
filamin B
Gene Aliases
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP
UniGene
Hs.476448
Gene Chromosome Location
Chr.3: 58008400-58172255 on Build GRCh38
Transcripts
NM_001164317.1 NM_001164318.1 NM_001164319.1 NM_001457.3 XM_005264978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3916515 c.581C>T p.P194L Chr.3: 58078756-58078756 on Build GRCh38
6417321 c.583G>A p.V195M Chr.3: 58078758-58078758 on Build GRCh38
5880700 c.548_549insT p.D185fs*13 Chr.3: 58078723-58078724 on Build GRCh38
6434265 c.624G>A p.W208* Chr.3: 58078799-58078799 on Build GRCh38
4654873 c.578A>G p.K193R Chr.3: 58078753-58078753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181769752 Chr.3: 58078752-58078752 on Build GRCh38
rs962901168 Chr.3: 58079085-58079085 on Build GRCh38
rs121908895 Chr.3: 58078779-58078779 on Build GRCh38
rs80356495 Chr.3: 58078717-58078717 on Build GRCh38
rs770152670 Chr.3: 58078675-58078675 on Build GRCh38
rs1022570393 Chr.3: 58078888-58078888 on Build GRCh38
rs781124586 Chr.3: 58078761-58078761 on Build GRCh38
rs891089276 Chr.3: 58079036-58079036 on Build GRCh38
rs768991851 Chr.3: 58078847-58078848 on Build GRCh38
rs761308482 Chr.3: 58078817-58078818 on Build GRCh38
rs984958925 Chr.3: 58079031-58079031 on Build GRCh38
rs745706435 Chr.3: 58078803-58078803 on Build GRCh38
rs997026587 Chr.3: 58078990-58078990 on Build GRCh38
rs919514645 Chr.3: 58078714-58078714 on Build GRCh38
rs769869203 Chr.3: 58079017-58079017 on Build GRCh38
rs775839237 Chr.3: 58078684-58078684 on Build GRCh38
rs934421931 Chr.3: 58078912-58078912 on Build GRCh38
rs780463121 Chr.3: 58078818-58078818 on Build GRCh38
rs139013348 Chr.3: 58078884-58078884 on Build GRCh38
rs558880551 Chr.3: 58078903-58078903 on Build GRCh38
rs1022917510 Chr.3: 58079107-58079107 on Build GRCh38
rs755969281 Chr.3: 58078814-58078814 on Build GRCh38
rs754328623 Chr.3: 58078894-58078894 on Build GRCh38
rs777288549 Chr.3: 58078748-58078748 on Build GRCh38
rs767004566 Chr.3: 58078867-58078867 on Build GRCh38
rs946884715 Chr.3: 58078952-58078952 on Build GRCh38
rs116551325 Chr.3: 58078968-58078968 on Build GRCh38
rs80356497 Chr.3: 58078783-58078783 on Build GRCh38
rs79735974 Chr.3: 58078911-58078911 on Build GRCh38
rs368472521 Chr.3: 58078747-58078747 on Build GRCh38
rs768946230 Chr.3: 58078835-58078835 on Build GRCh38
rs1015846205 Chr.3: 58078790-58078790 on Build GRCh38
rs571381668 Chr.3: 58078858-58078858 on Build GRCh38
rs372192183 Chr.3: 58078865-58078865 on Build GRCh38
rs768025435 Chr.3: 58078696-58078696 on Build GRCh38
rs1050933560 Chr.3: 58078877-58078877 on Build GRCh38
rs757953958 Chr.3: 58078745-58078745 on Build GRCh38
rs80356496 Chr.3: 58078724-58078724 on Build GRCh38
rs540424264 Chr.3: 58078944-58078948 on Build GRCh38
rs955656823 Chr.3: 58078709-58078709 on Build GRCh38
rs766711596 Chr.3: 58078856-58078856 on Build GRCh38
rs529273616 Chr.3: 58078746-58078746 on Build GRCh38
rs566497635 Chr.3: 58078721-58078721 on Build GRCh38
rs199977224 Chr.3: 58078755-58078755 on Build GRCh38
rs763191463 Chr.3: 58078687-58078687 on Build GRCh38
rs80356500 Chr.3: 58078804-58078804 on Build GRCh38
rs200285985 Chr.3: 58078862-58078862 on Build GRCh38
rs768821658 Chr.3: 58078689-58078689 on Build GRCh38
rs774966976 Chr.3: 58078692-58078692 on Build GRCh38
rs548961736 Chr.3: 58078751-58078751 on Build GRCh38
rs761089731 Chr.3: 58078711-58078711 on Build GRCh38
rs1004714290 Chr.3: 58079047-58079047 on Build GRCh38
rs752318884 Chr.3: 58078738-58078738 on Build GRCh38
rs748190199 Chr.3: 58078844-58078844 on Build GRCh38
rs1029732918 Chr.3: 58078690-58078690 on Build GRCh38
rs745611001 Chr.3: 58078668-58078668 on Build GRCh38
rs141550423 Chr.3: 58079058-58079058 on Build GRCh38
rs372274831 Chr.3: 58078841-58078841 on Build GRCh38
rs948390958 Chr.3: 58078879-58078879 on Build GRCh38
rs1043981126 Chr.3: 58078954-58078954 on Build GRCh38
rs886058759 Chr.3: 58078719-58078719 on Build GRCh38
rs374310262 Chr.3: 58078854-58078854 on Build GRCh38
rs909404502 Chr.3: 58079108-58079108 on Build GRCh38
rs751305617 Chr.3: 58078995-58078995 on Build GRCh38
rs763650772 Chr.3: 58078983-58078983 on Build GRCh38
rs973883689 Chr.3: 58079096-58079096 on Build GRCh38
rs1052832329 Chr.3: 58078933-58078933 on Build GRCh38
rs201563731 Chr.3: 58078820-58078820 on Build GRCh38
rs892387866 Chr.3: 58078883-58078883 on Build GRCh38
rs903590745 Chr.3: 58078898-58078898 on Build GRCh38
rs1015670430 Chr.3: 58079064-58079064 on Build GRCh38
rs772515705 Chr.3: 58078853-58078853 on Build GRCh38
rs555741628 Chr.3: 58078942-58078942 on Build GRCh38
rs1026538610 Chr.3: 58079004-58079004 on Build GRCh38
rs762246950 Chr.3: 58078693-58078693 on Build GRCh38
rs80356499 Chr.3: 58078777-58078777 on Build GRCh38
rs187469322 Chr.3: 58078923-58078923 on Build GRCh38
rs750769136 Chr.3: 58078697-58078697 on Build GRCh38
rs777025319 Chr.3: 58078858-58078858 on Build GRCh38

View Full Product Details