Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 63839326-63839822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf49
Entrez Gene ID
132200
Gene Name
chromosome 3 open reading frame 49
Gene Aliases
-
UniGene
Hs.506386
Gene Chromosome Location
Chr.3: 63819365-63848636 on Build GRCh38
Transcripts
NR_026866.1 NM_001285387.2 NM_001285404.1 NM_025075.3 XM_006713339.3

Gene Symbol
THOC7
Entrez Gene ID
80145
Gene Name
THO complex 7
Gene Aliases
NIF3L1BP1, fSAP24, hTREX30
UniGene
Hs.288151
Gene Chromosome Location
Chr.3: 63833870-63864478 on Build GRCh38
Transcripts
NR_026866.1 NM_001285387.2 NM_001285404.1 NM_025075.3 XM_006713339.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
331380 c.20-1G>T p.? Chr.3: 63839774-63839774 on Build GRCh38
5865196 c.37C>T p.R13C Chr.3: 63839756-63839756 on Build GRCh38
6801316 c.62C>T p.A21V Chr.3: 63839731-63839731 on Build GRCh38
4119781 c.126C>T p.S42S Chr.3: 63839667-63839667 on Build GRCh38
4119782 c.66A>C p.G22G Chr.3: 63839727-63839727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758517231 Chr.3: 63839735-63839736 on Build GRCh38
rs922584925 Chr.3: 63839524-63839524 on Build GRCh38
rs768983209 Chr.3: 63839631-63839631 on Build GRCh38
rs767420879 Chr.3: 63839573-63839574 on Build GRCh38
rs770808914 Chr.3: 63839772-63839772 on Build GRCh38
rs896878214 Chr.3: 63839675-63839675 on Build GRCh38
rs762509948 Chr.3: 63839692-63839692 on Build GRCh38
rs372905645 Chr.3: 63839660-63839660 on Build GRCh38
rs577187742 Chr.3: 63839747-63839747 on Build GRCh38
rs757561494 Chr.3: 63839714-63839714 on Build GRCh38
rs750726740 Chr.3: 63839734-63839735 on Build GRCh38
rs941175487 Chr.3: 63839655-63839655 on Build GRCh38
rs567448095 Chr.3: 63839624-63839624 on Build GRCh38
rs964353266 Chr.3: 63839512-63839512 on Build GRCh38
rs138554894 Chr.3: 63839574-63839577 on Build GRCh38
rs750907225 Chr.3: 63839739-63839739 on Build GRCh38
rs970711233 Chr.3: 63839402-63839402 on Build GRCh38
rs2637981 Chr.3: 63839384-63839384 on Build GRCh38
rs1001742076 Chr.3: 63839417-63839417 on Build GRCh38
rs775555606 Chr.3: 63839603-63839603 on Build GRCh38
rs146483827 Chr.3: 63839473-63839473 on Build GRCh38
rs1010682134 Chr.3: 63839536-63839536 on Build GRCh38
rs924763835 Chr.3: 63839646-63839646 on Build GRCh38
rs1047680545 Chr.3: 63839457-63839457 on Build GRCh38
rs748402561 Chr.3: 63839771-63839771 on Build GRCh38
rs868611333 Chr.3: 63839564-63839564 on Build GRCh38
rs760475085 Chr.3: 63839658-63839658 on Build GRCh38
rs551878613 Chr.3: 63839519-63839519 on Build GRCh38
rs536838633 Chr.3: 63839528-63839528 on Build GRCh38
rs763395731 Chr.3: 63839756-63839756 on Build GRCh38
rs1017296544 Chr.3: 63839499-63839499 on Build GRCh38
rs781435774 Chr.3: 63839716-63839716 on Build GRCh38
rs993729122 Chr.3: 63839452-63839452 on Build GRCh38
rs758992844 Chr.3: 63839708-63839708 on Build GRCh38
rs780238172 Chr.3: 63839739-63839740 on Build GRCh38
rs368913290 Chr.3: 63839608-63839608 on Build GRCh38
rs1036756819 Chr.3: 63839674-63839674 on Build GRCh38
rs369363957 Chr.3: 63839630-63839630 on Build GRCh38
rs913289747 Chr.3: 63839482-63839482 on Build GRCh38
rs547802104 Chr.3: 63839435-63839435 on Build GRCh38
rs377307863 Chr.3: 63839690-63839690 on Build GRCh38
rs779216036 Chr.3: 63839762-63839762 on Build GRCh38
rs73831959 Chr.3: 63839359-63839359 on Build GRCh38
rs768856410 Chr.3: 63839757-63839757 on Build GRCh38
rs774779546 Chr.3: 63839648-63839648 on Build GRCh38
rs781613067 Chr.3: 63839605-63839605 on Build GRCh38
rs765257484 Chr.3: 63839677-63839677 on Build GRCh38
rs534656257 Chr.3: 63839676-63839676 on Build GRCh38
rs555664821 Chr.3: 63839587-63839587 on Build GRCh38
rs756550008 Chr.3: 63839745-63839745 on Build GRCh38
rs779186148 Chr.3: 63839614-63839615 on Build GRCh38
rs776582006 Chr.3: 63839670-63839670 on Build GRCh38
rs932552908 Chr.3: 63839530-63839530 on Build GRCh38
rs186313466 Chr.3: 63839496-63839496 on Build GRCh38
rs569849065 Chr.3: 63839523-63839523 on Build GRCh38
rs552560637 Chr.3: 63839700-63839700 on Build GRCh38

View Full Product Details